Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113799254-113799509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255B
Entrez Gene ID
348013
Gene Name
transmembrane protein 255B
Gene Aliases
FAM70B
UniGene
Hs.280805
Gene Chromosome Location
Chr.13: 113759217-113813797 on Build GRCh38
Transcripts
NM_182614.2 XM_005272413.3 XM_005272415.2 XM_006719986.2 XM_017020558.1 XM_017020559.1 XM_017020560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320301 c.394G>T p.G132W Chr.13: 113799390-113799390 on Build GRCh38
3467443 c.348G>A p.P116P Chr.13: 113799344-113799344 on Build GRCh38
6049330 c.375G>A p.Q125Q Chr.13: 113799371-113799371 on Build GRCh38
3786146 c.415C>T p.Q139* Chr.13: 113799411-113799411 on Build GRCh38
3399258 c.354C>G p.P118P Chr.13: 113799350-113799350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779457156 Chr.13: 113799341-113799341 on Build GRCh38
rs201315236 Chr.13: 113799454-113799454 on Build GRCh38
rs199891050 Chr.13: 113799411-113799411 on Build GRCh38
rs763921937 Chr.13: 113799447-113799447 on Build GRCh38
rs369395059 Chr.13: 113799387-113799387 on Build GRCh38
rs760447272 Chr.13: 113799445-113799445 on Build GRCh38
rs752170142 Chr.13: 113799466-113799466 on Build GRCh38
rs541950367 Chr.13: 113799344-113799344 on Build GRCh38
rs376979844 Chr.13: 113799356-113799356 on Build GRCh38
rs182532286 Chr.13: 113799342-113799342 on Build GRCh38
rs7400029 Chr.13: 113799443-113799443 on Build GRCh38
rs750485711 Chr.13: 113799462-113799462 on Build GRCh38
rs527648771 Chr.13: 113799290-113799290 on Build GRCh38
rs139828029 Chr.13: 113799415-113799415 on Build GRCh38
rs145570963 Chr.13: 113799408-113799408 on Build GRCh38
rs568450748 Chr.13: 113799347-113799347 on Build GRCh38
rs373271717 Chr.13: 113799369-113799369 on Build GRCh38
rs202149127 Chr.13: 113799421-113799421 on Build GRCh38
rs369572917 Chr.13: 113799480-113799480 on Build GRCh38
rs1043701533 Chr.13: 113799300-113799300 on Build GRCh38
rs760817234 Chr.13: 113799380-113799380 on Build GRCh38
rs750907190 Chr.13: 113799402-113799402 on Build GRCh38
rs759422756 Chr.13: 113799365-113799365 on Build GRCh38
rs552396286 Chr.13: 113799291-113799291 on Build GRCh38
rs186855778 Chr.13: 113799477-113799477 on Build GRCh38
rs1031725608 Chr.13: 113799274-113799274 on Build GRCh38
rs946655566 Chr.13: 113799285-113799285 on Build GRCh38
rs372984958 Chr.13: 113799425-113799425 on Build GRCh38
rs1013145351 Chr.13: 113799431-113799431 on Build GRCh38
rs369542516 Chr.13: 113799359-113799359 on Build GRCh38
rs371905234 Chr.13: 113799464-113799464 on Build GRCh38
rs571962658 Chr.13: 113799405-113799405 on Build GRCh38
rs745758175 Chr.13: 113799433-113799433 on Build GRCh38
rs757345532 Chr.13: 113799332-113799332 on Build GRCh38
rs757333892 Chr.13: 113799389-113799389 on Build GRCh38
rs1025820279 Chr.13: 113799378-113799378 on Build GRCh38
rs755373819 Chr.13: 113799467-113799467 on Build GRCh38
rs141701439 Chr.13: 113799358-113799358 on Build GRCh38
rs755462147 Chr.13: 113799409-113799409 on Build GRCh38
rs141858027 Chr.13: 113799407-113799407 on Build GRCh38
rs541352760 Chr.13: 113799473-113799473 on Build GRCh38
rs1005800972 Chr.13: 113799395-113799395 on Build GRCh38
rs866224825 Chr.13: 113799444-113799444 on Build GRCh38
rs146672834 Chr.13: 113799385-113799385 on Build GRCh38
rs776877573 Chr.13: 113799451-113799451 on Build GRCh38
rs866521528 Chr.13: 113799364-113799364 on Build GRCh38
rs772783823 Chr.13: 113799348-113799348 on Build GRCh38
rs370896078 Chr.13: 113799470-113799470 on Build GRCh38
rs989290750 Chr.13: 113799310-113799310 on Build GRCh38
rs114871504 Chr.13: 113799343-113799343 on Build GRCh38
rs753916940 Chr.13: 113799295-113799295 on Build GRCh38
rs140209642 Chr.13: 113799401-113799401 on Build GRCh38
rs761975395 Chr.13: 113799453-113799453 on Build GRCh38
rs374064264 Chr.13: 113799479-113799479 on Build GRCh38
rs747702367 Chr.13: 113799416-113799417 on Build GRCh38
rs772000759 Chr.13: 113799441-113799441 on Build GRCh38
rs573851513 Chr.13: 113799469-113799469 on Build GRCh38
rs377144010 Chr.13: 113799371-113799371 on Build GRCh38

View Full Product Details