Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9076711-9076982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2M
Entrez Gene ID
2
Gene Name
alpha-2-macroglobulin
Gene Aliases
A2MD, CPAMD5, FWP007, S863-7
UniGene
Hs.212838
Gene Chromosome Location
Chr.12: 9067708-9115962 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Gene Symbol
KLRG1
Entrez Gene ID
10219
Gene Name
killer cell lectin like receptor G1
Gene Aliases
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE
UniGene
Hs.558446
Gene Chromosome Location
Chr.12: 8989079-9255692 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227228 c.3352-4A>C p.? Chr.12: 9076940-9076940 on Build GRCh38
1660323 c.3517G>C p.E1173Q Chr.12: 9076771-9076771 on Build GRCh38
6341174 c.3415G>T p.D1139Y Chr.12: 9076873-9076873 on Build GRCh38
3793031 c.3365G>A p.R1122H Chr.12: 9076923-9076923 on Build GRCh38
944405 c.3417C>A p.D1139E Chr.12: 9076871-9076871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374673182 Chr.12: 9076784-9076784 on Build GRCh38
rs754820133 Chr.12: 9076871-9076871 on Build GRCh38
rs376943047 Chr.12: 9076894-9076894 on Build GRCh38
rs750031856 Chr.12: 9076853-9076853 on Build GRCh38
rs61730087 Chr.12: 9076803-9076803 on Build GRCh38
rs755426499 Chr.12: 9076751-9076751 on Build GRCh38
rs773383299 Chr.12: 9076811-9076811 on Build GRCh38
rs772074669 Chr.12: 9076912-9076912 on Build GRCh38
rs756783772 Chr.12: 9076841-9076841 on Build GRCh38
rs748115332 Chr.12: 9076906-9076906 on Build GRCh38
rs755868906 Chr.12: 9076775-9076775 on Build GRCh38
rs756875905 Chr.12: 9076768-9076768 on Build GRCh38
rs975263560 Chr.12: 9076735-9076735 on Build GRCh38
rs139868527 Chr.12: 9076739-9076739 on Build GRCh38
rs1022108257 Chr.12: 9076818-9076818 on Build GRCh38
rs755711038 Chr.12: 9076856-9076856 on Build GRCh38
rs755680415 Chr.12: 9076792-9076795 on Build GRCh38
rs761083150 Chr.12: 9076786-9076786 on Build GRCh38
rs778775039 Chr.12: 9076896-9076896 on Build GRCh38
rs780939510 Chr.12: 9076769-9076769 on Build GRCh38
rs769128468 Chr.12: 9076805-9076805 on Build GRCh38
rs372050343 Chr.12: 9076832-9076832 on Build GRCh38
rs751082685 Chr.12: 9076767-9076767 on Build GRCh38
rs745534664 Chr.12: 9076770-9076770 on Build GRCh38
rs772162494 Chr.12: 9076810-9076810 on Build GRCh38
rs765910624 Chr.12: 9076765-9076765 on Build GRCh38
rs747179108 Chr.12: 9076924-9076924 on Build GRCh38
rs760813759 Chr.12: 9076813-9076813 on Build GRCh38
rs777730176 Chr.12: 9076923-9076923 on Build GRCh38
rs767403283 Chr.12: 9076789-9076789 on Build GRCh38
rs774316868 Chr.12: 9076802-9076802 on Build GRCh38
rs777608465 Chr.12: 9076890-9076890 on Build GRCh38
rs776853773 Chr.12: 9076831-9076831 on Build GRCh38
rs749009205 Chr.12: 9076859-9076859 on Build GRCh38
rs112722877 Chr.12: 9076938-9076938 on Build GRCh38
rs779723730 Chr.12: 9076785-9076785 on Build GRCh38
rs111718633 Chr.12: 9076937-9076937 on Build GRCh38
rs200419261 Chr.12: 9076794-9076794 on Build GRCh38
rs766679974 Chr.12: 9076815-9076815 on Build GRCh38
rs765815547 Chr.12: 9076837-9076837 on Build GRCh38
rs767137343 Chr.12: 9076851-9076851 on Build GRCh38
rs56280325 Chr.12: 9076921-9076921 on Build GRCh38
rs779825439 Chr.12: 9076857-9076857 on Build GRCh38
rs112499852 Chr.12: 9076754-9076754 on Build GRCh38
rs112122327 Chr.12: 9076755-9076755 on Build GRCh38

View Full Product Details