Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1817698-1817969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1
Entrez Gene ID
2782
Gene Name
G protein subunit beta 1
Gene Aliases
MRD42
UniGene
Hs.430425
Gene Chromosome Location
Chr.1: 1785285-1891117 on Build GRCh38
Transcripts
NM_001282538.1 NM_001282539.1 NM_002074.4 XM_017001059.1 XM_017001060.1 XM_017001061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030331 c.92C>G p.S31C Chr.1: 1817841-1817841 on Build GRCh38
5695257 c.76G>T p.A26S Chr.1: 1817857-1817857 on Build GRCh38
6419908 c.82G>A p.A28T Chr.1: 1817851-1817851 on Build GRCh38
4615474 c.93_94delTC p.Q32fs*46 Chr.1: 1817839-1817840 on Build GRCh38
463447 c.61G>T p.A21S Chr.1: 1817872-1817872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770350233 Chr.1: 1817904-1817904 on Build GRCh38
rs754937233 Chr.1: 1817842-1817842 on Build GRCh38
rs751238233 Chr.1: 1817867-1817867 on Build GRCh38
rs111709693 Chr.1: 1817877-1817877 on Build GRCh38
rs1030163591 Chr.1: 1817920-1817920 on Build GRCh38
rs955205372 Chr.1: 1817739-1817739 on Build GRCh38
rs111425912 Chr.1: 1817836-1817836 on Build GRCh38
rs774516809 Chr.1: 1817807-1817807 on Build GRCh38
rs949549482 Chr.1: 1817735-1817735 on Build GRCh38
rs887410990 Chr.1: 1817796-1817796 on Build GRCh38
rs751367871 Chr.1: 1817796-1817797 on Build GRCh38
rs753758297 Chr.1: 1817820-1817820 on Build GRCh38
rs542685700 Chr.1: 1817930-1817930 on Build GRCh38
rs764997309 Chr.1: 1817845-1817845 on Build GRCh38
rs143149194 Chr.1: 1817785-1817785 on Build GRCh38
rs113522501 Chr.1: 1817876-1817876 on Build GRCh38
rs768960857 Chr.1: 1817926-1817926 on Build GRCh38
rs780752397 Chr.1: 1817872-1817872 on Build GRCh38
rs758197716 Chr.1: 1817848-1817848 on Build GRCh38
rs977205994 Chr.1: 1817746-1817746 on Build GRCh38
rs780359561 Chr.1: 1817914-1817914 on Build GRCh38
rs367950986 Chr.1: 1817932-1817932 on Build GRCh38
rs964585696 Chr.1: 1817838-1817838 on Build GRCh38
rs749685560 Chr.1: 1817922-1817922 on Build GRCh38
rs116823785 Chr.1: 1817763-1817763 on Build GRCh38
rs764306642 Chr.1: 1817795-1817795 on Build GRCh38
rs756871637 Chr.1: 1817871-1817871 on Build GRCh38
rs140193175 Chr.1: 1817846-1817846 on Build GRCh38
rs761904502 Chr.1: 1817808-1817808 on Build GRCh38
rs775911869 Chr.1: 1817787-1817787 on Build GRCh38
rs957244856 Chr.1: 1817764-1817764 on Build GRCh38
rs993227790 Chr.1: 1817768-1817768 on Build GRCh38
rs567282331 Chr.1: 1817731-1817731 on Build GRCh38
rs561358754 Chr.1: 1817936-1817936 on Build GRCh38
rs895335116 Chr.1: 1817734-1817734 on Build GRCh38
rs755109902 Chr.1: 1817942-1817942 on Build GRCh38
rs1046550639 Chr.1: 1817759-1817759 on Build GRCh38
rs763171432 Chr.1: 1817794-1817794 on Build GRCh38
rs369342974 Chr.1: 1817861-1817861 on Build GRCh38
rs373243069 Chr.1: 1817918-1817918 on Build GRCh38
rs1006158856 Chr.1: 1817814-1817814 on Build GRCh38
rs369707527 Chr.1: 1817773-1817773 on Build GRCh38
rs199518971 Chr.1: 1817817-1817817 on Build GRCh38
rs1017496318 Chr.1: 1817828-1817828 on Build GRCh38
rs137888936 Chr.1: 1817873-1817873 on Build GRCh38

View Full Product Details