Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43656410-43656683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2BP
Entrez Gene ID
11077
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
UniGene
Hs.406157
Gene Chromosome Location
Chr.21: 43453987-43659499 on Build GRCh38
Transcripts
NM_007031.1 XM_011529443.2 XM_011529445.2 XM_011529446.2 XM_011529447.2 XM_017028267.1 XM_017028268.1 XM_017028269.1 XM_017028270.1 XM_017028271.1 XM_017028272.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7145234 c.118G>T p.D40Y Chr.21: 43656656-43656656 on Build GRCh38
6161651 c.144G>T p.G48G Chr.21: 43656630-43656630 on Build GRCh38
5366272 c.128C>T p.P43L Chr.21: 43656646-43656646 on Build GRCh38
5808253 c.145G>A p.E49K Chr.21: 43656629-43656629 on Build GRCh38
3551270 c.162C>T p.F54F Chr.21: 43656612-43656612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988682093 Chr.21: 43656445-43656445 on Build GRCh38
rs956906681 Chr.21: 43656438-43656438 on Build GRCh38
rs760742267 Chr.21: 43656617-43656617 on Build GRCh38
rs749274163 Chr.21: 43656567-43656567 on Build GRCh38
rs756859420 Chr.21: 43656568-43656571 on Build GRCh38
rs770483652 Chr.21: 43656591-43656592 on Build GRCh38
rs375853631 Chr.21: 43656572-43656572 on Build GRCh38
rs955415769 Chr.21: 43656510-43656510 on Build GRCh38
rs776812319 Chr.21: 43656620-43656620 on Build GRCh38
rs915497600 Chr.21: 43656451-43656451 on Build GRCh38
rs187478463 Chr.21: 43656469-43656469 on Build GRCh38
rs184584234 Chr.21: 43656585-43656585 on Build GRCh38
rs753195503 Chr.21: 43656630-43656630 on Build GRCh38
rs755676278 Chr.21: 43656651-43656651 on Build GRCh38
rs774403494 Chr.21: 43656580-43656580 on Build GRCh38
rs755946023 Chr.21: 43656552-43656552 on Build GRCh38
rs968263975 Chr.21: 43656486-43656486 on Build GRCh38
rs745503714 Chr.21: 43656545-43656545 on Build GRCh38
rs746391400 Chr.21: 43656573-43656580 on Build GRCh38
rs2838341 Chr.21: 43656562-43656562 on Build GRCh38
rs964508623 Chr.21: 43656543-43656543 on Build GRCh38
rs766950991 Chr.21: 43656647-43656647 on Build GRCh38
rs746458906 Chr.21: 43656566-43656566 on Build GRCh38
rs77621460 Chr.21: 43656532-43656532 on Build GRCh38
rs945099230 Chr.21: 43656563-43656563 on Build GRCh38
rs939048074 Chr.21: 43656577-43656577 on Build GRCh38
rs143769434 Chr.21: 43656648-43656648 on Build GRCh38
rs201717734 Chr.21: 43656602-43656602 on Build GRCh38
rs766796555 Chr.21: 43656618-43656618 on Build GRCh38
rs377019931 Chr.21: 43656634-43656634 on Build GRCh38
rs989306934 Chr.21: 43656559-43656559 on Build GRCh38
rs781737447 Chr.21: 43656568-43656569 on Build GRCh38
rs926944831 Chr.21: 43656496-43656496 on Build GRCh38
rs765771884 Chr.21: 43656623-43656623 on Build GRCh38
rs773582015 Chr.21: 43656610-43656610 on Build GRCh38
rs2838340 Chr.21: 43656492-43656492 on Build GRCh38
rs759920469 Chr.21: 43656622-43656622 on Build GRCh38
rs192200328 Chr.21: 43656507-43656507 on Build GRCh38
rs536785262 Chr.21: 43656528-43656528 on Build GRCh38
rs779739346 Chr.21: 43656658-43656658 on Build GRCh38
rs140803211 Chr.21: 43656506-43656506 on Build GRCh38
rs540584101 Chr.21: 43656568-43656568 on Build GRCh38

View Full Product Details