Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34656706-34656979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL11RA
Entrez Gene ID
3590
Gene Name
interleukin 11 receptor subunit alpha
Gene Aliases
CRSDA
UniGene
Hs.591088
Gene Chromosome Location
Chr.9: 34652185-34661902 on Build GRCh38
Transcripts
NM_001142784.2 NR_052010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3657060 c.204C>T p.L68L Chr.9: 34656781-34656781 on Build GRCh38
5800339 c.253G>T p.D85Y Chr.9: 34656830-34656830 on Build GRCh38
7330810 c.307G>T p.G103C Chr.9: 34656884-34656884 on Build GRCh38
1462154 c.231T>C p.H77H Chr.9: 34656808-34656808 on Build GRCh38
4800149 c.329G>C p.G110A Chr.9: 34656906-34656906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372764074 Chr.9: 34656938-34656938 on Build GRCh38
rs774380890 Chr.9: 34656900-34656900 on Build GRCh38
rs757675198 Chr.9: 34656950-34656950 on Build GRCh38
rs767164732 Chr.9: 34656793-34656793 on Build GRCh38
rs761678052 Chr.9: 34656790-34656790 on Build GRCh38
rs746029046 Chr.9: 34656752-34656752 on Build GRCh38
rs754479547 Chr.9: 34656825-34656825 on Build GRCh38
rs569085784 Chr.9: 34656758-34656758 on Build GRCh38
rs751970231 Chr.9: 34656838-34656838 on Build GRCh38
rs775518776 Chr.9: 34656759-34656759 on Build GRCh38
rs139359240 Chr.9: 34656744-34656744 on Build GRCh38
rs768570712 Chr.9: 34656766-34656766 on Build GRCh38
rs762947213 Chr.9: 34656765-34656765 on Build GRCh38
rs373056098 Chr.9: 34656760-34656760 on Build GRCh38
rs768658734 Chr.9: 34656899-34656899 on Build GRCh38
rs189071812 Chr.9: 34656818-34656818 on Build GRCh38
rs757663278 Chr.9: 34656847-34656847 on Build GRCh38
rs781388768 Chr.9: 34656848-34656848 on Build GRCh38
rs996434656 Chr.9: 34656767-34656767 on Build GRCh38
rs11575588 Chr.9: 34656731-34656731 on Build GRCh38
rs753383656 Chr.9: 34656820-34656820 on Build GRCh38
rs940379666 Chr.9: 34656881-34656881 on Build GRCh38
rs925134976 Chr.9: 34656894-34656894 on Build GRCh38
rs770001332 Chr.9: 34656854-34656854 on Build GRCh38
rs749387031 Chr.9: 34656859-34656859 on Build GRCh38
rs369630361 Chr.9: 34656858-34656858 on Build GRCh38
rs776944647 Chr.9: 34656745-34656745 on Build GRCh38
rs150731859 Chr.9: 34656905-34656905 on Build GRCh38
rs568165187 Chr.9: 34656923-34656923 on Build GRCh38
rs201179602 Chr.9: 34656901-34656901 on Build GRCh38
rs936532824 Chr.9: 34656895-34656895 on Build GRCh38
rs751155032 Chr.9: 34656817-34656817 on Build GRCh38
rs1055645332 Chr.9: 34656937-34656937 on Build GRCh38
rs11575589 Chr.9: 34656770-34656770 on Build GRCh38
rs764791779 Chr.9: 34656940-34656940 on Build GRCh38
rs979295919 Chr.9: 34656883-34656883 on Build GRCh38
rs911579661 Chr.9: 34656865-34656865 on Build GRCh38
rs376247869 Chr.9: 34656799-34656799 on Build GRCh38
rs778429139 Chr.9: 34656826-34656826 on Build GRCh38
rs752158662 Chr.9: 34656949-34656949 on Build GRCh38
rs750136207 Chr.9: 34656798-34656798 on Build GRCh38
rs764949466 Chr.9: 34656931-34656931 on Build GRCh38
rs954673482 Chr.9: 34656784-34656784 on Build GRCh38
rs866705580 Chr.9: 34656750-34656750 on Build GRCh38

View Full Product Details