Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72343940-72344213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555059 c.1579G>A p.E527K Chr.15: 72344088-72344088 on Build GRCh38
6317346 c.1527-10C>A p.? Chr.15: 72344150-72344150 on Build GRCh38
7290299 c.1528C>T p.R510* Chr.15: 72344139-72344139 on Build GRCh38
1708357 c.1546C>T p.P516S Chr.15: 72344121-72344121 on Build GRCh38
6231387 c.1586C>T p.T529I Chr.15: 72344081-72344081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997194852 Chr.15: 72344007-72344007 on Build GRCh38
rs369398952 Chr.15: 72344122-72344122 on Build GRCh38
rs769119115 Chr.15: 72344116-72344116 on Build GRCh38
rs536980660 Chr.15: 72344172-72344172 on Build GRCh38
rs763935887 Chr.15: 72344048-72344048 on Build GRCh38
rs991947603 Chr.15: 72344169-72344169 on Build GRCh38
rs761249905 Chr.15: 72344134-72344134 on Build GRCh38
rs759741558 Chr.15: 72343983-72343983 on Build GRCh38
rs751432099 Chr.15: 72343997-72343998 on Build GRCh38
rs750495717 Chr.15: 72344189-72344189 on Build GRCh38
rs1027298795 Chr.15: 72344176-72344176 on Build GRCh38
rs748656870 Chr.15: 72344121-72344121 on Build GRCh38
rs1054496232 Chr.15: 72343981-72343981 on Build GRCh38
rs762643178 Chr.15: 72344155-72344155 on Build GRCh38
rs1018731070 Chr.15: 72344123-72344123 on Build GRCh38
rs776140080 Chr.15: 72344136-72344136 on Build GRCh38
rs751574516 Chr.15: 72344058-72344058 on Build GRCh38
rs745432499 Chr.15: 72344138-72344138 on Build GRCh38
rs750636737 Chr.15: 72344064-72344064 on Build GRCh38
rs764962743 Chr.15: 72344032-72344032 on Build GRCh38
rs752413673 Chr.15: 72344035-72344035 on Build GRCh38
rs761877599 Chr.15: 72344184-72344184 on Build GRCh38
rs780359821 Chr.15: 72344076-72344076 on Build GRCh38
rs759216964 Chr.15: 72344028-72344028 on Build GRCh38
rs762847297 Chr.15: 72344037-72344037 on Build GRCh38
rs181233715 Chr.15: 72343998-72343998 on Build GRCh38
rs532797279 Chr.15: 72344065-72344065 on Build GRCh38
rs749609063 Chr.15: 72344113-72344113 on Build GRCh38
rs779228289 Chr.15: 72344117-72344117 on Build GRCh38
rs775281927 Chr.15: 72344150-72344150 on Build GRCh38
rs887509818 Chr.15: 72343982-72343982 on Build GRCh38
rs531838039 Chr.15: 72343996-72343996 on Build GRCh38
rs781430079 Chr.15: 72344060-72344060 on Build GRCh38
rs757357289 Chr.15: 72344059-72344059 on Build GRCh38
rs558918292 Chr.15: 72344001-72344001 on Build GRCh38
rs786204585 Chr.15: 72344139-72344139 on Build GRCh38
rs767660362 Chr.15: 72344187-72344187 on Build GRCh38
rs199914308 Chr.15: 72344146-72344146 on Build GRCh38
rs763965658 Chr.15: 72344164-72344164 on Build GRCh38
rs561029456 Chr.15: 72343966-72343966 on Build GRCh38
rs999012777 Chr.15: 72344057-72344057 on Build GRCh38
rs934899848 Chr.15: 72343970-72343970 on Build GRCh38
rs1030326678 Chr.15: 72344018-72344018 on Build GRCh38
rs866985944 Chr.15: 72343997-72343997 on Build GRCh38
rs774091540 Chr.15: 72344167-72344167 on Build GRCh38

View Full Product Details