Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8324540-8324785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK3
Entrez Gene ID
256949
Gene Name
KN motif and ankyrin repeat domains 3
Gene Aliases
ANKRD47
UniGene
Hs.745036
Gene Chromosome Location
Chr.19: 8322584-8343262 on Build GRCh38
Transcripts
NM_198471.2 XM_006722718.3 XM_011527884.2 XM_017026563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4083020 c.2276T>C p.L759P Chr.19: 8324637-8324637 on Build GRCh38
5365183 c.2222G>A p.R741H Chr.19: 8324691-8324691 on Build GRCh38
2730214 c.2181G>A p.A727A Chr.19: 8324732-8324732 on Build GRCh38
3971385 c.2195C>T p.A732V Chr.19: 8324718-8324718 on Build GRCh38
6690024 c.2246T>C p.L749P Chr.19: 8324667-8324667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748238931 Chr.19: 8324701-8324701 on Build GRCh38
rs200662076 Chr.19: 8324691-8324691 on Build GRCh38
rs145099833 Chr.19: 8324681-8324681 on Build GRCh38
rs61747967 Chr.19: 8324699-8324699 on Build GRCh38
rs752136296 Chr.19: 8324654-8324654 on Build GRCh38
rs756906983 Chr.19: 8324731-8324731 on Build GRCh38
rs771155846 Chr.19: 8324709-8324709 on Build GRCh38
rs528592054 Chr.19: 8324695-8324695 on Build GRCh38
rs543226825 Chr.19: 8324671-8324671 on Build GRCh38
rs765621259 Chr.19: 8324623-8324623 on Build GRCh38
rs149231331 Chr.19: 8324680-8324680 on Build GRCh38
rs368739546 Chr.19: 8324729-8324729 on Build GRCh38
rs748245756 Chr.19: 8324595-8324595 on Build GRCh38
rs749148570 Chr.19: 8324596-8324596 on Build GRCh38
rs778075778 Chr.19: 8324749-8324751 on Build GRCh38
rs1013364741 Chr.19: 8324666-8324666 on Build GRCh38
rs747628913 Chr.19: 8324593-8324593 on Build GRCh38
rs377532244 Chr.19: 8324715-8324715 on Build GRCh38
rs201161749 Chr.19: 8324741-8324741 on Build GRCh38
rs759934875 Chr.19: 8324718-8324718 on Build GRCh38
rs771120307 Chr.19: 8324762-8324762 on Build GRCh38
rs376598437 Chr.19: 8324617-8324617 on Build GRCh38
rs746731911 Chr.19: 8324743-8324743 on Build GRCh38
rs773588012 Chr.19: 8324702-8324702 on Build GRCh38
rs757052567 Chr.19: 8324756-8324756 on Build GRCh38
rs772364975 Chr.19: 8324765-8324765 on Build GRCh38
rs772012421 Chr.19: 8324619-8324619 on Build GRCh38
rs369325105 Chr.19: 8324594-8324594 on Build GRCh38
rs780642731 Chr.19: 8324592-8324592 on Build GRCh38
rs778900283 Chr.19: 8324565-8324568 on Build GRCh38
rs763042740 Chr.19: 8324703-8324703 on Build GRCh38
rs766599918 Chr.19: 8324649-8324649 on Build GRCh38
rs1019757 Chr.19: 8324636-8324636 on Build GRCh38
rs752268165 Chr.19: 8324618-8324618 on Build GRCh38
rs564028402 Chr.19: 8324610-8324610 on Build GRCh38
rs753136339 Chr.19: 8324722-8324722 on Build GRCh38
rs749369061 Chr.19: 8324754-8324754 on Build GRCh38
rs373370477 Chr.19: 8324690-8324690 on Build GRCh38
rs745651938 Chr.19: 8324692-8324692 on Build GRCh38
rs866128544 Chr.19: 8324635-8324635 on Build GRCh38
rs760191859 Chr.19: 8324723-8324723 on Build GRCh38
rs181309788 Chr.19: 8324621-8324621 on Build GRCh38
rs757098841 Chr.19: 8324684-8324684 on Build GRCh38
rs1001677914 Chr.19: 8324600-8324600 on Build GRCh38
rs201462799 Chr.19: 8324608-8324608 on Build GRCh38
rs145402858 Chr.19: 8324677-8324677 on Build GRCh38
rs891595781 Chr.19: 8324720-8324720 on Build GRCh38
rs199751780 Chr.19: 8324717-8324717 on Build GRCh38
rs753560105 Chr.19: 8324730-8324730 on Build GRCh38
rs369599827 Chr.19: 8324620-8324620 on Build GRCh38
rs758708258 Chr.19: 8324657-8324658 on Build GRCh38
rs200719099 Chr.19: 8324733-8324733 on Build GRCh38
rs201470304 Chr.19: 8324732-8324732 on Build GRCh38
rs567210074 Chr.19: 8324742-8324742 on Build GRCh38
rs952325653 Chr.19: 8324763-8324763 on Build GRCh38
rs142165107 Chr.19: 8324752-8324752 on Build GRCh38
rs369659674 Chr.19: 8324744-8324744 on Build GRCh38
rs755480025 Chr.19: 8324668-8324668 on Build GRCh38
rs370788910 Chr.19: 8324721-8324721 on Build GRCh38
rs774360351 Chr.19: 8324613-8324613 on Build GRCh38
rs373095585 Chr.19: 8324595-8324595 on Build GRCh38
rs372748157 Chr.19: 8324569-8324569 on Build GRCh38
rs763385894 Chr.19: 8324639-8324639 on Build GRCh38
rs1048407015 Chr.19: 8324759-8324759 on Build GRCh38
rs752265643 Chr.19: 8324676-8324676 on Build GRCh38
rs759828291 Chr.19: 8324764-8324764 on Build GRCh38

View Full Product Details