Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52801212-52801471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403591 c.763G>A p.E255K Chr.5: 52801445-52801445 on Build GRCh38
5130029 c.734_736delCCT p.S247delS Chr.5: 52801416-52801418 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964981464 Chr.5: 52801318-52801318 on Build GRCh38
rs746603199 Chr.5: 52801448-52801448 on Build GRCh38
rs371879238 Chr.5: 52801377-52801377 on Build GRCh38
rs113870011 Chr.5: 52801321-52801321 on Build GRCh38
rs764010359 Chr.5: 52801367-52801367 on Build GRCh38
rs1054158932 Chr.5: 52801255-52801255 on Build GRCh38
rs772915083 Chr.5: 52801428-52801429 on Build GRCh38
rs927508067 Chr.5: 52801449-52801449 on Build GRCh38
rs375508745 Chr.5: 52801368-52801368 on Build GRCh38
rs199703397 Chr.5: 52801371-52801371 on Build GRCh38
rs112573303 Chr.5: 52801277-52801277 on Build GRCh38
rs145721878 Chr.5: 52801420-52801420 on Build GRCh38
rs775449436 Chr.5: 52801359-52801359 on Build GRCh38
rs369963149 Chr.5: 52801408-52801408 on Build GRCh38
rs983095941 Chr.5: 52801376-52801376 on Build GRCh38
rs755238457 Chr.5: 52801440-52801440 on Build GRCh38
rs756957091 Chr.5: 52801450-52801450 on Build GRCh38
rs369308865 Chr.5: 52801393-52801393 on Build GRCh38
rs1025645893 Chr.5: 52801388-52801388 on Build GRCh38
rs756301643 Chr.5: 52801415-52801415 on Build GRCh38
rs990374199 Chr.5: 52801451-52801451 on Build GRCh38
rs202151315 Chr.5: 52801425-52801425 on Build GRCh38
rs2059202 Chr.5: 52801349-52801349 on Build GRCh38
rs779341584 Chr.5: 52801441-52801441 on Build GRCh38
rs762735875 Chr.5: 52801362-52801362 on Build GRCh38
rs186460209 Chr.5: 52801311-52801311 on Build GRCh38
rs375504463 Chr.5: 52801380-52801380 on Build GRCh38

View Full Product Details