Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41294295-41294526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938877 c.1307C>A p.T436K Chr.19: 41294378-41294378 on Build GRCh38
4938878 c.965C>A p.T322K Chr.19: 41294378-41294378 on Build GRCh38
6319159 c.941G>T p.G314V Chr.19: 41294354-41294354 on Build GRCh38
3990054 c.947C>G p.P316R Chr.19: 41294360-41294360 on Build GRCh38
6681472 c.941G>A p.G314D Chr.19: 41294354-41294354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535606398 Chr.19: 41294316-41294316 on Build GRCh38
rs961927036 Chr.19: 41294340-41294340 on Build GRCh38
rs749838914 Chr.19: 41294499-41294499 on Build GRCh38
rs371503869 Chr.19: 41294466-41294466 on Build GRCh38
rs781105837 Chr.19: 41294428-41294428 on Build GRCh38
rs751344376 Chr.19: 41294407-41294407 on Build GRCh38
rs780146476 Chr.19: 41294397-41294397 on Build GRCh38
rs374735263 Chr.19: 41294467-41294467 on Build GRCh38
rs774827461 Chr.19: 41294476-41294476 on Build GRCh38
rs540785702 Chr.19: 41294483-41294483 on Build GRCh38
rs748283525 Chr.19: 41294439-41294439 on Build GRCh38
rs376862674 Chr.19: 41294334-41294334 on Build GRCh38
rs764631374 Chr.19: 41294352-41294352 on Build GRCh38
rs775973037 Chr.19: 41294488-41294488 on Build GRCh38
rs772395072 Chr.19: 41294484-41294484 on Build GRCh38
rs1020008976 Chr.19: 41294475-41294475 on Build GRCh38
rs771359373 Chr.19: 41294473-41294473 on Build GRCh38
rs866591872 Chr.19: 41294458-41294458 on Build GRCh38
rs375451305 Chr.19: 41294492-41294492 on Build GRCh38
rs758375580 Chr.19: 41294386-41294386 on Build GRCh38
rs753429835 Chr.19: 41294321-41294321 on Build GRCh38
rs749974827 Chr.19: 41294358-41294358 on Build GRCh38
rs988908623 Chr.19: 41294403-41294403 on Build GRCh38
rs754804563 Chr.19: 41294415-41294415 on Build GRCh38

View Full Product Details