Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82194538-82194805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IBTK
Entrez Gene ID
25998
Gene Name
inhibitor of Bruton tyrosine kinase
Gene Aliases
BTBD26, BTKI
UniGene
Hs.306425
Gene Chromosome Location
Chr.6: 82169987-82248294 on Build GRCh38
Transcripts
NM_001300906.1 NM_015525.3 XM_006715453.2 XM_006715454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735487 c.3176C>T p.A1059V Chr.6: 82194641-82194641 on Build GRCh38
4758237 c.3206C>T p.S1069F Chr.6: 82194611-82194611 on Build GRCh38
3176339 c.3199G>C p.G1067R Chr.6: 82194618-82194618 on Build GRCh38
5676271 c.3200G>T p.G1067V Chr.6: 82194617-82194617 on Build GRCh38
4880507 c.3201T>A p.G1067G Chr.6: 82194616-82194616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757637803 Chr.6: 82194726-82194726 on Build GRCh38
rs754960032 Chr.6: 82194662-82194662 on Build GRCh38
rs776329788 Chr.6: 82194661-82194662 on Build GRCh38
rs774786142 Chr.6: 82194623-82194623 on Build GRCh38
rs777093081 Chr.6: 82194688-82194688 on Build GRCh38
rs372782350 Chr.6: 82194591-82194591 on Build GRCh38
rs1057209253 Chr.6: 82194725-82194725 on Build GRCh38
rs770879612 Chr.6: 82194652-82194653 on Build GRCh38
rs562764078 Chr.6: 82194701-82194701 on Build GRCh38
rs755675758 Chr.6: 82194680-82194680 on Build GRCh38
rs772951166 Chr.6: 82194597-82194597 on Build GRCh38
rs766230588 Chr.6: 82194660-82194660 on Build GRCh38
rs1044153518 Chr.6: 82194777-82194777 on Build GRCh38
rs754112848 Chr.6: 82194640-82194640 on Build GRCh38
rs531814084 Chr.6: 82194705-82194705 on Build GRCh38
rs760775848 Chr.6: 82194631-82194631 on Build GRCh38
rs754905839 Chr.6: 82194569-82194569 on Build GRCh38
rs1029232725 Chr.6: 82194773-82194773 on Build GRCh38
rs12662902 Chr.6: 82194624-82194624 on Build GRCh38
rs781079796 Chr.6: 82194668-82194668 on Build GRCh38
rs894335114 Chr.6: 82194642-82194642 on Build GRCh38
rs760184353 Chr.6: 82194628-82194628 on Build GRCh38
rs777798248 Chr.6: 82194627-82194634 on Build GRCh38
rs771343874 Chr.6: 82194619-82194619 on Build GRCh38
rs918858680 Chr.6: 82194711-82194711 on Build GRCh38
rs748971137 Chr.6: 82194693-82194693 on Build GRCh38
rs191661846 Chr.6: 82194641-82194641 on Build GRCh38
rs1029632243 Chr.6: 82194616-82194616 on Build GRCh38
rs746872926 Chr.6: 82194645-82194645 on Build GRCh38
rs780740496 Chr.6: 82194580-82194580 on Build GRCh38
rs745656942 Chr.6: 82194676-82194676 on Build GRCh38
rs751342784 Chr.6: 82194661-82194661 on Build GRCh38
rs939589680 Chr.6: 82194721-82194721 on Build GRCh38
rs554143374 Chr.6: 82194626-82194626 on Build GRCh38
rs752538735 Chr.6: 82194677-82194677 on Build GRCh38
rs747673396 Chr.6: 82194586-82194586 on Build GRCh38
rs748847598 Chr.6: 82194606-82194606 on Build GRCh38
rs948387873 Chr.6: 82194753-82194753 on Build GRCh38
rs764038171 Chr.6: 82194638-82194638 on Build GRCh38
rs548464161 Chr.6: 82194768-82194768 on Build GRCh38
rs78177295 Chr.6: 82194629-82194629 on Build GRCh38

View Full Product Details