Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32014077-32014318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386082 c.248C>T p.P83L Chr.1: 32014243-32014243 on Build GRCh38
6371072 c.197T>C p.L66P Chr.1: 32014192-32014192 on Build GRCh38
6540277 c.262C>T p.P88S Chr.1: 32014257-32014257 on Build GRCh38
4511497 c.86C>T p.S29L Chr.1: 32014081-32014081 on Build GRCh38
3667522 c.207C>T p.P69P Chr.1: 32014202-32014202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006416638 Chr.1: 32014161-32014161 on Build GRCh38
rs996023411 Chr.1: 32014124-32014124 on Build GRCh38
rs981028991 Chr.1: 32014240-32014240 on Build GRCh38
rs936680295 Chr.1: 32014261-32014261 on Build GRCh38
rs764132945 Chr.1: 32014239-32014239 on Build GRCh38
rs773057128 Chr.1: 32014202-32014202 on Build GRCh38
rs535621173 Chr.1: 32014260-32014260 on Build GRCh38
rs1040200101 Chr.1: 32014285-32014285 on Build GRCh38
rs201069205 Chr.1: 32014274-32014274 on Build GRCh38
rs375777010 Chr.1: 32014266-32014266 on Build GRCh38
rs373256621 Chr.1: 32014235-32014235 on Build GRCh38
rs867196638 Chr.1: 32014276-32014276 on Build GRCh38
rs551108017 Chr.1: 32014262-32014262 on Build GRCh38
rs1014672870 Chr.1: 32014190-32014190 on Build GRCh38
rs901779668 Chr.1: 32014113-32014113 on Build GRCh38
rs960009019 Chr.1: 32014147-32014147 on Build GRCh38
rs1012535544 Chr.1: 32014159-32014159 on Build GRCh38
rs750743983 Chr.1: 32014264-32014264 on Build GRCh38
rs986040812 Chr.1: 32014145-32014145 on Build GRCh38
rs758697117 Chr.1: 32014273-32014273 on Build GRCh38
rs762667767 Chr.1: 32014204-32014204 on Build GRCh38
rs780943655 Chr.1: 32014097-32014097 on Build GRCh38
rs912637857 Chr.1: 32014250-32014250 on Build GRCh38
rs112917271 Chr.1: 32014130-32014130 on Build GRCh38
rs1029097851 Chr.1: 32014128-32014128 on Build GRCh38
rs918062032 Chr.1: 32014217-32014217 on Build GRCh38
rs755592365 Chr.1: 32014295-32014295 on Build GRCh38
rs752135277 Chr.1: 32014282-32014282 on Build GRCh38
rs747814006 Chr.1: 32014109-32014109 on Build GRCh38
rs1004387150 Chr.1: 32014110-32014110 on Build GRCh38
rs1022548026 Chr.1: 32014166-32014166 on Build GRCh38
rs931565812 Chr.1: 32014298-32014298 on Build GRCh38
rs867892253 Chr.1: 32014252-32014252 on Build GRCh38
rs568532115 Chr.1: 32014247-32014247 on Build GRCh38
rs544360586 Chr.1: 32014189-32014189 on Build GRCh38
rs528464512 Chr.1: 32014228-32014228 on Build GRCh38
rs762086680 Chr.1: 32014251-32014251 on Build GRCh38
rs925306912 Chr.1: 32014249-32014249 on Build GRCh38
rs992335624 Chr.1: 32014215-32014215 on Build GRCh38
rs961750842 Chr.1: 32014200-32014200 on Build GRCh38
rs760696661 Chr.1: 32014236-32014236 on Build GRCh38

View Full Product Details