Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23364572-23364799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZF1
Entrez Gene ID
64412
Gene Name
GDNF inducible zinc finger protein 1
Gene Aliases
ZBTB23, ZNF336
UniGene
Hs.709462
Gene Chromosome Location
Chr.20: 23361585-23373062 on Build GRCh38
Transcripts
NM_001317012.1 NM_001317019.1 NM_022482.4 XM_005260800.2 XM_005260802.3 XM_011529321.2 XM_011529322.1 XM_017028017.1 XM_017028018.1 XM_017028019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239042 c.260T>C p.F87S Chr.20: 23364643-23364643 on Build GRCh38
4917264 c.315G>T p.R105R Chr.20: 23364698-23364698 on Build GRCh38
4945634 c.297A>G p.V99V Chr.20: 23364680-23364680 on Build GRCh38
6270421 c.295G>T p.V99L Chr.20: 23364678-23364678 on Build GRCh38
7260294 c.359A>T p.D120V Chr.20: 23364742-23364742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767776459 Chr.20: 23364665-23364665 on Build GRCh38
rs976318007 Chr.20: 23364633-23364633 on Build GRCh38
rs200516081 Chr.20: 23364740-23364740 on Build GRCh38
rs773053982 Chr.20: 23364757-23364757 on Build GRCh38
rs747704995 Chr.20: 23364705-23364705 on Build GRCh38
rs370627382 Chr.20: 23364695-23364695 on Build GRCh38
rs545992812 Chr.20: 23364610-23364610 on Build GRCh38
rs749264121 Chr.20: 23364697-23364697 on Build GRCh38
rs563699571 Chr.20: 23364600-23364600 on Build GRCh38
rs376361468 Chr.20: 23364698-23364698 on Build GRCh38
rs746210854 Chr.20: 23364768-23364768 on Build GRCh38
rs750206786 Chr.20: 23364668-23364668 on Build GRCh38
rs779791091 Chr.20: 23364693-23364693 on Build GRCh38
rs377379164 Chr.20: 23364644-23364644 on Build GRCh38
rs143111256 Chr.20: 23364663-23364663 on Build GRCh38
rs374755807 Chr.20: 23364696-23364696 on Build GRCh38
rs753311654 Chr.20: 23364641-23364641 on Build GRCh38
rs879374128 Chr.20: 23364655-23364655 on Build GRCh38
rs764171758 Chr.20: 23364650-23364650 on Build GRCh38
rs189316929 Chr.20: 23364760-23364760 on Build GRCh38
rs748548388 Chr.20: 23364767-23364770 on Build GRCh38
rs374275229 Chr.20: 23364632-23364632 on Build GRCh38
rs756005236 Chr.20: 23364677-23364677 on Build GRCh38
rs138988702 Chr.20: 23364606-23364606 on Build GRCh38
rs988499404 Chr.20: 23364765-23364765 on Build GRCh38
rs370629303 Chr.20: 23364629-23364629 on Build GRCh38
rs367955768 Chr.20: 23364701-23364701 on Build GRCh38

View Full Product Details