Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22171101-22171348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA6
Entrez Gene ID
2627
Gene Name
GATA binding protein 6
Gene Aliases
-
UniGene
Hs.514746
Gene Chromosome Location
Chr.18: 22169437-22202528 on Build GRCh38
Transcripts
NM_005257.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388389 c.134C>G p.S45C Chr.18: 22171278-22171278 on Build GRCh38
7244346 c.43G>C p.G15R Chr.18: 22171187-22171187 on Build GRCh38
4632742 c.48C>T p.A16A Chr.18: 22171192-22171192 on Build GRCh38
6229049 c.66C>T p.S22S Chr.18: 22171210-22171210 on Build GRCh38
6669584 c.165C>T p.G55G Chr.18: 22171309-22171309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747871809 Chr.18: 22171166-22171166 on Build GRCh38
rs750899923 Chr.18: 22171326-22171326 on Build GRCh38
rs777486467 Chr.18: 22171246-22171246 on Build GRCh38
rs764489452 Chr.18: 22171201-22171201 on Build GRCh38
rs758992870 Chr.18: 22171255-22171255 on Build GRCh38
rs1030316600 Chr.18: 22171194-22171194 on Build GRCh38
rs377217497 Chr.18: 22171327-22171327 on Build GRCh38
rs767886137 Chr.18: 22171318-22171318 on Build GRCh38
rs924843423 Chr.18: 22171316-22171316 on Build GRCh38
rs755180659 Chr.18: 22171286-22171286 on Build GRCh38
rs139666654 Chr.18: 22171206-22171206 on Build GRCh38
rs757420762 Chr.18: 22171123-22171123 on Build GRCh38
rs758867858 Chr.18: 22171200-22171200 on Build GRCh38
rs762294271 Chr.18: 22171315-22171315 on Build GRCh38
rs771802924 Chr.18: 22171175-22171175 on Build GRCh38
rs993471660 Chr.18: 22171193-22171193 on Build GRCh38
rs746782842 Chr.18: 22171292-22171292 on Build GRCh38
rs770825513 Chr.18: 22171251-22171251 on Build GRCh38
rs746156401 Chr.18: 22171156-22171156 on Build GRCh38
rs750435307 Chr.18: 22171218-22171218 on Build GRCh38
rs781366792 Chr.18: 22171147-22171147 on Build GRCh38
rs770662152 Chr.18: 22171295-22171295 on Build GRCh38
rs762765948 Chr.18: 22171313-22171313 on Build GRCh38
rs756406083 Chr.18: 22171233-22171233 on Build GRCh38
rs776000244 Chr.18: 22171188-22171188 on Build GRCh38
rs764776127 Chr.18: 22171192-22171192 on Build GRCh38
rs1035980210 Chr.18: 22171190-22171190 on Build GRCh38
rs762641247 Chr.18: 22171267-22171267 on Build GRCh38
rs572030089 Chr.18: 22171260-22171260 on Build GRCh38
rs373655784 Chr.18: 22171122-22171122 on Build GRCh38
rs978912698 Chr.18: 22171308-22171308 on Build GRCh38
rs200519946 Chr.18: 22171297-22171297 on Build GRCh38
rs761329606 Chr.18: 22171293-22171293 on Build GRCh38
rs767885101 Chr.18: 22171280-22171280 on Build GRCh38
rs773163815 Chr.18: 22171177-22171177 on Build GRCh38
rs768396712 Chr.18: 22171314-22171314 on Build GRCh38
rs541367007 Chr.18: 22171212-22171212 on Build GRCh38
rs747016928 Chr.18: 22171249-22171249 on Build GRCh38
rs968804836 Chr.18: 22171272-22171272 on Build GRCh38
rs780116515 Chr.18: 22171283-22171283 on Build GRCh38
rs774239468 Chr.18: 22171161-22171161 on Build GRCh38
rs139750927 Chr.18: 22171160-22171160 on Build GRCh38
rs541150137 Chr.18: 22171299-22171299 on Build GRCh38
rs769310811 Chr.18: 22171256-22171256 on Build GRCh38
rs116262672 Chr.18: 22171187-22171187 on Build GRCh38
rs762445989 Chr.18: 22171273-22171273 on Build GRCh38
rs752129361 Chr.18: 22171205-22171205 on Build GRCh38
rs780160108 Chr.18: 22171235-22171235 on Build GRCh38
rs764615856 Chr.18: 22171269-22171269 on Build GRCh38
rs752348718 Chr.18: 22171197-22171197 on Build GRCh38
rs1006472004 Chr.18: 22171144-22171144 on Build GRCh38
rs759274902 Chr.18: 22171189-22171189 on Build GRCh38
rs781485109 Chr.18: 22171216-22171216 on Build GRCh38
rs745748080 Chr.18: 22171302-22171302 on Build GRCh38
rs1005892039 Chr.18: 22171125-22171125 on Build GRCh38
rs749485622 Chr.18: 22171236-22171236 on Build GRCh38
rs766427554 Chr.18: 22171330-22171330 on Build GRCh38
rs776255080 Chr.18: 22171254-22171254 on Build GRCh38
rs756181712 Chr.18: 22171281-22171281 on Build GRCh38
rs771857951 Chr.18: 22171245-22171245 on Build GRCh38
rs754073585 Chr.18: 22171285-22171285 on Build GRCh38
rs559705145 Chr.18: 22171331-22171331 on Build GRCh38
rs777584552 Chr.18: 22171291-22171291 on Build GRCh38
rs769197296 Chr.18: 22171304-22171304 on Build GRCh38
rs36041440 Chr.18: 22171329-22171330 on Build GRCh38

View Full Product Details