Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51004165-51004398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNG1
Entrez Gene ID
3755
Gene Name
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases
K13, KCNG, KV6.1, kH2
UniGene
Hs.118695
Gene Chromosome Location
Chr.20: 51003656-51023176 on Build GRCh38
Transcripts
NM_002237.3 XM_006723785.2 XM_011528800.1 XM_011528801.1 XM_011528802.1 XM_011528803.2 XM_011528804.1 XM_011528805.2 XM_011528806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707485 c.1268C>T p.T423M Chr.20: 51004313-51004313 on Build GRCh38
1565721 c.1247G>T p.W416L Chr.20: 51004334-51004334 on Build GRCh38
347188 c.1222T>A p.F408I Chr.20: 51004359-51004359 on Build GRCh38
7429530 c.1311C>T p.G437G Chr.20: 51004270-51004270 on Build GRCh38
7411278 c.1247G>A p.W416* Chr.20: 51004334-51004334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377479756 Chr.20: 51004196-51004196 on Build GRCh38
rs1054267 Chr.20: 51004304-51004304 on Build GRCh38
rs139519744 Chr.20: 51004210-51004210 on Build GRCh38
rs759794959 Chr.20: 51004224-51004224 on Build GRCh38
rs777199424 Chr.20: 51004371-51004371 on Build GRCh38
rs994612200 Chr.20: 51004262-51004262 on Build GRCh38
rs372529899 Chr.20: 51004280-51004280 on Build GRCh38
rs747818898 Chr.20: 51004341-51004341 on Build GRCh38
rs17857320 Chr.20: 51004242-51004242 on Build GRCh38
rs868859085 Chr.20: 51004361-51004361 on Build GRCh38
rs764148815 Chr.20: 51004264-51004264 on Build GRCh38
rs760814669 Chr.20: 51004249-51004249 on Build GRCh38
rs1054268 Chr.20: 51004246-51004246 on Build GRCh38
rs8192536 Chr.20: 51004231-51004231 on Build GRCh38
rs770129076 Chr.20: 51004187-51004187 on Build GRCh38
rs755634559 Chr.20: 51004348-51004348 on Build GRCh38
rs763609987 Chr.20: 51004199-51004199 on Build GRCh38
rs751377534 Chr.20: 51004312-51004312 on Build GRCh38
rs774900169 Chr.20: 51004215-51004215 on Build GRCh38
rs868244534 Chr.20: 51004238-51004238 on Build GRCh38
rs780897638 Chr.20: 51004321-51004321 on Build GRCh38
rs867655754 Chr.20: 51004373-51004373 on Build GRCh38
rs11554012 Chr.20: 51004364-51004364 on Build GRCh38
rs754530150 Chr.20: 51004318-51004318 on Build GRCh38
rs1019111340 Chr.20: 51004315-51004315 on Build GRCh38
rs375631930 Chr.20: 51004347-51004347 on Build GRCh38
rs766121713 Chr.20: 51004300-51004300 on Build GRCh38
rs186604143 Chr.20: 51004288-51004288 on Build GRCh38
rs150548464 Chr.20: 51004372-51004372 on Build GRCh38

View Full Product Details