Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156624110-156624371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN2
Entrez Gene ID
60484
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
BRAL1
UniGene
Hs.410719
Gene Chromosome Location
Chr.1: 156601504-156626608 on Build GRCh38
Transcripts
NM_021817.2 XM_005245415.2 XM_011509853.2 XM_017002020.1 XM_017002021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6858411 c.398G>T p.G133V Chr.1: 156624119-156624119 on Build GRCh38
6229629 c.440-5C>A p.? Chr.1: 156624346-156624346 on Build GRCh38
1719028 c.451C>T p.P151S Chr.1: 156624362-156624362 on Build GRCh38
897277 c.440-2A>G p.? Chr.1: 156624349-156624349 on Build GRCh38
5789069 c.402C>G p.I134M Chr.1: 156624123-156624123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747400863 Chr.1: 156624130-156624130 on Build GRCh38
rs747311207 Chr.1: 156624212-156624212 on Build GRCh38
rs112755050 Chr.1: 156624237-156624237 on Build GRCh38
rs776749289 Chr.1: 156624346-156624346 on Build GRCh38
rs149573918 Chr.1: 156624136-156624136 on Build GRCh38
rs543932163 Chr.1: 156624232-156624232 on Build GRCh38
rs376296474 Chr.1: 156624327-156624327 on Build GRCh38
rs781729246 Chr.1: 156624304-156624304 on Build GRCh38
rs747800991 Chr.1: 156624306-156624306 on Build GRCh38
rs769375024 Chr.1: 156624317-156624317 on Build GRCh38
rs757486382 Chr.1: 156624179-156624179 on Build GRCh38
rs367871357 Chr.1: 156624299-156624299 on Build GRCh38
rs770830866 Chr.1: 156624331-156624331 on Build GRCh38
rs1029265185 Chr.1: 156624220-156624220 on Build GRCh38
rs750516645 Chr.1: 156624195-156624195 on Build GRCh38
rs762920807 Chr.1: 156624147-156624147 on Build GRCh38
rs926447540 Chr.1: 156624329-156624329 on Build GRCh38
rs755408840 Chr.1: 156624302-156624302 on Build GRCh38
rs758889838 Chr.1: 156624203-156624203 on Build GRCh38
rs887321524 Chr.1: 156624161-156624161 on Build GRCh38
rs374417527 Chr.1: 156624205-156624205 on Build GRCh38
rs955404462 Chr.1: 156624279-156624279 on Build GRCh38
rs533551649 Chr.1: 156624230-156624230 on Build GRCh38
rs780443838 Chr.1: 156624129-156624129 on Build GRCh38
rs774282053 Chr.1: 156624335-156624335 on Build GRCh38
rs369574575 Chr.1: 156624340-156624340 on Build GRCh38
rs765328626 Chr.1: 156624192-156624192 on Build GRCh38
rs759327164 Chr.1: 156624157-156624157 on Build GRCh38
rs1030136819 Chr.1: 156624256-156624256 on Build GRCh38
rs959441276 Chr.1: 156624343-156624343 on Build GRCh38
rs372076575 Chr.1: 156624318-156624318 on Build GRCh38
rs754044721 Chr.1: 156624174-156624174 on Build GRCh38
rs769335710 Chr.1: 156624141-156624141 on Build GRCh38
rs981191927 Chr.1: 156624312-156624312 on Build GRCh38
rs898421912 Chr.1: 156624328-156624328 on Build GRCh38
rs747605463 Chr.1: 156624140-156624140 on Build GRCh38
rs764239515 Chr.1: 156624173-156624173 on Build GRCh38
rs774087986 Chr.1: 156624153-156624153 on Build GRCh38
rs781413994 Chr.1: 156624131-156624131 on Build GRCh38
rs772666969 Chr.1: 156624142-156624142 on Build GRCh38
rs766280263 Chr.1: 156624148-156624148 on Build GRCh38

View Full Product Details