Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193101130-193101397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX2
Entrez Gene ID
51022
Gene Name
glutaredoxin 2
Gene Aliases
CGI-133, GRX2
UniGene
Hs.458283
Gene Chromosome Location
Chr.1: 193096465-193106535 on Build GRCh38
Transcripts
NM_001243399.1 NM_001319291.1 NM_016066.4 NM_197962.2 XM_017001397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6862784 c.161C>G p.T54R Chr.1: 193101163-193101163 on Build GRCh38
2124129 c.166G>A p.A56T Chr.1: 193101161-193101161 on Build GRCh38
4687238 c.180C>T p.I60I Chr.1: 193101144-193101144 on Build GRCh38
2124128 c.168G>A p.A56A Chr.1: 193101159-193101159 on Build GRCh38
4861269 c.167C>A p.P56H Chr.1: 193101157-193101157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976604109 Chr.1: 193101341-193101341 on Build GRCh38
rs371533088 Chr.1: 193101167-193101167 on Build GRCh38
rs537135562 Chr.1: 193101274-193101274 on Build GRCh38
rs535022976 Chr.1: 193101163-193101163 on Build GRCh38
rs776030642 Chr.1: 193101165-193101165 on Build GRCh38
rs762682354 Chr.1: 193101176-193101176 on Build GRCh38
rs1023423625 Chr.1: 193101264-193101264 on Build GRCh38
rs979440400 Chr.1: 193101262-193101262 on Build GRCh38
rs956083592 Chr.1: 193101322-193101322 on Build GRCh38
rs182259915 Chr.1: 193101190-193101190 on Build GRCh38
rs750791522 Chr.1: 193101248-193101248 on Build GRCh38
rs942639056 Chr.1: 193101274-193101274 on Build GRCh38
rs755184994 Chr.1: 193101227-193101227 on Build GRCh38
rs975367230 Chr.1: 193101303-193101303 on Build GRCh38
rs770888932 Chr.1: 193101232-193101232 on Build GRCh38
rs754143505 Chr.1: 193101217-193101217 on Build GRCh38
rs756254930 Chr.1: 193101207-193101207 on Build GRCh38
rs746762033 Chr.1: 193101229-193101229 on Build GRCh38
rs566116135 Chr.1: 193101154-193101154 on Build GRCh38
rs192512448 Chr.1: 193101282-193101282 on Build GRCh38
rs762077841 Chr.1: 193101159-193101159 on Build GRCh38
rs751904576 Chr.1: 193101189-193101189 on Build GRCh38
rs199693744 Chr.1: 193101211-193101211 on Build GRCh38
rs771900567 Chr.1: 193101160-193101160 on Build GRCh38
rs772973929 Chr.1: 193101162-193101162 on Build GRCh38
rs376344623 Chr.1: 193101249-193101249 on Build GRCh38
rs768017253 Chr.1: 193101196-193101196 on Build GRCh38
rs756413752 Chr.1: 193101215-193101215 on Build GRCh38
rs745773638 Chr.1: 193101242-193101242 on Build GRCh38
rs758764158 Chr.1: 193101261-193101261 on Build GRCh38
rs750498194 Chr.1: 193101205-193101205 on Build GRCh38
rs764620170 Chr.1: 193101185-193101185 on Build GRCh38
rs764623308 Chr.1: 193101166-193101166 on Build GRCh38
rs777424988 Chr.1: 193101300-193101300 on Build GRCh38
rs1031766234 Chr.1: 193101305-193101305 on Build GRCh38
rs557149799 Chr.1: 193101275-193101275 on Build GRCh38
rs752483012 Chr.1: 193101181-193101183 on Build GRCh38
rs896862655 Chr.1: 193101184-193101184 on Build GRCh38
rs552858842 Chr.1: 193101327-193101327 on Build GRCh38
rs187643708 Chr.1: 193101281-193101281 on Build GRCh38
rs146558056 Chr.1: 193101257-193101257 on Build GRCh38

View Full Product Details