Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 20670998-20671223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF7
Entrez Gene ID
151449
Gene Name
growth differentiation factor 7
Gene Aliases
BMP12
UniGene
Hs.447688
Gene Chromosome Location
Chr.2: 20666664-20671490 on Build GRCh38
Transcripts
NM_182828.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161235 c.1119C>T p.Y373Y Chr.2: 20671191-20671191 on Build GRCh38
1614211 c.1053C>T p.R351R Chr.2: 20671125-20671125 on Build GRCh38
228304 c.1144T>A p.C382S Chr.2: 20671216-20671216 on Build GRCh38
2714537 c.1134C>A p.C378* Chr.2: 20671206-20671206 on Build GRCh38
2714539 c.1147G>A p.D383N Chr.2: 20671219-20671219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768216472 Chr.2: 20671188-20671188 on Build GRCh38
rs146317060 Chr.2: 20671162-20671162 on Build GRCh38
rs758288030 Chr.2: 20671104-20671104 on Build GRCh38
rs761497794 Chr.2: 20671206-20671206 on Build GRCh38
rs551505619 Chr.2: 20671115-20671115 on Build GRCh38
rs528385339 Chr.2: 20671108-20671108 on Build GRCh38
rs929428292 Chr.2: 20671191-20671191 on Build GRCh38
rs750203935 Chr.2: 20671097-20671097 on Build GRCh38
rs771479381 Chr.2: 20671032-20671032 on Build GRCh38
rs367794645 Chr.2: 20671127-20671127 on Build GRCh38
rs765086458 Chr.2: 20671096-20671096 on Build GRCh38
rs936221550 Chr.2: 20671125-20671125 on Build GRCh38
rs756893365 Chr.2: 20671095-20671095 on Build GRCh38
rs770169112 Chr.2: 20671027-20671027 on Build GRCh38
rs779193439 Chr.2: 20671140-20671140 on Build GRCh38
rs776117428 Chr.2: 20671197-20671197 on Build GRCh38
rs753433038 Chr.2: 20671087-20671087 on Build GRCh38
rs202010435 Chr.2: 20671164-20671164 on Build GRCh38
rs768045674 Chr.2: 20671051-20671051 on Build GRCh38
rs906704049 Chr.2: 20671048-20671048 on Build GRCh38
rs758354252 Chr.2: 20671017-20671017 on Build GRCh38
rs542280550 Chr.2: 20671038-20671038 on Build GRCh38
rs749586960 Chr.2: 20671123-20671123 on Build GRCh38
rs974975582 Chr.2: 20671102-20671102 on Build GRCh38
rs776001362 Chr.2: 20671165-20671165 on Build GRCh38
rs752722308 Chr.2: 20671114-20671114 on Build GRCh38
rs973624243 Chr.2: 20671167-20671167 on Build GRCh38
rs571155374 Chr.2: 20671116-20671116 on Build GRCh38
rs748206775 Chr.2: 20671021-20671021 on Build GRCh38
rs774772874 Chr.2: 20671045-20671045 on Build GRCh38
rs1047896816 Chr.2: 20671198-20671198 on Build GRCh38
rs759999217 Chr.2: 20671046-20671046 on Build GRCh38
rs78872016 Chr.2: 20671060-20671060 on Build GRCh38
rs144349838 Chr.2: 20671182-20671182 on Build GRCh38
rs886089938 Chr.2: 20671025-20671025 on Build GRCh38
rs1033905416 Chr.2: 20671081-20671081 on Build GRCh38
rs773497984 Chr.2: 20671028-20671028 on Build GRCh38
rs369980607 Chr.2: 20671018-20671018 on Build GRCh38
rs763741972 Chr.2: 20671080-20671080 on Build GRCh38
rs760140572 Chr.2: 20671074-20671074 on Build GRCh38

View Full Product Details