Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78465262-78465497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IREB2
Entrez Gene ID
3658
Gene Name
iron responsive element binding protein 2
Gene Aliases
ACO3, IRP2, IRP2AD
UniGene
Hs.436031
Gene Chromosome Location
Chr.15: 78438176-78501456 on Build GRCh38
Transcripts
NM_001320941.1 NM_001320942.1 NM_001320943.1 NM_004136.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
434368 c.306G>A p.M102I Chr.15: 78465284-78465284 on Build GRCh38
701731 c.367G>C p.D123H Chr.15: 78465345-78465345 on Build GRCh38
3741686 c.408A>G p.K136K Chr.15: 78465386-78465386 on Build GRCh38
4057139 c.299C>T p.A100V Chr.15: 78465277-78465277 on Build GRCh38
4757945 c.400T>G p.F134V Chr.15: 78465378-78465378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780324273 Chr.15: 78465299-78465299 on Build GRCh38
rs973645854 Chr.15: 78465447-78465447 on Build GRCh38
rs780842947 Chr.15: 78465367-78465367 on Build GRCh38
rs929654985 Chr.15: 78465465-78465465 on Build GRCh38
rs762507638 Chr.15: 78465375-78465375 on Build GRCh38
rs374515393 Chr.15: 78465341-78465341 on Build GRCh38
rs368758572 Chr.15: 78465291-78465291 on Build GRCh38
rs377751652 Chr.15: 78465423-78465423 on Build GRCh38
rs980958681 Chr.15: 78465379-78465379 on Build GRCh38
rs1019338688 Chr.15: 78465326-78465326 on Build GRCh38
rs768540922 Chr.15: 78465395-78465395 on Build GRCh38
rs146357212 Chr.15: 78465320-78465320 on Build GRCh38
rs371672348 Chr.15: 78465307-78465307 on Build GRCh38
rs755496176 Chr.15: 78465322-78465322 on Build GRCh38
rs919555878 Chr.15: 78465464-78465464 on Build GRCh38
rs868258073 Chr.15: 78465364-78465364 on Build GRCh38
rs965065343 Chr.15: 78465374-78465374 on Build GRCh38
rs750215754 Chr.15: 78465430-78465430 on Build GRCh38
rs774276221 Chr.15: 78465396-78465396 on Build GRCh38
rs371029182 Chr.15: 78465340-78465340 on Build GRCh38
rs570560882 Chr.15: 78465371-78465371 on Build GRCh38
rs187072238 Chr.15: 78465425-78465425 on Build GRCh38
rs761539604 Chr.15: 78465413-78465413 on Build GRCh38
rs772422852 Chr.15: 78465350-78465350 on Build GRCh38
rs547881233 Chr.15: 78465321-78465321 on Build GRCh38
rs373390703 Chr.15: 78465399-78465399 on Build GRCh38
rs778274026 Chr.15: 78465444-78465444 on Build GRCh38
rs745481790 Chr.15: 78465368-78465368 on Build GRCh38
rs756297456 Chr.15: 78465294-78465294 on Build GRCh38
rs377005450 Chr.15: 78465435-78465435 on Build GRCh38

View Full Product Details