Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53509063-53509336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS1
Entrez Gene ID
148979
Gene Name
GLIS family zinc finger 1
Gene Aliases
-
UniGene
Hs.306691
Gene Chromosome Location
Chr.1: 53506233-53739171 on Build GRCh38
Transcripts
NM_147193.2 XM_017000408.1 XM_017000409.1 XM_017000410.1 XM_017000411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369548 c.1573T>A p.F525I Chr.1: 53509252-53509252 on Build GRCh38
5001530 c.1661G>A p.R554Q Chr.1: 53509164-53509164 on Build GRCh38
7369549 c.1642C>A p.H548N Chr.1: 53509183-53509183 on Build GRCh38
6252027 c.1671C>A p.G557G Chr.1: 53509154-53509154 on Build GRCh38
5479309 c.1667A>C p.N556T Chr.1: 53509158-53509158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150825393 Chr.1: 53509230-53509230 on Build GRCh38
rs767901688 Chr.1: 53509238-53509238 on Build GRCh38
rs747978637 Chr.1: 53509202-53509202 on Build GRCh38
rs200322187 Chr.1: 53509164-53509164 on Build GRCh38
rs142921936 Chr.1: 53509165-53509165 on Build GRCh38
rs777833515 Chr.1: 53509296-53509296 on Build GRCh38
rs140697112 Chr.1: 53509211-53509211 on Build GRCh38
rs757043440 Chr.1: 53509306-53509306 on Build GRCh38
rs776950446 Chr.1: 53509213-53509213 on Build GRCh38
rs183328786 Chr.1: 53509241-53509241 on Build GRCh38
rs906325786 Chr.1: 53509098-53509098 on Build GRCh38
rs764642194 Chr.1: 53509190-53509190 on Build GRCh38
rs957879297 Chr.1: 53509231-53509231 on Build GRCh38
rs10749691 Chr.1: 53509314-53509314 on Build GRCh38
rs201181155 Chr.1: 53509240-53509240 on Build GRCh38
rs146608107 Chr.1: 53509180-53509180 on Build GRCh38
rs888611587 Chr.1: 53509175-53509175 on Build GRCh38
rs542743733 Chr.1: 53509192-53509192 on Build GRCh38
rs867294148 Chr.1: 53509222-53509222 on Build GRCh38
rs555101142 Chr.1: 53509090-53509090 on Build GRCh38
rs753266799 Chr.1: 53509104-53509104 on Build GRCh38
rs757889592 Chr.1: 53509284-53509284 on Build GRCh38
rs779746938 Chr.1: 53509137-53509137 on Build GRCh38
rs761426568 Chr.1: 53509296-53509296 on Build GRCh38
rs745364686 Chr.1: 53509101-53509101 on Build GRCh38
rs777036352 Chr.1: 53509172-53509172 on Build GRCh38
rs781143987 Chr.1: 53509311-53509311 on Build GRCh38
rs149886396 Chr.1: 53509144-53509144 on Build GRCh38
rs146824733 Chr.1: 53509199-53509199 on Build GRCh38
rs763525078 Chr.1: 53509187-53509187 on Build GRCh38
rs775625661 Chr.1: 53509220-53509220 on Build GRCh38
rs966583330 Chr.1: 53509217-53509217 on Build GRCh38
rs977880370 Chr.1: 53509080-53509080 on Build GRCh38
rs760607934 Chr.1: 53509168-53509168 on Build GRCh38
rs370061053 Chr.1: 53509170-53509170 on Build GRCh38
rs758646195 Chr.1: 53509105-53509105 on Build GRCh38
rs746773227 Chr.1: 53509210-53509210 on Build GRCh38
rs779656693 Chr.1: 53509194-53509194 on Build GRCh38
rs769485921 Chr.1: 53509119-53509119 on Build GRCh38
rs947954641 Chr.1: 53509256-53509256 on Build GRCh38
rs372691554 Chr.1: 53509174-53509174 on Build GRCh38
rs769700551 Chr.1: 53509218-53509218 on Build GRCh38
rs561127762 Chr.1: 53509234-53509234 on Build GRCh38
rs759933200 Chr.1: 53509219-53509219 on Build GRCh38
rs41302766 Chr.1: 53509209-53509209 on Build GRCh38
rs759618371 Chr.1: 53509242-53509242 on Build GRCh38
rs773158514 Chr.1: 53509166-53509166 on Build GRCh38
rs753957924 Chr.1: 53509085-53509085 on Build GRCh38
rs757657556 Chr.1: 53509193-53509193 on Build GRCh38
rs778200016 Chr.1: 53509110-53509110 on Build GRCh38
rs767952393 Chr.1: 53509113-53509114 on Build GRCh38
rs566746351 Chr.1: 53509312-53509312 on Build GRCh38
rs753230967 Chr.1: 53509255-53509255 on Build GRCh38
rs1005201769 Chr.1: 53509179-53509179 on Build GRCh38
rs752108105 Chr.1: 53509233-53509233 on Build GRCh38
rs373577732 Chr.1: 53509259-53509259 on Build GRCh38
rs997155148 Chr.1: 53509204-53509204 on Build GRCh38
rs200798419 Chr.1: 53509112-53509112 on Build GRCh38
rs982036881 Chr.1: 53509310-53509310 on Build GRCh38
rs778490328 Chr.1: 53509201-53509201 on Build GRCh38
rs778667678 Chr.1: 53509149-53509149 on Build GRCh38
rs199891850 Chr.1: 53509140-53509140 on Build GRCh38
rs866261589 Chr.1: 53509280-53509280 on Build GRCh38
rs763009701 Chr.1: 53509228-53509228 on Build GRCh38
rs949492016 Chr.1: 53509298-53509298 on Build GRCh38
rs375795005 Chr.1: 53509181-53509181 on Build GRCh38
rs112558798 Chr.1: 53509083-53509083 on Build GRCh38
rs369357255 Chr.1: 53509216-53509216 on Build GRCh38
rs540910687 Chr.1: 53509129-53509129 on Build GRCh38
rs746971724 Chr.1: 53509300-53509300 on Build GRCh38
rs755177966 Chr.1: 53509087-53509087 on Build GRCh38
rs935352637 Chr.1: 53509215-53509215 on Build GRCh38

View Full Product Details