Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94045612-94045839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
-
UniGene
Hs.669645
Gene Chromosome Location
Chr.11: 94021212-94114208 on Build GRCh38
Transcripts
NM_001098672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3454702 c.190G>C p.E64Q Chr.11: 94045692-94045692 on Build GRCh38
275452 c.207G>A p.R69R Chr.11: 94045709-94045709 on Build GRCh38
415693 c.312G>A p.L104L Chr.11: 94045814-94045814 on Build GRCh38
6328064 c.281C>T p.P94L Chr.11: 94045783-94045783 on Build GRCh38
3953990 c.267A>G p.I89M Chr.11: 94045769-94045769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762564811 Chr.11: 94045682-94045682 on Build GRCh38
rs368479317 Chr.11: 94045798-94045798 on Build GRCh38
rs766928747 Chr.11: 94045696-94045696 on Build GRCh38
rs753062282 Chr.11: 94045762-94045762 on Build GRCh38
rs779746221 Chr.11: 94045810-94045810 on Build GRCh38
rs1024586682 Chr.11: 94045716-94045716 on Build GRCh38
rs780967618 Chr.11: 94045665-94045665 on Build GRCh38
rs181178133 Chr.11: 94045691-94045691 on Build GRCh38
rs765533033 Chr.11: 94045702-94045702 on Build GRCh38
rs765538086 Chr.11: 94045760-94045760 on Build GRCh38
rs768315885 Chr.11: 94045739-94045739 on Build GRCh38
rs769933661 Chr.11: 94045735-94045735 on Build GRCh38
rs757198049 Chr.11: 94045717-94045717 on Build GRCh38
rs199720861 Chr.11: 94045759-94045759 on Build GRCh38
rs185760944 Chr.11: 94045744-94045744 on Build GRCh38
rs905488680 Chr.11: 94045806-94045806 on Build GRCh38
rs567827686 Chr.11: 94045698-94045698 on Build GRCh38
rs751570193 Chr.11: 94045710-94045710 on Build GRCh38
rs751694796 Chr.11: 94045774-94045774 on Build GRCh38
rs749046975 Chr.11: 94045811-94045811 on Build GRCh38
rs748917040 Chr.11: 94045731-94045731 on Build GRCh38
rs370633871 Chr.11: 94045700-94045700 on Build GRCh38
rs565317106 Chr.11: 94045751-94045751 on Build GRCh38
rs769543857 Chr.11: 94045674-94045674 on Build GRCh38
rs745603580 Chr.11: 94045666-94045666 on Build GRCh38
rs777277535 Chr.11: 94045755-94045755 on Build GRCh38
rs373750549 Chr.11: 94045706-94045706 on Build GRCh38
rs143861250 Chr.11: 94045743-94045743 on Build GRCh38
rs758594442 Chr.11: 94045709-94045709 on Build GRCh38
rs376541115 Chr.11: 94045683-94045683 on Build GRCh38
rs777812350 Chr.11: 94045651-94045651 on Build GRCh38
rs775000201 Chr.11: 94045681-94045681 on Build GRCh38
rs1051130690 Chr.11: 94045799-94045799 on Build GRCh38
rs971910805 Chr.11: 94045767-94045767 on Build GRCh38
rs757323955 Chr.11: 94045789-94045789 on Build GRCh38
rs779806912 Chr.11: 94045730-94045730 on Build GRCh38
rs189753078 Chr.11: 94045662-94045662 on Build GRCh38
rs865890538 Chr.11: 94045708-94045708 on Build GRCh38
rs781147374 Chr.11: 94045790-94045790 on Build GRCh38
rs995169156 Chr.11: 94045816-94045816 on Build GRCh38
rs769446236 Chr.11: 94045726-94045726 on Build GRCh38
rs539757301 Chr.11: 94045768-94045768 on Build GRCh38
rs763087711 Chr.11: 94045763-94045763 on Build GRCh38
rs761297134 Chr.11: 94045692-94045692 on Build GRCh38
rs185301555 Chr.11: 94045655-94045655 on Build GRCh38
rs1018505438 Chr.11: 94045657-94045657 on Build GRCh38
rs745730257 Chr.11: 94045720-94045720 on Build GRCh38

View Full Product Details