Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 11399480-11399737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST1
Entrez Gene ID
9957
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 1
Gene Aliases
3OST, 3OST1
UniGene
Hs.507348
Gene Chromosome Location
Chr.4: 11398364-11434398 on Build GRCh38
Transcripts
NM_005114.2 XM_011513913.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3006917 c.424C>T p.L142L Chr.4: 11399582-11399582 on Build GRCh38
5644328 c.378G>A p.S126S Chr.4: 11399628-11399628 on Build GRCh38
3006916 c.429G>A p.L143L Chr.4: 11399577-11399577 on Build GRCh38
3006915 c.439C>T p.R147* Chr.4: 11399567-11399567 on Build GRCh38
3916986 c.306G>A p.W102* Chr.4: 11399700-11399700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867570414 Chr.4: 11399651-11399651 on Build GRCh38
rs778649153 Chr.4: 11399550-11399550 on Build GRCh38
rs749725679 Chr.4: 11399669-11399669 on Build GRCh38
rs764624970 Chr.4: 11399714-11399714 on Build GRCh38
rs764361372 Chr.4: 11399593-11399593 on Build GRCh38
rs985680970 Chr.4: 11399589-11399589 on Build GRCh38
rs777308770 Chr.4: 11399556-11399556 on Build GRCh38
rs747203335 Chr.4: 11399559-11399559 on Build GRCh38
rs544551937 Chr.4: 11399505-11399505 on Build GRCh38
rs370992760 Chr.4: 11399602-11399602 on Build GRCh38
rs769864503 Chr.4: 11399583-11399583 on Build GRCh38
rs190695013 Chr.4: 11399628-11399628 on Build GRCh38
rs867060972 Chr.4: 11399594-11399594 on Build GRCh38
rs759333668 Chr.4: 11399711-11399711 on Build GRCh38
rs556962591 Chr.4: 11399555-11399555 on Build GRCh38
rs762292381 Chr.4: 11399596-11399596 on Build GRCh38
rs151069090 Chr.4: 11399574-11399574 on Build GRCh38
rs999595859 Chr.4: 11399504-11399504 on Build GRCh38
rs771189478 Chr.4: 11399567-11399567 on Build GRCh38
rs374145151 Chr.4: 11399511-11399511 on Build GRCh38
rs753547060 Chr.4: 11399604-11399604 on Build GRCh38
rs777406089 Chr.4: 11399640-11399640 on Build GRCh38
rs267599978 Chr.4: 11399679-11399679 on Build GRCh38
rs775031273 Chr.4: 11399675-11399675 on Build GRCh38
rs974239157 Chr.4: 11399665-11399665 on Build GRCh38
rs1017583740 Chr.4: 11399584-11399584 on Build GRCh38
rs756922415 Chr.4: 11399716-11399716 on Build GRCh38
rs141543396 Chr.4: 11399717-11399717 on Build GRCh38
rs769159638 Chr.4: 11399673-11399673 on Build GRCh38
rs775803601 Chr.4: 11399585-11399585 on Build GRCh38
rs199950801 Chr.4: 11399586-11399586 on Build GRCh38
rs147865098 Chr.4: 11399667-11399667 on Build GRCh38
rs776413113 Chr.4: 11399708-11399708 on Build GRCh38
rs945185944 Chr.4: 11399684-11399684 on Build GRCh38
rs757071088 Chr.4: 11399647-11399647 on Build GRCh38
rs754567643 Chr.4: 11399549-11399549 on Build GRCh38
rs200959205 Chr.4: 11399557-11399557 on Build GRCh38
rs146387142 Chr.4: 11399661-11399661 on Build GRCh38
rs975454953 Chr.4: 11399629-11399629 on Build GRCh38
rs758403883 Chr.4: 11399633-11399633 on Build GRCh38
rs572962784 Chr.4: 11399600-11399600 on Build GRCh38
rs955068145 Chr.4: 11399645-11399645 on Build GRCh38
rs539367868 Chr.4: 11399580-11399580 on Build GRCh38
rs894531484 Chr.4: 11399510-11399510 on Build GRCh38
rs763572146 Chr.4: 11399592-11399592 on Build GRCh38
rs556539659 Chr.4: 11399561-11399561 on Build GRCh38
rs140972324 Chr.4: 11399615-11399615 on Build GRCh38
rs746103348 Chr.4: 11399658-11399658 on Build GRCh38
rs375593864 Chr.4: 11399694-11399694 on Build GRCh38
rs781322372 Chr.4: 11399654-11399654 on Build GRCh38
rs566542208 Chr.4: 11399547-11399547 on Build GRCh38
rs765967863 Chr.4: 11399523-11399523 on Build GRCh38
rs762202394 Chr.4: 11399687-11399687 on Build GRCh38

View Full Product Details