Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58196160-58196409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPMK
Entrez Gene ID
253430
Gene Name
inositol polyphosphate multikinase
Gene Aliases
-
UniGene
Hs.30280
Gene Chromosome Location
Chr.10: 58191517-58269837 on Build GRCh38
Transcripts
NM_152230.4 XM_011539565.2 XM_017016013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295722 c.1085T>C p.L362P Chr.10: 58196242-58196242 on Build GRCh38
391624 c.986delA p.H329fs*7 Chr.10: 58196341-58196341 on Build GRCh38
3998037 c.1047G>A p.M349I Chr.10: 58196280-58196280 on Build GRCh38
6066276 c.919G>A p.E307K Chr.10: 58196408-58196408 on Build GRCh38
401048 c.1132G>T p.E378* Chr.10: 58196195-58196195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025935796 Chr.10: 58196218-58196218 on Build GRCh38
rs375383787 Chr.10: 58196371-58196371 on Build GRCh38
rs775609595 Chr.10: 58196219-58196219 on Build GRCh38
rs764957765 Chr.10: 58196228-58196228 on Build GRCh38
rs201334665 Chr.10: 58196189-58196189 on Build GRCh38
rs755724483 Chr.10: 58196284-58196284 on Build GRCh38
rs199822749 Chr.10: 58196224-58196224 on Build GRCh38
rs762947078 Chr.10: 58196238-58196238 on Build GRCh38
rs749648025 Chr.10: 58196292-58196292 on Build GRCh38
rs770946028 Chr.10: 58196325-58196325 on Build GRCh38
rs759410890 Chr.10: 58196369-58196369 on Build GRCh38
rs2275443 Chr.10: 58196280-58196280 on Build GRCh38
rs765751052 Chr.10: 58196343-58196343 on Build GRCh38
rs766290279 Chr.10: 58196244-58196244 on Build GRCh38
rs759662679 Chr.10: 58196206-58196206 on Build GRCh38
rs772305007 Chr.10: 58196338-58196338 on Build GRCh38
rs772940182 Chr.10: 58196353-58196353 on Build GRCh38
rs774628464 Chr.10: 58196203-58196203 on Build GRCh38
rs750168956 Chr.10: 58196233-58196233 on Build GRCh38
rs150092014 Chr.10: 58196249-58196249 on Build GRCh38
rs535168447 Chr.10: 58196368-58196368 on Build GRCh38
rs746340310 Chr.10: 58196324-58196324 on Build GRCh38
rs762715336 Chr.10: 58196363-58196363 on Build GRCh38
rs772459420 Chr.10: 58196216-58196216 on Build GRCh38
rs371083086 Chr.10: 58196252-58196252 on Build GRCh38
rs766345094 Chr.10: 58196365-58196365 on Build GRCh38
rs766938208 Chr.10: 58196370-58196370 on Build GRCh38
rs149331771 Chr.10: 58196282-58196282 on Build GRCh38
rs779527924 Chr.10: 58196309-58196309 on Build GRCh38
rs185185082 Chr.10: 58196339-58196339 on Build GRCh38
rs144385789 Chr.10: 58196186-58196186 on Build GRCh38
rs146242677 Chr.10: 58196296-58196296 on Build GRCh38
rs778062653 Chr.10: 58196288-58196288 on Build GRCh38

View Full Product Details