Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37880010-37880283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
UniGene
Hs.449942
Gene Chromosome Location
Chr.1: 37860697-37947094 on Build GRCh38
Transcripts
NM_001297434.1 NM_005540.2 XM_006710621.3 XM_011541392.2 XM_011541393.2 XM_017001206.1 XM_017001207.1 XM_017001208.1 XM_017001209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7091963 c.1709G>A p.G570D Chr.1: 37880157-37880157 on Build GRCh38
2167664 c.1690G>A p.A564T Chr.1: 37880176-37880176 on Build GRCh38
426220 c.1717G>C p.E573Q Chr.1: 37880149-37880149 on Build GRCh38
2167665 c.1689C>T p.A563A Chr.1: 37880177-37880177 on Build GRCh38
3400746 c.1675A>G p.K559E Chr.1: 37880191-37880191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764227212 Chr.1: 37880073-37880073 on Build GRCh38
rs373766336 Chr.1: 37880177-37880177 on Build GRCh38
rs765611992 Chr.1: 37880099-37880099 on Build GRCh38
rs143220912 Chr.1: 37880195-37880195 on Build GRCh38
rs748877627 Chr.1: 37880223-37880223 on Build GRCh38
rs751902255 Chr.1: 37880214-37880214 on Build GRCh38
rs770684608 Chr.1: 37880050-37880050 on Build GRCh38
rs758871943 Chr.1: 37880108-37880108 on Build GRCh38
rs755631150 Chr.1: 37880238-37880238 on Build GRCh38
rs763538557 Chr.1: 37880207-37880207 on Build GRCh38
rs751311651 Chr.1: 37880113-37880113 on Build GRCh38
rs747237998 Chr.1: 37880169-37880169 on Build GRCh38
rs750811924 Chr.1: 37880089-37880095 on Build GRCh38
rs773068622 Chr.1: 37880242-37880242 on Build GRCh38
rs754151170 Chr.1: 37880077-37880077 on Build GRCh38
rs777299247 Chr.1: 37880214-37880214 on Build GRCh38
rs1018530326 Chr.1: 37880082-37880082 on Build GRCh38
rs780542769 Chr.1: 37880109-37880109 on Build GRCh38
rs762397016 Chr.1: 37880093-37880093 on Build GRCh38
rs201915324 Chr.1: 37880222-37880222 on Build GRCh38
rs758941281 Chr.1: 37880097-37880101 on Build GRCh38
rs749203715 Chr.1: 37880157-37880157 on Build GRCh38
rs747549758 Chr.1: 37880139-37880139 on Build GRCh38
rs550008668 Chr.1: 37880057-37880057 on Build GRCh38
rs375882714 Chr.1: 37880166-37880166 on Build GRCh38
rs766837693 Chr.1: 37880147-37880148 on Build GRCh38
rs754670086 Chr.1: 37880120-37880120 on Build GRCh38
rs907265515 Chr.1: 37880122-37880122 on Build GRCh38
rs766914596 Chr.1: 37880208-37880208 on Build GRCh38
rs754688158 Chr.1: 37880221-37880221 on Build GRCh38
rs1010776433 Chr.1: 37880079-37880079 on Build GRCh38
rs1017795943 Chr.1: 37880226-37880226 on Build GRCh38
rs777615820 Chr.1: 37880147-37880147 on Build GRCh38
rs765326925 Chr.1: 37880182-37880182 on Build GRCh38
rs756870065 Chr.1: 37880237-37880237 on Build GRCh38
rs753793706 Chr.1: 37880086-37880086 on Build GRCh38
rs377591947 Chr.1: 37880206-37880206 on Build GRCh38
rs372827762 Chr.1: 37880167-37880167 on Build GRCh38
rs371250267 Chr.1: 37880049-37880049 on Build GRCh38
rs374188779 Chr.1: 37880239-37880239 on Build GRCh38
rs368979685 Chr.1: 37880176-37880176 on Build GRCh38
rs2306427 Chr.1: 37880224-37880224 on Build GRCh38
rs755575012 Chr.1: 37880213-37880214 on Build GRCh38
rs571641776 Chr.1: 37880107-37880107 on Build GRCh38
rs976654140 Chr.1: 37880127-37880127 on Build GRCh38
rs760874649 Chr.1: 37880064-37880064 on Build GRCh38
rs752183819 Chr.1: 37880159-37880159 on Build GRCh38
rs756111839 Chr.1: 37880145-37880145 on Build GRCh38
rs780754308 Chr.1: 37880121-37880121 on Build GRCh38
rs907991560 Chr.1: 37880179-37880179 on Build GRCh38
rs767041963 Chr.1: 37880223-37880223 on Build GRCh38

View Full Product Details