Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29596415-29596682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIK1
Entrez Gene ID
2897
Gene Name
glutamate ionotropic receptor kainate type subunit 1
Gene Aliases
EAA3, EEA3, GLR5, GLUR5, GluK1, gluR-5
UniGene
Hs.664641
Gene Chromosome Location
Chr.21: 29536933-29940052 on Build GRCh38
Transcripts
NM_000830.4 NM_001320616.1 NM_001320618.1 NM_001320621.1 NM_175611.2 XM_005260942.1 XM_005260944.3 XM_006723991.1 NR_033368.1

Gene Symbol
GRIK1-AS2
Entrez Gene ID
100379661
Gene Name
GRIK1 antisense RNA 2
Gene Aliases
C21orf41, NCRNA00258
UniGene
Hs.659368
Gene Chromosome Location
Chr.21: 29596040-29630747 on Build GRCh38
Transcripts
NM_000830.4 NM_001320616.1 NM_001320618.1 NM_001320621.1 NM_175611.2 XM_005260942.1 XM_005260944.3 XM_006723991.1 NR_033368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668517 c.1235A>C p.H412P Chr.21: 29596548-29596548 on Build GRCh38
4914056 c.1220G>T p.G407V Chr.21: 29596563-29596563 on Build GRCh38
5673673 c.1240T>C p.Y414H Chr.21: 29596543-29596543 on Build GRCh38
184900 c.1234T>C p.Y412H Chr.21: 29596543-29596543 on Build GRCh38
1592765 c.71A>C p.N24T Chr.21: 29596422-29596422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73348595 Chr.21: 29596639-29596639 on Build GRCh38
rs201283754 Chr.21: 29596481-29596481 on Build GRCh38
rs765157708 Chr.21: 29596458-29596458 on Build GRCh38
rs372810146 Chr.21: 29596596-29596596 on Build GRCh38
rs769686738 Chr.21: 29596532-29596532 on Build GRCh38
rs761636328 Chr.21: 29596442-29596442 on Build GRCh38
rs757684799 Chr.21: 29596454-29596454 on Build GRCh38
rs748406656 Chr.21: 29596526-29596526 on Build GRCh38
rs768626937 Chr.21: 29596522-29596522 on Build GRCh38
rs918922383 Chr.21: 29596447-29596447 on Build GRCh38
rs781466907 Chr.21: 29596603-29596603 on Build GRCh38
rs752325028 Chr.21: 29596564-29596564 on Build GRCh38
rs754573148 Chr.21: 29596473-29596473 on Build GRCh38
rs371401872 Chr.21: 29596561-29596561 on Build GRCh38
rs774304958 Chr.21: 29596619-29596619 on Build GRCh38
rs770205803 Chr.21: 29596605-29596605 on Build GRCh38
rs762792023 Chr.21: 29596544-29596544 on Build GRCh38
rs773178926 Chr.21: 29596541-29596541 on Build GRCh38
rs752041893 Chr.21: 29596588-29596588 on Build GRCh38
rs565558729 Chr.21: 29596465-29596465 on Build GRCh38
rs554393676 Chr.21: 29596624-29596624 on Build GRCh38
rs759131327 Chr.21: 29596536-29596536 on Build GRCh38
rs777694556 Chr.21: 29596607-29596607 on Build GRCh38
rs946206531 Chr.21: 29596521-29596521 on Build GRCh38
rs781700964 Chr.21: 29596472-29596472 on Build GRCh38
rs374777692 Chr.21: 29596562-29596562 on Build GRCh38
rs774168985 Chr.21: 29596549-29596549 on Build GRCh38
rs200846672 Chr.21: 29596490-29596490 on Build GRCh38
rs555415857 Chr.21: 29596463-29596463 on Build GRCh38
rs780546010 Chr.21: 29596581-29596581 on Build GRCh38
rs759010911 Chr.21: 29596499-29596499 on Build GRCh38
rs746331501 Chr.21: 29596495-29596495 on Build GRCh38
rs746711582 Chr.21: 29596559-29596559 on Build GRCh38
rs773231917 Chr.21: 29596459-29596459 on Build GRCh38
rs770634207 Chr.21: 29596615-29596615 on Build GRCh38
rs749052947 Chr.21: 29596612-29596612 on Build GRCh38
rs760361593 Chr.21: 29596567-29596567 on Build GRCh38
rs779985372 Chr.21: 29596505-29596505 on Build GRCh38
rs372845109 Chr.21: 29596625-29596625 on Build GRCh38
rs554173363 Chr.21: 29596485-29596485 on Build GRCh38
rs3026024 Chr.21: 29596513-29596513 on Build GRCh38
rs116188011 Chr.21: 29596523-29596523 on Build GRCh38
rs770354408 Chr.21: 29596589-29596589 on Build GRCh38
rs374931169 Chr.21: 29596533-29596533 on Build GRCh38
rs973180590 Chr.21: 29596445-29596445 on Build GRCh38
rs751144777 Chr.21: 29596471-29596471 on Build GRCh38
rs758707069 Chr.21: 29596579-29596579 on Build GRCh38
rs368557467 Chr.21: 29596574-29596574 on Build GRCh38
rs928914787 Chr.21: 29596452-29596452 on Build GRCh38
rs367688924 Chr.21: 29596545-29596545 on Build GRCh38
rs200024509 Chr.21: 29596570-29596570 on Build GRCh38
rs748345362 Chr.21: 29596604-29596604 on Build GRCh38
rs773051065 Chr.21: 29596506-29596508 on Build GRCh38
rs918973312 Chr.21: 29596649-29596649 on Build GRCh38
rs374195550 Chr.21: 29596493-29596493 on Build GRCh38

View Full Product Details