Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122998496-122998723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
UniGene
Hs.501289
Gene Chromosome Location
Chr.10: 122990806-123008850 on Build GRCh38
Transcripts
NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2020917 c.15_16insA p.P6fs*29 Chr.10: 122998610-122998611 on Build GRCh38
2020916 c.33C>T p.F11F Chr.10: 122998593-122998593 on Build GRCh38
1346516 c.15delA p.K5fs*8 Chr.10: 122998611-122998611 on Build GRCh38
682934 c.104G>A p.G35E Chr.10: 122998522-122998522 on Build GRCh38
4012366 c.115G>A p.A39T Chr.10: 122998511-122998511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757971262 Chr.10: 122998530-122998530 on Build GRCh38
rs200964765 Chr.10: 122998578-122998578 on Build GRCh38
rs746261510 Chr.10: 122998567-122998567 on Build GRCh38
rs763593826 Chr.10: 122998671-122998671 on Build GRCh38
rs768696087 Chr.10: 122998597-122998597 on Build GRCh38
rs913403838 Chr.10: 122998624-122998624 on Build GRCh38
rs199959505 Chr.10: 122998593-122998593 on Build GRCh38
rs774504855 Chr.10: 122998608-122998608 on Build GRCh38
rs759528056 Chr.10: 122998626-122998626 on Build GRCh38
rs765178014 Chr.10: 122998636-122998636 on Build GRCh38
rs936984388 Chr.10: 122998690-122998690 on Build GRCh38
rs535220324 Chr.10: 122998663-122998663 on Build GRCh38
rs202154688 Chr.10: 122998610-122998610 on Build GRCh38
rs781626712 Chr.10: 122998551-122998551 on Build GRCh38
rs776753682 Chr.10: 122998611-122998611 on Build GRCh38
rs978997089 Chr.10: 122998685-122998685 on Build GRCh38
rs775417868 Chr.10: 122998641-122998641 on Build GRCh38
rs749592041 Chr.10: 122998595-122998595 on Build GRCh38
rs776071988 Chr.10: 122998687-122998687 on Build GRCh38
rs1029063246 Chr.10: 122998592-122998592 on Build GRCh38
rs763723497 Chr.10: 122998533-122998533 on Build GRCh38
rs757794183 Chr.10: 122998547-122998547 on Build GRCh38
rs752442150 Chr.10: 122998529-122998529 on Build GRCh38
rs750965585 Chr.10: 122998539-122998539 on Build GRCh38
rs367823215 Chr.10: 122998647-122998647 on Build GRCh38

View Full Product Details