Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34913116-34913389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS2
Entrez Gene ID
55902
Gene Name
acyl-CoA synthetase short-chain family member 2
Gene Aliases
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1
UniGene
Hs.517034
Gene Chromosome Location
Chr.20: 34874942-34927966 on Build GRCh38
Transcripts
NM_001076552.2 NM_001242393.1 NM_018677.3 XM_005260455.1 XM_005260456.1 XM_006723826.1 XM_011528905.1 XM_011528906.1 XM_011528907.1 XM_011528908.1 XM_011528909.2 XM_011528911.1 XM_011528912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545587 c.458G>A p.R153Q Chr.20: 34913179-34913179 on Build GRCh38
7351466 c.316+1G>A p.? Chr.20: 34913188-34913188 on Build GRCh38
3545585 c.456C>T p.L152L Chr.20: 34913177-34913177 on Build GRCh38
3545583 c.403C>T p.Q135* Chr.20: 34913124-34913124 on Build GRCh38
6592764 c.308G>A p.R103Q Chr.20: 34913179-34913179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777719878 Chr.20: 34913216-34913216 on Build GRCh38
rs1027702834 Chr.20: 34913312-34913312 on Build GRCh38
rs896802660 Chr.20: 34913257-34913257 on Build GRCh38
rs993840506 Chr.20: 34913275-34913275 on Build GRCh38
rs745770950 Chr.20: 34913236-34913236 on Build GRCh38
rs1023876655 Chr.20: 34913351-34913351 on Build GRCh38
rs1009127579 Chr.20: 34913223-34913223 on Build GRCh38
rs763781277 Chr.20: 34913179-34913179 on Build GRCh38
rs369200323 Chr.20: 34913161-34913161 on Build GRCh38
rs960903040 Chr.20: 34913313-34913313 on Build GRCh38
rs183847650 Chr.20: 34913158-34913158 on Build GRCh38
rs765627693 Chr.20: 34913200-34913200 on Build GRCh38
rs1034724904 Chr.20: 34913302-34913302 on Build GRCh38
rs372789849 Chr.20: 34913202-34913202 on Build GRCh38
rs973895642 Chr.20: 34913303-34913303 on Build GRCh38
rs538187001 Chr.20: 34913234-34913234 on Build GRCh38
rs747045202 Chr.20: 34913217-34913217 on Build GRCh38
rs1038140611 Chr.20: 34913254-34913254 on Build GRCh38
rs767072404 Chr.20: 34913316-34913316 on Build GRCh38
rs544619206 Chr.20: 34913277-34913277 on Build GRCh38
rs562270723 Chr.20: 34913170-34913170 on Build GRCh38
rs761178819 Chr.20: 34913182-34913182 on Build GRCh38
rs745607403 Chr.20: 34913150-34913150 on Build GRCh38
rs769544838 Chr.20: 34913362-34913362 on Build GRCh38
rs769692584 Chr.20: 34913156-34913156 on Build GRCh38
rs754226756 Chr.20: 34913188-34913188 on Build GRCh38
rs950788196 Chr.20: 34913323-34913323 on Build GRCh38
rs776598687 Chr.20: 34913142-34913142 on Build GRCh38
rs1018177115 Chr.20: 34913238-34913238 on Build GRCh38
rs879242447 Chr.20: 34913317-34913317 on Build GRCh38
rs757281988 Chr.20: 34913219-34913219 on Build GRCh38
rs199714251 Chr.20: 34913206-34913206 on Build GRCh38
rs766834295 Chr.20: 34913187-34913187 on Build GRCh38
rs761186205 Chr.20: 34913245-34913245 on Build GRCh38
rs530201978 Chr.20: 34913178-34913178 on Build GRCh38

View Full Product Details