Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99391076-99391319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC1B
Entrez Gene ID
10095
Gene Name
actin related protein 2/3 complex subunit 1B
Gene Aliases
ARC41, p40-ARC, p41-ARC
UniGene
Hs.489284
Gene Chromosome Location
Chr.7: 99374675-99394801 on Build GRCh38
Transcripts
NM_005720.3 XM_006715825.1 XM_006715826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998573 c.757A>G p.I253V Chr.7: 99391227-99391227 on Build GRCh38
673969 c.707+3G>C p.? Chr.7: 99391102-99391102 on Build GRCh38
6300240 c.694G>T p.D232Y Chr.7: 99391086-99391086 on Build GRCh38
3883804 c.747G>A p.A249A Chr.7: 99391217-99391217 on Build GRCh38
1093784 c.694G>A p.D232N Chr.7: 99391086-99391086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944457556 Chr.7: 99391241-99391241 on Build GRCh38
rs753242090 Chr.7: 99391143-99391143 on Build GRCh38
rs774520107 Chr.7: 99391133-99391133 on Build GRCh38
rs377169175 Chr.7: 99391175-99391175 on Build GRCh38
rs776629816 Chr.7: 99391106-99391106 on Build GRCh38
rs764805361 Chr.7: 99391218-99391218 on Build GRCh38
rs778558824 Chr.7: 99391161-99391161 on Build GRCh38
rs759673375 Chr.7: 99391139-99391139 on Build GRCh38
rs768768149 Chr.7: 99391178-99391178 on Build GRCh38
rs772222428 Chr.7: 99391293-99391293 on Build GRCh38
rs779597975 Chr.7: 99391253-99391253 on Build GRCh38
rs768495702 Chr.7: 99391105-99391105 on Build GRCh38
rs776539765 Chr.7: 99391179-99391179 on Build GRCh38
rs114065699 Chr.7: 99391209-99391209 on Build GRCh38
rs769705016 Chr.7: 99391282-99391282 on Build GRCh38
rs754153254 Chr.7: 99391216-99391216 on Build GRCh38
rs756864602 Chr.7: 99391158-99391158 on Build GRCh38
rs763249515 Chr.7: 99391111-99391111 on Build GRCh38
rs747975319 Chr.7: 99391181-99391181 on Build GRCh38
rs147238850 Chr.7: 99391182-99391182 on Build GRCh38
rs746717899 Chr.7: 99391099-99391099 on Build GRCh38
rs766803763 Chr.7: 99391264-99391268 on Build GRCh38
rs750089172 Chr.7: 99391222-99391222 on Build GRCh38
rs373141340 Chr.7: 99391163-99391163 on Build GRCh38
rs1022139359 Chr.7: 99391104-99391104 on Build GRCh38
rs746078510 Chr.7: 99391288-99391288 on Build GRCh38
rs757307668 Chr.7: 99391184-99391184 on Build GRCh38
rs760191638 Chr.7: 99391209-99391213 on Build GRCh38
rs758046980 Chr.7: 99391245-99391245 on Build GRCh38
rs761214014 Chr.7: 99391201-99391201 on Build GRCh38
rs527770230 Chr.7: 99391152-99391152 on Build GRCh38
rs751074024 Chr.7: 99391265-99391265 on Build GRCh38
rs781285072 Chr.7: 99391271-99391271 on Build GRCh38
rs906746979 Chr.7: 99391123-99391123 on Build GRCh38
rs749887267 Chr.7: 99391126-99391130 on Build GRCh38
rs550507060 Chr.7: 99391269-99391269 on Build GRCh38
rs1052158523 Chr.7: 99391264-99391264 on Build GRCh38
rs766735622 Chr.7: 99391122-99391122 on Build GRCh38
rs200511413 Chr.7: 99391301-99391301 on Build GRCh38
rs756635642 Chr.7: 99391146-99391146 on Build GRCh38
rs375752684 Chr.7: 99391280-99391280 on Build GRCh38
rs757633382 Chr.7: 99391217-99391217 on Build GRCh38
rs775479028 Chr.7: 99391191-99391191 on Build GRCh38
rs371905376 Chr.7: 99391153-99391153 on Build GRCh38
rs767620205 Chr.7: 99391190-99391190 on Build GRCh38
rs998903467 Chr.7: 99391230-99391230 on Build GRCh38
rs961108790 Chr.7: 99391171-99391171 on Build GRCh38
rs941306421 Chr.7: 99391180-99391180 on Build GRCh38
rs775598851 Chr.7: 99391297-99391297 on Build GRCh38
rs1034846429 Chr.7: 99391151-99391151 on Build GRCh38
rs368603633 Chr.7: 99391275-99391275 on Build GRCh38
rs369966813 Chr.7: 99391183-99391183 on Build GRCh38
rs746956120 Chr.7: 99391176-99391176 on Build GRCh38
rs531693021 Chr.7: 99391232-99391232 on Build GRCh38
rs777485188 Chr.7: 99391287-99391287 on Build GRCh38
rs969211896 Chr.7: 99391112-99391112 on Build GRCh38
rs767842063 Chr.7: 99391141-99391141 on Build GRCh38

View Full Product Details