Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3659485-3659752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART1
Entrez Gene ID
417
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ART2, ARTC1, CD296, RT6
UniGene
Hs.382188
Gene Chromosome Location
Chr.11: 3645110-3665595 on Build GRCh38
Transcripts
NM_004314.2 XM_011520114.2 XM_017017763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231802 c.134C>T p.S45F Chr.11: 3659653-3659653 on Build GRCh38
268711 c.130G>A p.A44T Chr.11: 3659649-3659649 on Build GRCh38
131101 c.149A>G p.Y50C Chr.11: 3659668-3659668 on Build GRCh38
1321694 c.89G>A p.R30Q Chr.11: 3659608-3659608 on Build GRCh38
278993 c.108G>T p.E36D Chr.11: 3659627-3659627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376115062 Chr.11: 3659549-3659549 on Build GRCh38
rs527878673 Chr.11: 3659719-3659719 on Build GRCh38
rs761928695 Chr.11: 3659571-3659571 on Build GRCh38
rs573354342 Chr.11: 3659528-3659528 on Build GRCh38
rs778468551 Chr.11: 3659620-3659620 on Build GRCh38
rs763258503 Chr.11: 3659536-3659536 on Build GRCh38
rs560779596 Chr.11: 3659688-3659688 on Build GRCh38
rs775699699 Chr.11: 3659535-3659535 on Build GRCh38
rs755973140 Chr.11: 3659606-3659606 on Build GRCh38
rs562108762 Chr.11: 3659640-3659640 on Build GRCh38
rs764183700 Chr.11: 3659561-3659561 on Build GRCh38
rs777237646 Chr.11: 3659647-3659647 on Build GRCh38
rs202064026 Chr.11: 3659645-3659645 on Build GRCh38
rs749791342 Chr.11: 3659704-3659704 on Build GRCh38
rs779943962 Chr.11: 3659607-3659607 on Build GRCh38
rs780369791 Chr.11: 3659702-3659702 on Build GRCh38
rs768939051 Chr.11: 3659661-3659661 on Build GRCh38
rs907576007 Chr.11: 3659584-3659584 on Build GRCh38
rs1051671427 Chr.11: 3659700-3659700 on Build GRCh38
rs983031312 Chr.11: 3659525-3659525 on Build GRCh38
rs975386471 Chr.11: 3659665-3659665 on Build GRCh38
rs746466114 Chr.11: 3659690-3659690 on Build GRCh38
rs892941585 Chr.11: 3659703-3659703 on Build GRCh38
rs754686900 Chr.11: 3659611-3659611 on Build GRCh38
rs200793958 Chr.11: 3659632-3659632 on Build GRCh38
rs376176124 Chr.11: 3659720-3659720 on Build GRCh38
rs368360706 Chr.11: 3659670-3659670 on Build GRCh38
rs756652182 Chr.11: 3659692-3659692 on Build GRCh38
rs753669568 Chr.11: 3659673-3659673 on Build GRCh38
rs747787811 Chr.11: 3659635-3659635 on Build GRCh38
rs760941868 Chr.11: 3659666-3659666 on Build GRCh38
rs777286910 Chr.11: 3659689-3659689 on Build GRCh38
rs766186226 Chr.11: 3659672-3659672 on Build GRCh38
rs773457083 Chr.11: 3659721-3659721 on Build GRCh38
rs201412190 Chr.11: 3659568-3659568 on Build GRCh38
rs774822641 Chr.11: 3659717-3659717 on Build GRCh38
rs767742248 Chr.11: 3659669-3659669 on Build GRCh38
rs750450780 Chr.11: 3659605-3659605 on Build GRCh38
rs763163415 Chr.11: 3659654-3659654 on Build GRCh38
rs1020008360 Chr.11: 3659727-3659727 on Build GRCh38
rs990039760 Chr.11: 3659641-3659641 on Build GRCh38
rs563782581 Chr.11: 3659731-3659731 on Build GRCh38
rs769207726 Chr.11: 3659705-3659705 on Build GRCh38
rs199916833 Chr.11: 3659604-3659604 on Build GRCh38
rs779072169 Chr.11: 3659723-3659723 on Build GRCh38
rs543890617 Chr.11: 3659683-3659683 on Build GRCh38
rs139244300 Chr.11: 3659726-3659726 on Build GRCh38
rs765013277 Chr.11: 3659682-3659682 on Build GRCh38
rs754669485 Chr.11: 3659677-3659677 on Build GRCh38
rs35318694 Chr.11: 3659648-3659648 on Build GRCh38
rs372301018 Chr.11: 3659680-3659680 on Build GRCh38
rs775958574 Chr.11: 3659653-3659653 on Build GRCh38

View Full Product Details