Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169674532-169674771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC173
Entrez Gene ID
129881
Gene Name
coiled-coil domain containing 173
Gene Aliases
C2orf77
UniGene
Hs.370111
Gene Chromosome Location
Chr.2: 169645425-169694421 on Build GRCh38
Transcripts
NM_001085447.1 XM_011510589.1 XM_011510590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6206971 c.503G>A p.R168H Chr.2: 169674735-169674735 on Build GRCh38
4667650 c.618delA p.K206fs*14 Chr.2: 169674620-169674620 on Build GRCh38
1009335 c.598G>T p.E200* Chr.2: 169674640-169674640 on Build GRCh38
6010378 c.652C>T p.H218Y Chr.2: 169674586-169674586 on Build GRCh38
5394917 c.559G>A p.E187K Chr.2: 169674679-169674679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763715447 Chr.2: 169674704-169674704 on Build GRCh38
rs375356931 Chr.2: 169674575-169674575 on Build GRCh38
rs928638821 Chr.2: 169674716-169674716 on Build GRCh38
rs750531318 Chr.2: 169674685-169674686 on Build GRCh38
rs199751298 Chr.2: 169674729-169674729 on Build GRCh38
rs369756121 Chr.2: 169674619-169674619 on Build GRCh38
rs200589066 Chr.2: 169674652-169674652 on Build GRCh38
rs749362148 Chr.2: 169674657-169674657 on Build GRCh38
rs765271237 Chr.2: 169674635-169674637 on Build GRCh38
rs1053578874 Chr.2: 169674685-169674685 on Build GRCh38
rs766379493 Chr.2: 169674728-169674728 on Build GRCh38
rs267599001 Chr.2: 169674715-169674715 on Build GRCh38
rs368546077 Chr.2: 169674735-169674735 on Build GRCh38
rs764737812 Chr.2: 169674713-169674713 on Build GRCh38
rs754874295 Chr.2: 169674736-169674736 on Build GRCh38
rs767780209 Chr.2: 169674598-169674598 on Build GRCh38
rs761309902 Chr.2: 169674709-169674709 on Build GRCh38
rs951452516 Chr.2: 169674690-169674690 on Build GRCh38
rs763558909 Chr.2: 169674606-169674606 on Build GRCh38
rs774929204 Chr.2: 169674664-169674664 on Build GRCh38
rs748361850 Chr.2: 169674650-169674650 on Build GRCh38
rs985701209 Chr.2: 169674572-169674572 on Build GRCh38
rs376884574 Chr.2: 169674673-169674673 on Build GRCh38
rs765147831 Chr.2: 169674618-169674618 on Build GRCh38
rs758152092 Chr.2: 169674620-169674620 on Build GRCh38
rs775752655 Chr.2: 169674684-169674684 on Build GRCh38
rs753314829 Chr.2: 169674607-169674607 on Build GRCh38
rs776324974 Chr.2: 169674705-169674705 on Build GRCh38
rs779734131 Chr.2: 169674633-169674633 on Build GRCh38
rs761146574 Chr.2: 169674686-169674686 on Build GRCh38
rs756809578 Chr.2: 169674613-169674613 on Build GRCh38
rs925942525 Chr.2: 169674626-169674626 on Build GRCh38
rs772435536 Chr.2: 169674674-169674674 on Build GRCh38
rs760271101 Chr.2: 169674605-169674605 on Build GRCh38
rs778133148 Chr.2: 169674740-169674740 on Build GRCh38
rs759938720 Chr.2: 169674578-169674578 on Build GRCh38
rs750324427 Chr.2: 169674714-169674714 on Build GRCh38
rs771199136 Chr.2: 169674660-169674660 on Build GRCh38
rs775154607 Chr.2: 169674604-169674604 on Build GRCh38

View Full Product Details