Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90286295-90286552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRDE2
Entrez Gene ID
55051
Gene Name
NRDE-2, necessary for RNA interference, domain containing
Gene Aliases
C14orf102
UniGene
Hs.528131
Gene Chromosome Location
Chr.14: 90274276-90332137 on Build GRCh38
Transcripts
NM_017970.3 XM_011536886.1 XM_017021394.1 XM_017021395.1 XM_017021396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7287034 c.3207G>A p.M1069I Chr.14: 90286444-90286444 on Build GRCh38
6035098 c.3192T>C p.P1064P Chr.14: 90286459-90286459 on Build GRCh38
3744470 c.3242G>A p.R1081H Chr.14: 90286409-90286409 on Build GRCh38
7086690 c.3241C>T p.R1081C Chr.14: 90286410-90286410 on Build GRCh38
6729517 c.3159-2A>G p.? Chr.14: 90286494-90286494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768603112 Chr.14: 90286494-90286494 on Build GRCh38
rs963578834 Chr.14: 90286394-90286394 on Build GRCh38
rs376374829 Chr.14: 90286496-90286496 on Build GRCh38
rs147836268 Chr.14: 90286471-90286471 on Build GRCh38
rs747614894 Chr.14: 90286502-90286502 on Build GRCh38
rs763650416 Chr.14: 90286337-90286337 on Build GRCh38
rs756440981 Chr.14: 90286373-90286373 on Build GRCh38
rs377535383 Chr.14: 90286522-90286522 on Build GRCh38
rs370281803 Chr.14: 90286452-90286452 on Build GRCh38
rs142496380 Chr.14: 90286408-90286408 on Build GRCh38
rs371946013 Chr.14: 90286342-90286342 on Build GRCh38
rs368794564 Chr.14: 90286509-90286509 on Build GRCh38
rs752736603 Chr.14: 90286459-90286459 on Build GRCh38
rs769634843 Chr.14: 90286321-90286321 on Build GRCh38
rs754457316 Chr.14: 90286387-90286387 on Build GRCh38
rs749271366 Chr.14: 90286412-90286412 on Build GRCh38
rs148200991 Chr.14: 90286453-90286453 on Build GRCh38
rs201924235 Chr.14: 90286485-90286485 on Build GRCh38
rs116980182 Chr.14: 90286410-90286410 on Build GRCh38
rs148410546 Chr.14: 90286390-90286390 on Build GRCh38
rs868195426 Chr.14: 90286451-90286451 on Build GRCh38
rs767154198 Chr.14: 90286369-90286369 on Build GRCh38
rs758440381 Chr.14: 90286460-90286460 on Build GRCh38
rs952024314 Chr.14: 90286331-90286331 on Build GRCh38
rs766650892 Chr.14: 90286381-90286381 on Build GRCh38
rs761867417 Chr.14: 90286447-90286448 on Build GRCh38
rs750772758 Chr.14: 90286371-90286371 on Build GRCh38
rs267604083 Chr.14: 90286441-90286441 on Build GRCh38
rs900283211 Chr.14: 90286464-90286464 on Build GRCh38
rs1015182923 Chr.14: 90286506-90286506 on Build GRCh38
rs748199867 Chr.14: 90286401-90286401 on Build GRCh38
rs369683933 Chr.14: 90286526-90286526 on Build GRCh38
rs373947973 Chr.14: 90286511-90286511 on Build GRCh38
rs779179256 Chr.14: 90286399-90286399 on Build GRCh38
rs777995436 Chr.14: 90286405-90286405 on Build GRCh38
rs373072769 Chr.14: 90286528-90286528 on Build GRCh38
rs772885445 Chr.14: 90286501-90286501 on Build GRCh38
rs775336995 Chr.14: 90286324-90286324 on Build GRCh38
rs113463686 Chr.14: 90286502-90286502 on Build GRCh38
rs766089728 Chr.14: 90286514-90286514 on Build GRCh38
rs766071717 Chr.14: 90286516-90286517 on Build GRCh38
rs768550083 Chr.14: 90286428-90286428 on Build GRCh38
rs376798752 Chr.14: 90286404-90286404 on Build GRCh38
rs751046463 Chr.14: 90286341-90286341 on Build GRCh38
rs762871589 Chr.14: 90286330-90286330 on Build GRCh38
rs980113434 Chr.14: 90286406-90286406 on Build GRCh38
rs528197535 Chr.14: 90286409-90286409 on Build GRCh38
rs567915458 Chr.14: 90286456-90286456 on Build GRCh38
rs749043037 Chr.14: 90286486-90286486 on Build GRCh38
rs767087430 Chr.14: 90286440-90286440 on Build GRCh38
rs765206507 Chr.14: 90286484-90286484 on Build GRCh38
rs773877964 Chr.14: 90286435-90286435 on Build GRCh38
rs776437992 Chr.14: 90286499-90286499 on Build GRCh38
rs144412283 Chr.14: 90286470-90286470 on Build GRCh38
rs761336895 Chr.14: 90286439-90286439 on Build GRCh38
rs986110791 Chr.14: 90286351-90286351 on Build GRCh38

View Full Product Details