Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47078624-47078869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO2
Entrez Gene ID
347902
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
ALI1, AMIGO-2, DEGA
UniGene
Hs.121520
Gene Chromosome Location
Chr.12: 47075707-47080826 on Build GRCh38
Transcripts
NM_001143668.1 NM_181847.4 XM_005268837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1705502 c.341C>T p.S114F Chr.12: 47078662-47078662 on Build GRCh38
3461118 c.282G>A p.K94K Chr.12: 47078721-47078721 on Build GRCh38
273961 c.301C>T p.R101C Chr.12: 47078702-47078702 on Build GRCh38
3461117 c.291C>T p.T97T Chr.12: 47078712-47078712 on Build GRCh38
1734613 c.377C>G p.S126C Chr.12: 47078626-47078626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779485456 Chr.12: 47078760-47078760 on Build GRCh38
rs147425809 Chr.12: 47078742-47078742 on Build GRCh38
rs772598061 Chr.12: 47078786-47078786 on Build GRCh38
rs750188293 Chr.12: 47078738-47078738 on Build GRCh38
rs759522612 Chr.12: 47078701-47078701 on Build GRCh38
rs765180978 Chr.12: 47078814-47078814 on Build GRCh38
rs771979287 Chr.12: 47078700-47078700 on Build GRCh38
rs112239025 Chr.12: 47078735-47078735 on Build GRCh38
rs761778115 Chr.12: 47078820-47078820 on Build GRCh38
rs996517583 Chr.12: 47078662-47078662 on Build GRCh38
rs747623308 Chr.12: 47078656-47078656 on Build GRCh38
rs751545427 Chr.12: 47078819-47078819 on Build GRCh38
rs952582741 Chr.12: 47078730-47078730 on Build GRCh38
rs376626328 Chr.12: 47078772-47078772 on Build GRCh38
rs757065070 Chr.12: 47078733-47078733 on Build GRCh38
rs557234455 Chr.12: 47078838-47078838 on Build GRCh38
rs537520309 Chr.12: 47078812-47078812 on Build GRCh38
rs139701778 Chr.12: 47078748-47078748 on Build GRCh38
rs766682220 Chr.12: 47078671-47078671 on Build GRCh38
rs761102831 Chr.12: 47078796-47078796 on Build GRCh38
rs779664782 Chr.12: 47078745-47078745 on Build GRCh38
rs371910751 Chr.12: 47078731-47078731 on Build GRCh38
rs963705194 Chr.12: 47078651-47078651 on Build GRCh38
rs576902763 Chr.12: 47078811-47078811 on Build GRCh38
rs773755936 Chr.12: 47078791-47078791 on Build GRCh38
rs865785572 Chr.12: 47078702-47078702 on Build GRCh38
rs764160800 Chr.12: 47078708-47078708 on Build GRCh38
rs751388566 Chr.12: 47078718-47078718 on Build GRCh38
rs754000418 Chr.12: 47078678-47078678 on Build GRCh38
rs767418969 Chr.12: 47078822-47078822 on Build GRCh38
rs767982849 Chr.12: 47078756-47078756 on Build GRCh38

View Full Product Details