Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189667141-189667396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
UniGene
Hs.101364
Gene Chromosome Location
Chr.2: 189661399-189670831 on Build GRCh38
Transcripts
NM_019048.2 XM_017004381.1 XM_017004382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695161 c.1169C>T p.A390V Chr.2: 189667301-189667301 on Build GRCh38
1404046 c.1217G>A p.R406Q Chr.2: 189667349-189667349 on Build GRCh38
1014013 c.1122G>T p.Q374H Chr.2: 189667254-189667254 on Build GRCh38
6841023 c.1167_1169delTGC p.A394delA Chr.2: 189667299-189667301 on Build GRCh38
7209149 c.1181_1183delCTG p.A394delA Chr.2: 189667313-189667315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141853606 Chr.2: 189667291-189667291 on Build GRCh38
rs750758362 Chr.2: 189667356-189667356 on Build GRCh38
rs201954770 Chr.2: 189667322-189667322 on Build GRCh38
rs536138565 Chr.2: 189667353-189667353 on Build GRCh38
rs959291123 Chr.2: 189667336-189667336 on Build GRCh38
rs756610478 Chr.2: 189667259-189667259 on Build GRCh38
rs746723583 Chr.2: 189667168-189667168 on Build GRCh38
rs746661540 Chr.2: 189667306-189667306 on Build GRCh38
rs770757193 Chr.2: 189667174-189667174 on Build GRCh38
rs765306857 Chr.2: 189667198-189667198 on Build GRCh38
rs370637526 Chr.2: 189667207-189667207 on Build GRCh38
rs755538097 Chr.2: 189667371-189667371 on Build GRCh38
rs751837652 Chr.2: 189667223-189667223 on Build GRCh38
rs939801659 Chr.2: 189667368-189667368 on Build GRCh38
rs372794498 Chr.2: 189667364-189667364 on Build GRCh38
rs779854033 Chr.2: 189667226-189667227 on Build GRCh38
rs992055118 Chr.2: 189667337-189667337 on Build GRCh38
rs200325293 Chr.2: 189667211-189667211 on Build GRCh38
rs775675550 Chr.2: 189667202-189667202 on Build GRCh38
rs754345548 Chr.2: 189667366-189667366 on Build GRCh38
rs750799341 Chr.2: 189667247-189667247 on Build GRCh38
rs140770284 Chr.2: 189667299-189667301 on Build GRCh38
rs889361933 Chr.2: 189667179-189667179 on Build GRCh38
rs142010572 Chr.2: 189667330-189667330 on Build GRCh38
rs148271409 Chr.2: 189667181-189667181 on Build GRCh38
rs757484556 Chr.2: 189667232-189667232 on Build GRCh38
rs777339697 Chr.2: 189667305-189667305 on Build GRCh38
rs752153380 Chr.2: 189667274-189667274 on Build GRCh38
rs144391161 Chr.2: 189667349-189667349 on Build GRCh38
rs768856061 Chr.2: 189667341-189667341 on Build GRCh38
rs777310473 Chr.2: 189667362-189667362 on Build GRCh38
rs374486614 Chr.2: 189667261-189667261 on Build GRCh38
rs917760084 Chr.2: 189667358-189667358 on Build GRCh38
rs772872475 Chr.2: 189667164-189667164 on Build GRCh38
rs758018468 Chr.2: 189667182-189667184 on Build GRCh38
rs1437881 Chr.2: 189667205-189667205 on Build GRCh38
rs746823522 Chr.2: 189667298-189667299 on Build GRCh38
rs199846958 Chr.2: 189667310-189667310 on Build GRCh38
rs747499215 Chr.2: 189667212-189667212 on Build GRCh38
rs766811766 Chr.2: 189667365-189667365 on Build GRCh38
rs139656772 Chr.2: 189667333-189667333 on Build GRCh38
rs775585791 Chr.2: 189667331-189667331 on Build GRCh38
rs767908739 Chr.2: 189667236-189667236 on Build GRCh38
rs774350824 Chr.2: 189667348-189667348 on Build GRCh38
rs759495798 Chr.2: 189667193-189667193 on Build GRCh38
rs926205893 Chr.2: 189667302-189667302 on Build GRCh38
rs776349243 Chr.2: 189667189-189667189 on Build GRCh38

View Full Product Details