Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38125679-38125910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA1
Entrez Gene ID
30
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
ACAA, PTHIO, THIO
UniGene
Hs.643487
Gene Chromosome Location
Chr.3: 38122710-38137242 on Build GRCh38
Transcripts
NM_001130410.1 NM_001607.3 NR_024024.1 XM_006713122.1 XM_006713123.2 XM_011533650.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
169022 c.1026C>T p.F342F Chr.3: 38125853-38125853 on Build GRCh38
2985994 c.1014C>T p.D338D Chr.3: 38125865-38125865 on Build GRCh38
1044145 c.1078C>T p.R360* Chr.3: 38125686-38125686 on Build GRCh38
6388276 c.1006G>A p.V336M Chr.3: 38125873-38125873 on Build GRCh38
3593153 c.1084C>T p.P362S Chr.3: 38125680-38125680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759862498 Chr.3: 38125865-38125865 on Build GRCh38
rs776152437 Chr.3: 38125749-38125749 on Build GRCh38
rs146607820 Chr.3: 38125770-38125770 on Build GRCh38
rs528682842 Chr.3: 38125845-38125845 on Build GRCh38
rs747828124 Chr.3: 38125868-38125868 on Build GRCh38
rs117484375 Chr.3: 38125864-38125864 on Build GRCh38
rs111568656 Chr.3: 38125823-38125823 on Build GRCh38
rs867839524 Chr.3: 38125701-38125701 on Build GRCh38
rs192098597 Chr.3: 38125736-38125736 on Build GRCh38
rs373564026 Chr.3: 38125750-38125750 on Build GRCh38
rs149958808 Chr.3: 38125880-38125880 on Build GRCh38
rs779812329 Chr.3: 38125744-38125744 on Build GRCh38
rs929301187 Chr.3: 38125820-38125820 on Build GRCh38
rs750386233 Chr.3: 38125848-38125848 on Build GRCh38
rs766732536 Chr.3: 38125725-38125727 on Build GRCh38
rs746770119 Chr.3: 38125861-38125861 on Build GRCh38
rs370654139 Chr.3: 38125769-38125769 on Build GRCh38
rs977140646 Chr.3: 38125777-38125777 on Build GRCh38
rs765838107 Chr.3: 38125881-38125881 on Build GRCh38
rs559474003 Chr.3: 38125800-38125800 on Build GRCh38
rs781630235 Chr.3: 38125735-38125735 on Build GRCh38
rs151138541 Chr.3: 38125852-38125852 on Build GRCh38
rs548538092 Chr.3: 38125758-38125758 on Build GRCh38
rs200744998 Chr.3: 38125791-38125791 on Build GRCh38
rs973499758 Chr.3: 38125811-38125811 on Build GRCh38
rs942270679 Chr.3: 38125883-38125883 on Build GRCh38
rs759867596 Chr.3: 38125808-38125808 on Build GRCh38
rs371052867 Chr.3: 38125703-38125703 on Build GRCh38
rs1000588245 Chr.3: 38125733-38125733 on Build GRCh38
rs372279103 Chr.3: 38125783-38125783 on Build GRCh38
rs144462889 Chr.3: 38125741-38125741 on Build GRCh38
rs543482153 Chr.3: 38125755-38125755 on Build GRCh38
rs766663223 Chr.3: 38125784-38125784 on Build GRCh38
rs956581427 Chr.3: 38125735-38125735 on Build GRCh38
rs752936627 Chr.3: 38125813-38125814 on Build GRCh38
rs752944703 Chr.3: 38125834-38125834 on Build GRCh38
rs779832294 Chr.3: 38125853-38125853 on Build GRCh38
rs184192813 Chr.3: 38125793-38125793 on Build GRCh38
rs370778921 Chr.3: 38125746-38125746 on Build GRCh38
rs535924556 Chr.3: 38125832-38125832 on Build GRCh38
rs377017874 Chr.3: 38125704-38125704 on Build GRCh38
rs778765436 Chr.3: 38125717-38125717 on Build GRCh38
rs768232951 Chr.3: 38125748-38125748 on Build GRCh38
rs139767196 Chr.3: 38125699-38125699 on Build GRCh38
rs377639371 Chr.3: 38125702-38125702 on Build GRCh38
rs756188064 Chr.3: 38125838-38125838 on Build GRCh38

View Full Product Details