Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100993269-100993538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf2
Entrez Gene ID
56652
Gene Name
chromosome 10 open reading frame 2
Gene Aliases
ATXN8, IOSCA, MTDPS7, PEO, PEO1, PEOA3, PRLTS5, SANDO, SCA8, TWINL
UniGene
Hs.22678
Gene Chromosome Location
Chr.10: 100987527-100994403 on Build GRCh38
Transcripts
NM_001163812.1 NM_001163813.1 NM_001163814.1 NM_021830.4 XM_011539975.2 XM_017016437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6450097 c.1853C>T p.P618L Chr.10: 100993308-100993308 on Build GRCh38
4954491 c.1935T>C p.T645T Chr.10: 100993390-100993390 on Build GRCh38
363623 c.1958C>G p.S653C Chr.10: 100993413-100993413 on Build GRCh38
1702932 c.2014C>T p.P672S Chr.10: 100993469-100993469 on Build GRCh38
3686478 c.1975G>A p.A659T Chr.10: 100993430-100993430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055355481 Chr.10: 100993422-100993422 on Build GRCh38
rs760735184 Chr.10: 100993438-100993438 on Build GRCh38
rs761514855 Chr.10: 100993336-100993338 on Build GRCh38
rs760156011 Chr.10: 100993515-100993518 on Build GRCh38
rs190487176 Chr.10: 100993397-100993397 on Build GRCh38
rs557221500 Chr.10: 100993336-100993336 on Build GRCh38
rs571312130 Chr.10: 100993434-100993434 on Build GRCh38
rs747328996 Chr.10: 100993467-100993467 on Build GRCh38
rs143659587 Chr.10: 100993512-100993512 on Build GRCh38
rs913331026 Chr.10: 100993323-100993323 on Build GRCh38
rs200335475 Chr.10: 100993516-100993516 on Build GRCh38
rs755985090 Chr.10: 100993364-100993364 on Build GRCh38
rs764752550 Chr.10: 100993299-100993299 on Build GRCh38
rs755045003 Chr.10: 100993447-100993447 on Build GRCh38
rs370814108 Chr.10: 100993430-100993430 on Build GRCh38
rs267602340 Chr.10: 100993409-100993409 on Build GRCh38
rs542793145 Chr.10: 100993394-100993394 on Build GRCh38
rs757018348 Chr.10: 100993337-100993337 on Build GRCh38
rs752911053 Chr.10: 100993457-100993457 on Build GRCh38
rs974370773 Chr.10: 100993317-100993317 on Build GRCh38
rs753984523 Chr.10: 100993444-100993444 on Build GRCh38
rs771310512 Chr.10: 100993408-100993408 on Build GRCh38
rs747597649 Chr.10: 100993403-100993403 on Build GRCh38
rs762436636 Chr.10: 100993419-100993419 on Build GRCh38
rs758995089 Chr.10: 100993292-100993292 on Build GRCh38
rs778236767 Chr.10: 100993465-100993465 on Build GRCh38
rs781620751 Chr.10: 100993472-100993472 on Build GRCh38
rs746515280 Chr.10: 100993507-100993507 on Build GRCh38
rs755312970 Chr.10: 100993366-100993366 on Build GRCh38
rs139162317 Chr.10: 100993435-100993435 on Build GRCh38
rs754165856 Chr.10: 100993508-100993508 on Build GRCh38
rs771621115 Chr.10: 100993496-100993496 on Build GRCh38
rs758523933 Chr.10: 100993463-100993463 on Build GRCh38
rs575489870 Chr.10: 100993365-100993365 on Build GRCh38
rs758267849 Chr.10: 100993478-100993478 on Build GRCh38
rs374146818 Chr.10: 100993455-100993455 on Build GRCh38
rs544684533 Chr.10: 100993309-100993309 on Build GRCh38
rs766409426 Chr.10: 100993442-100993442 on Build GRCh38
rs373741697 Chr.10: 100993477-100993477 on Build GRCh38
rs182559752 Chr.10: 100993500-100993500 on Build GRCh38
rs886046632 Chr.10: 100993308-100993308 on Build GRCh38
rs763748512 Chr.10: 100993420-100993420 on Build GRCh38
rs944443717 Chr.10: 100993425-100993425 on Build GRCh38
rs377392391 Chr.10: 100993330-100993330 on Build GRCh38
rs766667279 Chr.10: 100993370-100993372 on Build GRCh38
rs748463127 Chr.10: 100993380-100993380 on Build GRCh38
rs761569674 Chr.10: 100993481-100993481 on Build GRCh38
rs773953603 Chr.10: 100993429-100993429 on Build GRCh38
rs62626293 Chr.10: 100993357-100993357 on Build GRCh38
rs369223258 Chr.10: 100993505-100993505 on Build GRCh38
rs868535097 Chr.10: 100993427-100993427 on Build GRCh38
rs775046032 Chr.10: 100993413-100993413 on Build GRCh38
rs199583659 Chr.10: 100993470-100993470 on Build GRCh38
rs1013913162 Chr.10: 100993454-100993454 on Build GRCh38
rs779222579 Chr.10: 100993368-100993368 on Build GRCh38
rs765453709 Chr.10: 100993513-100993513 on Build GRCh38
rs774956040 Chr.10: 100993303-100993303 on Build GRCh38
rs891782757 Chr.10: 100993484-100993484 on Build GRCh38

View Full Product Details