Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22225881-22226154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO5
Entrez Gene ID
203859
Gene Name
anoctamin 5
Gene Aliases
GDD1, LGMD2L, TMEM16E
UniGene
Hs.154329
Gene Chromosome Location
Chr.11: 22192485-22283367 on Build GRCh38
Transcripts
NM_001142649.1 NM_213599.2 XM_005252820.3 XM_005252821.3 XM_005252822.4 XM_011519949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925970 c.342G>T p.E114D Chr.11: 22226031-22226031 on Build GRCh38
3446313 c.337C>G p.L113V Chr.11: 22226026-22226026 on Build GRCh38
3979518 c.340G>C p.E114Q Chr.11: 22226029-22226029 on Build GRCh38
4974665 c.295-2A>G p.? Chr.11: 22225982-22225982 on Build GRCh38
6601092 c.322C>A p.L108I Chr.11: 22226011-22226011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776859202 Chr.11: 22225989-22225993 on Build GRCh38
rs753416246 Chr.11: 22225977-22225977 on Build GRCh38
rs745616578 Chr.11: 22226024-22226024 on Build GRCh38
rs886041551 Chr.11: 22225993-22225997 on Build GRCh38
rs771699875 Chr.11: 22226087-22226087 on Build GRCh38
rs774989555 Chr.11: 22226031-22226031 on Build GRCh38
rs748664107 Chr.11: 22225937-22225941 on Build GRCh38
rs759034458 Chr.11: 22225960-22225961 on Build GRCh38
rs750707593 Chr.11: 22225980-22225980 on Build GRCh38
rs771598823 Chr.11: 22225955-22225955 on Build GRCh38
rs778956037 Chr.11: 22226020-22226020 on Build GRCh38
rs765819272 Chr.11: 22226076-22226076 on Build GRCh38
rs780109230 Chr.11: 22225983-22225983 on Build GRCh38
rs796065330 Chr.11: 22225976-22225977 on Build GRCh38
rs776244183 Chr.11: 22226070-22226070 on Build GRCh38
rs761434385 Chr.11: 22226074-22226074 on Build GRCh38
rs987506786 Chr.11: 22225924-22225924 on Build GRCh38
rs71490613 Chr.11: 22226050-22226050 on Build GRCh38
rs763750946 Chr.11: 22225974-22225974 on Build GRCh38
rs755037890 Chr.11: 22225997-22225997 on Build GRCh38
rs748258258 Chr.11: 22226003-22226003 on Build GRCh38
rs766826838 Chr.11: 22226091-22226091 on Build GRCh38
rs954701076 Chr.11: 22225986-22225986 on Build GRCh38
rs551218752 Chr.11: 22225928-22225928 on Build GRCh38
rs775246611 Chr.11: 22225958-22225958 on Build GRCh38
rs398124623 Chr.11: 22225992-22225992 on Build GRCh38
rs770803394 Chr.11: 22226016-22226016 on Build GRCh38
rs975757101 Chr.11: 22226034-22226034 on Build GRCh38
rs758847803 Chr.11: 22225982-22225982 on Build GRCh38
rs530467287 Chr.11: 22226123-22226123 on Build GRCh38
rs771226980 Chr.11: 22225976-22225977 on Build GRCh38
rs571318959 Chr.11: 22225985-22225985 on Build GRCh38
rs746125372 Chr.11: 22226089-22226089 on Build GRCh38
rs760350070 Chr.11: 22226064-22226064 on Build GRCh38
rs746448319 Chr.11: 22225968-22225968 on Build GRCh38
rs967425453 Chr.11: 22226115-22226115 on Build GRCh38
rs878854367 Chr.11: 22226041-22226041 on Build GRCh38
rs775086127 Chr.11: 22226131-22226131 on Build GRCh38
rs954832309 Chr.11: 22225904-22225904 on Build GRCh38
rs771965186 Chr.11: 22226029-22226029 on Build GRCh38
rs137854522 Chr.11: 22226056-22226056 on Build GRCh38
rs778435515 Chr.11: 22225968-22225968 on Build GRCh38
rs781180716 Chr.11: 22225999-22225999 on Build GRCh38
rs913291854 Chr.11: 22226077-22226077 on Build GRCh38
rs912437906 Chr.11: 22225927-22225927 on Build GRCh38
rs763223849 Chr.11: 22226079-22226079 on Build GRCh38
rs919753027 Chr.11: 22226067-22226067 on Build GRCh38
rs1046815059 Chr.11: 22226109-22226109 on Build GRCh38

View Full Product Details