Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39100224-39100497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3H
Entrez Gene ID
164668
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases
A3H, ARP-10, ARP10
UniGene
Hs.440515
Gene Chromosome Location
Chr.22: 39097204-39104067 on Build GRCh38
Transcripts
NM_001166002.2 NM_001166003.2 NM_001166004.2 NM_181773.4 XM_011529990.2 XM_011529991.2 XM_011529992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662517 c.19G>A p.E7K Chr.22: 39100297-39100297 on Build GRCh38
4884042 c.125C>G p.P42R Chr.22: 39100403-39100403 on Build GRCh38
3759196 c.53G>T p.R18L Chr.22: 39100331-39100331 on Build GRCh38
3759197 c.129G>C p.T43T Chr.22: 39100407-39100407 on Build GRCh38
1416316 c.40_42delAAC p.N15delN Chr.22: 39100318-39100320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140936762 Chr.22: 39100318-39100320 on Build GRCh38
rs563921695 Chr.22: 39100438-39100438 on Build GRCh38
rs761854928 Chr.22: 39100440-39100440 on Build GRCh38
rs139293 Chr.22: 39100331-39100331 on Build GRCh38
rs189764428 Chr.22: 39100359-39100359 on Build GRCh38
rs376048091 Chr.22: 39100410-39100410 on Build GRCh38
rs774051298 Chr.22: 39100337-39100337 on Build GRCh38
rs146649681 Chr.22: 39100412-39100412 on Build GRCh38
rs776130540 Chr.22: 39100253-39100253 on Build GRCh38
rs760113060 Chr.22: 39100297-39100297 on Build GRCh38
rs139292 Chr.22: 39100323-39100325 on Build GRCh38
rs750601192 Chr.22: 39100385-39100385 on Build GRCh38
rs749078969 Chr.22: 39100323-39100323 on Build GRCh38
rs5757475 Chr.22: 39100389-39100389 on Build GRCh38
rs764240600 Chr.22: 39100336-39100337 on Build GRCh38
rs747509986 Chr.22: 39100328-39100328 on Build GRCh38
rs201177427 Chr.22: 39100322-39100324 on Build GRCh38
rs759128070 Chr.22: 39100341-39100341 on Build GRCh38
rs765341469 Chr.22: 39100443-39100443 on Build GRCh38
rs749399007 Chr.22: 39100311-39100311 on Build GRCh38
rs762492011 Chr.22: 39100332-39100332 on Build GRCh38
rs369126907 Chr.22: 39100383-39100383 on Build GRCh38
rs766070770 Chr.22: 39100281-39100281 on Build GRCh38
rs764738270 Chr.22: 39100267-39100267 on Build GRCh38
rs149415555 Chr.22: 39100382-39100382 on Build GRCh38
rs772425911 Chr.22: 39100249-39100249 on Build GRCh38
rs764174253 Chr.22: 39100360-39100360 on Build GRCh38
rs772740027 Chr.22: 39100277-39100277 on Build GRCh38
rs374766186 Chr.22: 39100327-39100327 on Build GRCh38
rs767346992 Chr.22: 39100306-39100306 on Build GRCh38
rs750491882 Chr.22: 39100445-39100445 on Build GRCh38
rs867564302 Chr.22: 39100380-39100380 on Build GRCh38
rs202012802 Chr.22: 39100400-39100400 on Build GRCh38
rs779316580 Chr.22: 39100318-39100318 on Build GRCh38
rs747383920 Chr.22: 39100401-39100401 on Build GRCh38
rs937862707 Chr.22: 39100308-39100308 on Build GRCh38
rs372088508 Chr.22: 39100307-39100307 on Build GRCh38
rs113212259 Chr.22: 39100460-39100460 on Build GRCh38
rs150949141 Chr.22: 39100397-39100397 on Build GRCh38
rs754870937 Chr.22: 39100297-39100297 on Build GRCh38
rs369700866 Chr.22: 39100466-39100466 on Build GRCh38
rs759184375 Chr.22: 39100421-39100421 on Build GRCh38
rs767272509 Chr.22: 39100346-39100346 on Build GRCh38
rs761214813 Chr.22: 39100256-39100256 on Build GRCh38
rs545736738 Chr.22: 39100437-39100437 on Build GRCh38
rs931374735 Chr.22: 39100314-39100314 on Build GRCh38
rs771641702 Chr.22: 39100425-39100425 on Build GRCh38
rs746229527 Chr.22: 39100325-39100325 on Build GRCh38
rs139294 Chr.22: 39100407-39100407 on Build GRCh38
rs777865663 Chr.22: 39100309-39100309 on Build GRCh38
rs753936907 Chr.22: 39100378-39100378 on Build GRCh38
rs991057105 Chr.22: 39100312-39100312 on Build GRCh38
rs539789572 Chr.22: 39100296-39100296 on Build GRCh38
rs142563386 Chr.22: 39100352-39100352 on Build GRCh38
rs76049870 Chr.22: 39100289-39100289 on Build GRCh38
rs1010218879 Chr.22: 39100270-39100270 on Build GRCh38
rs374189433 Chr.22: 39100246-39100246 on Build GRCh38
rs780182369 Chr.22: 39100458-39100458 on Build GRCh38
rs139620128 Chr.22: 39100336-39100336 on Build GRCh38
rs757527498 Chr.22: 39100313-39100313 on Build GRCh38
rs758625079 Chr.22: 39100452-39100452 on Build GRCh38
rs760023124 Chr.22: 39100273-39100274 on Build GRCh38
rs571817129 Chr.22: 39100436-39100436 on Build GRCh38
rs763213711 Chr.22: 39100322-39100323 on Build GRCh38
rs368714381 Chr.22: 39100278-39100278 on Build GRCh38
rs751950985 Chr.22: 39100459-39100459 on Build GRCh38
rs769176395 Chr.22: 39100406-39100406 on Build GRCh38

View Full Product Details