Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73498749-73498998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTF3
Entrez Gene ID
689
Gene Name
basic transcription factor 3
Gene Aliases
BETA-NAC, BTF3a, BTF3b, NACB
UniGene
Hs.591768
Gene Chromosome Location
Chr.5: 73498425-73505623 on Build GRCh38
Transcripts
NM_001037637.1 NM_001207.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416081 c.122A>T p.Q41L Chr.5: 73498789-73498789 on Build GRCh38
3393659 c.132+8G>A p.? Chr.5: 73498807-73498807 on Build GRCh38
5433182 c.116G>A p.R39Q Chr.5: 73498783-73498783 on Build GRCh38
6431095 c.91C>T p.Q31* Chr.5: 73498758-73498758 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958149151 Chr.5: 73498880-73498880 on Build GRCh38
rs768751845 Chr.5: 73498852-73498853 on Build GRCh38
rs891186498 Chr.5: 73498960-73498960 on Build GRCh38
rs916611357 Chr.5: 73498913-73498913 on Build GRCh38
rs3057010 Chr.5: 73498927-73498928 on Build GRCh38
rs1052803478 Chr.5: 73498857-73498857 on Build GRCh38
rs546479065 Chr.5: 73498852-73498852 on Build GRCh38
rs923731366 Chr.5: 73498878-73498878 on Build GRCh38
rs188464958 Chr.5: 73498895-73498895 on Build GRCh38
rs36032413 Chr.5: 73498953-73498953 on Build GRCh38
rs1141021 Chr.5: 73498788-73498788 on Build GRCh38
rs1007245004 Chr.5: 73498858-73498863 on Build GRCh38
rs911238120 Chr.5: 73498767-73498767 on Build GRCh38
rs991930087 Chr.5: 73498874-73498874 on Build GRCh38
rs760904650 Chr.5: 73498926-73498927 on Build GRCh38
rs1044425838 Chr.5: 73498876-73498876 on Build GRCh38
rs896915700 Chr.5: 73498774-73498774 on Build GRCh38
rs1029844438 Chr.5: 73498942-73498942 on Build GRCh38
rs532461640 Chr.5: 73498877-73498877 on Build GRCh38
rs771068947 Chr.5: 73498947-73498947 on Build GRCh38
rs374787775 Chr.5: 73498928-73498929 on Build GRCh38
rs528335034 Chr.5: 73498827-73498827 on Build GRCh38
rs897321367 Chr.5: 73498914-73498914 on Build GRCh38
rs978723605 Chr.5: 73498828-73498828 on Build GRCh38
rs934527099 Chr.5: 73498856-73498856 on Build GRCh38
rs928250289 Chr.5: 73498976-73498976 on Build GRCh38
rs916960419 Chr.5: 73498870-73498870 on Build GRCh38
rs917771272 Chr.5: 73498961-73498961 on Build GRCh38
rs774205468 Chr.5: 73498839-73498839 on Build GRCh38
rs994297154 Chr.5: 73498915-73498915 on Build GRCh38
rs535957770 Chr.5: 73498974-73498974 on Build GRCh38
rs769856280 Chr.5: 73498791-73498791 on Build GRCh38
rs146342908 Chr.5: 73498970-73498970 on Build GRCh38
rs747069228 Chr.5: 73498939-73498939 on Build GRCh38
rs142006356 Chr.5: 73498875-73498875 on Build GRCh38
rs905547587 Chr.5: 73498883-73498883 on Build GRCh38
rs995176698 Chr.5: 73498835-73498835 on Build GRCh38
rs1038645287 Chr.5: 73498898-73498898 on Build GRCh38

View Full Product Details