Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99074851-99075124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH5
Entrez Gene ID
128
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Gene Aliases
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p
UniGene
Hs.78989
Gene Chromosome Location
Chr.4: 99070978-99088788 on Build GRCh38
Transcripts
NM_000671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512594 c.908C>T p.T303I Chr.4: 99074967-99074967 on Build GRCh38
6203178 c.925G>A p.V309I Chr.4: 99074950-99074950 on Build GRCh38
6101490 c.826-1G>C p.? Chr.4: 99075050-99075050 on Build GRCh38
4126907 c.905C>T p.A302V Chr.4: 99074970-99074970 on Build GRCh38
7312086 c.918C>A p.F306L Chr.4: 99074957-99074957 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs28730629 Chr.4: 99074885-99074885 on Build GRCh38
rs771824754 Chr.4: 99074906-99074906 on Build GRCh38
rs78820902 Chr.4: 99075069-99075069 on Build GRCh38
rs749866392 Chr.4: 99075073-99075073 on Build GRCh38
rs747862881 Chr.4: 99075053-99075053 on Build GRCh38
rs368604169 Chr.4: 99075057-99075057 on Build GRCh38
rs762072471 Chr.4: 99074977-99074977 on Build GRCh38
rs770013922 Chr.4: 99075057-99075058 on Build GRCh38
rs570631002 Chr.4: 99075103-99075103 on Build GRCh38
rs374481239 Chr.4: 99074941-99074941 on Build GRCh38
rs754853545 Chr.4: 99075043-99075043 on Build GRCh38
rs772887965 Chr.4: 99074907-99074907 on Build GRCh38
rs753876606 Chr.4: 99075030-99075030 on Build GRCh38
rs552228974 Chr.4: 99075008-99075008 on Build GRCh38
rs774821719 Chr.4: 99075083-99075083 on Build GRCh38
rs962060219 Chr.4: 99074976-99074976 on Build GRCh38
rs374579100 Chr.4: 99075085-99075085 on Build GRCh38
rs28730627 Chr.4: 99075102-99075104 on Build GRCh38
rs776001954 Chr.4: 99075068-99075068 on Build GRCh38
rs756141527 Chr.4: 99075004-99075004 on Build GRCh38
rs778563437 Chr.4: 99074881-99074881 on Build GRCh38
rs372874345 Chr.4: 99075064-99075064 on Build GRCh38
rs750535620 Chr.4: 99074995-99074995 on Build GRCh38
rs560307792 Chr.4: 99074964-99074964 on Build GRCh38
rs767752183 Chr.4: 99075098-99075098 on Build GRCh38
rs778814924 Chr.4: 99075050-99075050 on Build GRCh38
rs912381311 Chr.4: 99074975-99074975 on Build GRCh38
rs770446863 Chr.4: 99074936-99074936 on Build GRCh38
rs112298116 Chr.4: 99074962-99074962 on Build GRCh38
rs28730628 Chr.4: 99074950-99074950 on Build GRCh38
rs769023481 Chr.4: 99075078-99075078 on Build GRCh38
rs770499965 Chr.4: 99075066-99075066 on Build GRCh38
rs527917175 Chr.4: 99074966-99074966 on Build GRCh38
rs763232284 Chr.4: 99074947-99074947 on Build GRCh38
rs760221275 Chr.4: 99074908-99074908 on Build GRCh38
rs773369050 Chr.4: 99075099-99075099 on Build GRCh38
rs767698666 Chr.4: 99074982-99074982 on Build GRCh38

View Full Product Details