Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4899667-4899926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf107
Entrez Gene ID
100130311
Gene Name
chromosome 17 open reading frame 107
Gene Aliases
-
UniGene
Hs.579243
Gene Chromosome Location
Chr.17: 4899653-4902932 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832550 c.39G>C p.W13C Chr.17: 4899801-4899801 on Build GRCh38
5972265 c.31C>T p.L11L Chr.17: 4899793-4899793 on Build GRCh38
7214470 c.9G>C p.G3G Chr.17: 4899771-4899771 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746102360 Chr.17: 4899905-4899920 on Build GRCh38
rs765978613 Chr.17: 4899896-4899896 on Build GRCh38
rs969186963 Chr.17: 4899699-4899699 on Build GRCh38
rs1023519124 Chr.17: 4899886-4899886 on Build GRCh38
rs757103797 Chr.17: 4899793-4899793 on Build GRCh38
rs995149076 Chr.17: 4899725-4899725 on Build GRCh38
rs763258127 Chr.17: 4899876-4899876 on Build GRCh38
rs765735484 Chr.17: 4899870-4899870 on Build GRCh38
rs187059456 Chr.17: 4899855-4899855 on Build GRCh38
rs752694493 Chr.17: 4899891-4899892 on Build GRCh38
rs762331553 Chr.17: 4899713-4899713 on Build GRCh38
rs916430553 Chr.17: 4899693-4899693 on Build GRCh38
rs778410074 Chr.17: 4899807-4899807 on Build GRCh38
rs971778405 Chr.17: 4899731-4899731 on Build GRCh38
rs558951496 Chr.17: 4899776-4899776 on Build GRCh38
rs1048601616 Chr.17: 4899781-4899781 on Build GRCh38
rs752528236 Chr.17: 4899759-4899759 on Build GRCh38
rs540854905 Chr.17: 4899860-4899860 on Build GRCh38
rs771447345 Chr.17: 4899826-4899826 on Build GRCh38
rs758376347 Chr.17: 4899727-4899727 on Build GRCh38
rs920241546 Chr.17: 4899756-4899756 on Build GRCh38
rs554023721 Chr.17: 4899873-4899873 on Build GRCh38
rs766311064 Chr.17: 4899887-4899887 on Build GRCh38
rs991296636 Chr.17: 4899842-4899842 on Build GRCh38
rs756059704 Chr.17: 4899763-4899763 on Build GRCh38
rs890612632 Chr.17: 4899689-4899689 on Build GRCh38
rs143413388 Chr.17: 4899700-4899700 on Build GRCh38
rs182418908 Chr.17: 4899687-4899687 on Build GRCh38
rs569156733 Chr.17: 4899839-4899839 on Build GRCh38
rs893785612 Chr.17: 4899882-4899882 on Build GRCh38
rs377353872 Chr.17: 4899714-4899714 on Build GRCh38
rs931578136 Chr.17: 4899764-4899764 on Build GRCh38
rs555087310 Chr.17: 4899853-4899853 on Build GRCh38
rs76421344 Chr.17: 4899850-4899850 on Build GRCh38
rs951324756 Chr.17: 4899787-4899787 on Build GRCh38
rs766510474 Chr.17: 4899740-4899740 on Build GRCh38
rs1041599982 Chr.17: 4899818-4899818 on Build GRCh38
rs375052652 Chr.17: 4899843-4899843 on Build GRCh38
rs1010847156 Chr.17: 4899883-4899883 on Build GRCh38
rs539314116 Chr.17: 4899771-4899771 on Build GRCh38
rs369152087 Chr.17: 4899834-4899834 on Build GRCh38
rs745578004 Chr.17: 4899813-4899813 on Build GRCh38
rs899557194 Chr.17: 4899723-4899723 on Build GRCh38

View Full Product Details