Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81441759-81442032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHCC1
Entrez Gene ID
57597
Gene Name
BAH domain and coiled-coil containing 1
Gene Aliases
BAHD2
UniGene
Hs.514580
Gene Chromosome Location
Chr.17: 81395431-81466332 on Build GRCh38
Transcripts
NM_001291324.1 XM_011525063.2 XM_017024898.1 XM_017024899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6580139 c.482-8C>A p.? Chr.17: 81441823-81441823 on Build GRCh38
6312912 c.528G>T p.P176P Chr.17: 81441877-81441877 on Build GRCh38
4919649 c.660G>A p.V220V Chr.17: 81442009-81442009 on Build GRCh38
7105236 c.550G>A p.V184M Chr.17: 81441899-81441899 on Build GRCh38
4632731 c.580G>T p.G194C Chr.17: 81441929-81441929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782085804 Chr.17: 81441845-81441845 on Build GRCh38
rs369843602 Chr.17: 81441995-81441995 on Build GRCh38
rs782018075 Chr.17: 81441917-81441917 on Build GRCh38
rs781999039 Chr.17: 81441844-81441844 on Build GRCh38
rs782421130 Chr.17: 81441988-81441988 on Build GRCh38
rs782537173 Chr.17: 81441942-81441942 on Build GRCh38
rs116138822 Chr.17: 81441898-81441898 on Build GRCh38
rs781916395 Chr.17: 81441997-81441997 on Build GRCh38
rs782515354 Chr.17: 81441856-81441856 on Build GRCh38
rs782674396 Chr.17: 81441948-81441948 on Build GRCh38
rs542765721 Chr.17: 81441954-81441954 on Build GRCh38
rs782589241 Chr.17: 81441955-81441955 on Build GRCh38
rs782784821 Chr.17: 81441999-81441999 on Build GRCh38
rs577913810 Chr.17: 81441855-81441855 on Build GRCh38
rs782376513 Chr.17: 81441794-81441794 on Build GRCh38
rs370107300 Chr.17: 81441923-81441923 on Build GRCh38
rs782430717 Chr.17: 81441868-81441868 on Build GRCh38
rs545200214 Chr.17: 81441877-81441877 on Build GRCh38
rs758323235 Chr.17: 81441992-81441992 on Build GRCh38
rs376051584 Chr.17: 81441905-81441905 on Build GRCh38
rs199920012 Chr.17: 81441957-81441957 on Build GRCh38
rs782576981 Chr.17: 81441788-81441788 on Build GRCh38
rs782051705 Chr.17: 81441816-81441816 on Build GRCh38
rs781973096 Chr.17: 81441908-81441908 on Build GRCh38
rs782281838 Chr.17: 81441986-81441986 on Build GRCh38
rs781895622 Chr.17: 81442000-81442000 on Build GRCh38
rs782404709 Chr.17: 81441797-81441797 on Build GRCh38
rs782057281 Chr.17: 81441911-81441911 on Build GRCh38
rs782013292 Chr.17: 81441822-81441822 on Build GRCh38
rs200849322 Chr.17: 81441994-81441994 on Build GRCh38
rs782645349 Chr.17: 81441864-81441864 on Build GRCh38
rs782608042 Chr.17: 81441810-81441810 on Build GRCh38
rs201597980 Chr.17: 81441899-81441899 on Build GRCh38
rs782765812 Chr.17: 81441931-81441931 on Build GRCh38
rs782554430 Chr.17: 81441784-81441785 on Build GRCh38
rs372977595 Chr.17: 81441867-81441867 on Build GRCh38
rs782674826 Chr.17: 81441789-81441790 on Build GRCh38
rs782476877 Chr.17: 81441939-81441939 on Build GRCh38
rs782704662 Chr.17: 81441941-81441941 on Build GRCh38
rs782722714 Chr.17: 81441915-81441915 on Build GRCh38
rs782281202 Chr.17: 81441896-81441896 on Build GRCh38
rs781969774 Chr.17: 81441815-81441815 on Build GRCh38
rs377738105 Chr.17: 81441906-81441906 on Build GRCh38
rs782241889 Chr.17: 81441953-81441953 on Build GRCh38
rs782213915 Chr.17: 81441819-81441819 on Build GRCh38
rs553003334 Chr.17: 81441850-81441850 on Build GRCh38
rs372078407 Chr.17: 81441965-81441965 on Build GRCh38
rs782329221 Chr.17: 81441821-81441821 on Build GRCh38
rs782494007 Chr.17: 81441784-81441784 on Build GRCh38
rs782116373 Chr.17: 81442006-81442006 on Build GRCh38
rs374129514 Chr.17: 81441979-81441979 on Build GRCh38
rs782153654 Chr.17: 81441824-81441824 on Build GRCh38
rs782749686 Chr.17: 81441828-81441828 on Build GRCh38
rs782063768 Chr.17: 81441998-81441998 on Build GRCh38
rs782798125 Chr.17: 81441853-81441853 on Build GRCh38
rs782198198 Chr.17: 81441791-81441791 on Build GRCh38
rs782339010 Chr.17: 81441807-81441807 on Build GRCh38
rs369477787 Chr.17: 81441805-81441805 on Build GRCh38
rs990731433 Chr.17: 81441897-81441897 on Build GRCh38
rs782692718 Chr.17: 81441798-81441798 on Build GRCh38
rs782753355 Chr.17: 81441846-81441846 on Build GRCh38
rs374363244 Chr.17: 81441937-81441937 on Build GRCh38

View Full Product Details