Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38327497-38327754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
UniGene
Hs.529019
Gene Chromosome Location
Chr.20: 38304150-38337503 on Build GRCh38
Transcripts
NM_001725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4637844 c.1204G>A p.E402K Chr.20: 38327618-38327618 on Build GRCh38
4134525 c.1210A>G p.N404D Chr.20: 38327624-38327624 on Build GRCh38
5926082 c.1199G>A p.S400N Chr.20: 38327613-38327613 on Build GRCh38
79549 c.1188C>T p.S396S Chr.20: 38327602-38327602 on Build GRCh38
2761336 c.1229T>A p.L410H Chr.20: 38327643-38327643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745705452 Chr.20: 38327577-38327577 on Build GRCh38
rs769893829 Chr.20: 38327692-38327692 on Build GRCh38
rs936480285 Chr.20: 38327665-38327665 on Build GRCh38
rs572597261 Chr.20: 38327546-38327546 on Build GRCh38
rs760378805 Chr.20: 38327592-38327592 on Build GRCh38
rs758742697 Chr.20: 38327537-38327537 on Build GRCh38
rs768577314 Chr.20: 38327594-38327594 on Build GRCh38
rs5741809 Chr.20: 38327624-38327624 on Build GRCh38
rs34137603 Chr.20: 38327590-38327590 on Build GRCh38
rs980192085 Chr.20: 38327614-38327614 on Build GRCh38
rs760548784 Chr.20: 38327591-38327591 on Build GRCh38
rs368846296 Chr.20: 38327552-38327552 on Build GRCh38
rs5743529 Chr.20: 38327723-38327723 on Build GRCh38
rs373213999 Chr.20: 38327608-38327608 on Build GRCh38
rs376936419 Chr.20: 38327629-38327629 on Build GRCh38
rs866533360 Chr.20: 38327690-38327690 on Build GRCh38
rs752039728 Chr.20: 38327551-38327551 on Build GRCh38
rs531238626 Chr.20: 38327662-38327662 on Build GRCh38
rs748270566 Chr.20: 38327685-38327685 on Build GRCh38
rs961563514 Chr.20: 38327526-38327526 on Build GRCh38
rs386814027 Chr.20: 38327707-38327708 on Build GRCh38
rs183212486 Chr.20: 38327708-38327708 on Build GRCh38
rs779723515 Chr.20: 38327670-38327670 on Build GRCh38
rs548563166 Chr.20: 38327678-38327678 on Build GRCh38
rs762336689 Chr.20: 38327667-38327668 on Build GRCh38
rs778745918 Chr.20: 38327667-38327667 on Build GRCh38
rs781763349 Chr.20: 38327648-38327648 on Build GRCh38
rs770774033 Chr.20: 38327568-38327568 on Build GRCh38
rs766677571 Chr.20: 38327618-38327618 on Build GRCh38
rs530448991 Chr.20: 38327677-38327677 on Build GRCh38
rs5743527 Chr.20: 38327617-38327617 on Build GRCh38
rs5743528 Chr.20: 38327707-38327707 on Build GRCh38
rs563848454 Chr.20: 38327664-38327664 on Build GRCh38
rs563914333 Chr.20: 38327695-38327695 on Build GRCh38
rs151281669 Chr.20: 38327589-38327589 on Build GRCh38
rs778693174 Chr.20: 38327572-38327572 on Build GRCh38
rs546999507 Chr.20: 38327727-38327727 on Build GRCh38
rs771701688 Chr.20: 38327668-38327668 on Build GRCh38
rs1008195381 Chr.20: 38327721-38327721 on Build GRCh38
rs766172249 Chr.20: 38327615-38327615 on Build GRCh38
rs776762822 Chr.20: 38327598-38327598 on Build GRCh38
rs753455111 Chr.20: 38327663-38327663 on Build GRCh38
rs541774002 Chr.20: 38327656-38327656 on Build GRCh38
rs563886979 Chr.20: 38327532-38327532 on Build GRCh38
rs5741810 Chr.20: 38327698-38327698 on Build GRCh38
rs748739821 Chr.20: 38327555-38327555 on Build GRCh38
rs776530009 Chr.20: 38327682-38327682 on Build GRCh38

View Full Product Details