Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89188124-89188397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD2
Entrez Gene ID
11057
Gene Name
abhydrolase domain containing 2
Gene Aliases
HS1-2, LABH2, PHPS1-2
UniGene
Hs.122337
Gene Chromosome Location
Chr.15: 89088150-89202360 on Build GRCh38
Transcripts
NM_007011.7 NM_152924.4 XM_005254832.1 XM_005254833.1 XM_011521165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
305550 c.823C>T p.L275F Chr.15: 89188200-89188200 on Build GRCh38
4333196 c.861A>T p.E287D Chr.15: 89188238-89188238 on Build GRCh38
2015165 c.851A>G p.Q284R Chr.15: 89188228-89188228 on Build GRCh38
434507 c.848C>T p.P283L Chr.15: 89188225-89188225 on Build GRCh38
1608798 c.883T>C p.Y295H Chr.15: 89188260-89188260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189147436 Chr.15: 89188218-89188218 on Build GRCh38
rs767644116 Chr.15: 89188344-89188344 on Build GRCh38
rs924775630 Chr.15: 89188254-89188254 on Build GRCh38
rs765464096 Chr.15: 89188231-89188231 on Build GRCh38
rs561242753 Chr.15: 89188244-89188244 on Build GRCh38
rs544347453 Chr.15: 89188164-89188164 on Build GRCh38
rs184637482 Chr.15: 89188200-89188200 on Build GRCh38
rs777443677 Chr.15: 89188315-89188315 on Build GRCh38
rs754955538 Chr.15: 89188311-89188311 on Build GRCh38
rs71405636 Chr.15: 89188192-89188192 on Build GRCh38
rs772591124 Chr.15: 89188189-89188189 on Build GRCh38
rs771760672 Chr.15: 89188186-89188186 on Build GRCh38
rs934821209 Chr.15: 89188267-89188269 on Build GRCh38
rs746310569 Chr.15: 89188330-89188330 on Build GRCh38
rs1031566592 Chr.15: 89188181-89188181 on Build GRCh38
rs774673321 Chr.15: 89188340-89188340 on Build GRCh38
rs764438488 Chr.15: 89188259-89188259 on Build GRCh38
rs1049917580 Chr.15: 89188331-89188331 on Build GRCh38
rs928634124 Chr.15: 89188260-89188260 on Build GRCh38
rs747696520 Chr.15: 89188166-89188166 on Build GRCh38
rs763085663 Chr.15: 89188245-89188245 on Build GRCh38
rs757451013 Chr.15: 89188263-89188263 on Build GRCh38
rs528631828 Chr.15: 89188294-89188294 on Build GRCh38
rs192520435 Chr.15: 89188314-89188314 on Build GRCh38
rs750633826 Chr.15: 89188339-89188339 on Build GRCh38
rs756188283 Chr.15: 89188236-89188238 on Build GRCh38
rs368935537 Chr.15: 89188224-89188224 on Build GRCh38
rs763427038 Chr.15: 89188334-89188334 on Build GRCh38
rs561978329 Chr.15: 89188199-89188199 on Build GRCh38
rs776132086 Chr.15: 89188320-89188320 on Build GRCh38
rs28565281 Chr.15: 89188333-89188333 on Build GRCh38
rs770403421 Chr.15: 89188332-89188332 on Build GRCh38
rs78193744 Chr.15: 89188210-89188210 on Build GRCh38
rs753775299 Chr.15: 89188275-89188275 on Build GRCh38
rs1040096424 Chr.15: 89188361-89188361 on Build GRCh38
rs754867511 Chr.15: 89188348-89188348 on Build GRCh38
rs915740489 Chr.15: 89188226-89188226 on Build GRCh38
rs760058843 Chr.15: 89188229-89188229 on Build GRCh38
rs751663877 Chr.15: 89188262-89188262 on Build GRCh38
rs770379924 Chr.15: 89188319-89188319 on Build GRCh38
rs767732232 Chr.15: 89188274-89188274 on Build GRCh38
rs201349282 Chr.15: 89188312-89188312 on Build GRCh38
rs116127688 Chr.15: 89188197-89188197 on Build GRCh38
rs752843590 Chr.15: 89188169-89188172 on Build GRCh38
rs373053634 Chr.15: 89188232-89188232 on Build GRCh38
rs184752726 Chr.15: 89188317-89188317 on Build GRCh38
rs765275609 Chr.15: 89188350-89188350 on Build GRCh38
rs778384944 Chr.15: 89188322-89188322 on Build GRCh38

View Full Product Details