Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17294281-17294552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711274 c.1050C>T p.G350G Chr.19: 17294387-17294387 on Build GRCh38
564910 c.1147G>T p.E383* Chr.19: 17294290-17294290 on Build GRCh38
4838001 c.1045G>A p.E349K Chr.19: 17294392-17294392 on Build GRCh38
6373338 c.978G>T p.K326N Chr.19: 17294459-17294459 on Build GRCh38
6590919 c.934G>A p.A312T Chr.19: 17294503-17294503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776978981 Chr.19: 17294360-17294360 on Build GRCh38
rs1017154349 Chr.19: 17294441-17294441 on Build GRCh38
rs745919339 Chr.19: 17294324-17294324 on Build GRCh38
rs374194413 Chr.19: 17294371-17294371 on Build GRCh38
rs746055933 Chr.19: 17294491-17294491 on Build GRCh38
rs1039561725 Chr.19: 17294299-17294299 on Build GRCh38
rs1028990706 Chr.19: 17294494-17294494 on Build GRCh38
rs774142596 Chr.19: 17294438-17294439 on Build GRCh38
rs771183001 Chr.19: 17294335-17294335 on Build GRCh38
rs777956154 Chr.19: 17294330-17294330 on Build GRCh38
rs199521781 Chr.19: 17294316-17294316 on Build GRCh38
rs375406198 Chr.19: 17294521-17294521 on Build GRCh38
rs138969474 Chr.19: 17294383-17294383 on Build GRCh38
rs773133263 Chr.19: 17294524-17294524 on Build GRCh38
rs760613065 Chr.19: 17294531-17294531 on Build GRCh38
rs536645024 Chr.19: 17294387-17294387 on Build GRCh38
rs145137671 Chr.19: 17294321-17294321 on Build GRCh38
rs747053634 Chr.19: 17294334-17294334 on Build GRCh38
rs761490842 Chr.19: 17294520-17294520 on Build GRCh38
rs753031036 Chr.19: 17294435-17294435 on Build GRCh38
rs77887041 Chr.19: 17294533-17294533 on Build GRCh38
rs1005725052 Chr.19: 17294425-17294425 on Build GRCh38
rs556911878 Chr.19: 17294393-17294393 on Build GRCh38
rs998316977 Chr.19: 17294317-17294317 on Build GRCh38
rs140686610 Chr.19: 17294462-17294462 on Build GRCh38
rs758833512 Chr.19: 17294438-17294438 on Build GRCh38
rs779284386 Chr.19: 17294433-17294433 on Build GRCh38
rs775823503 Chr.19: 17294512-17294512 on Build GRCh38
rs753466792 Chr.19: 17294408-17294408 on Build GRCh38
rs368480144 Chr.19: 17294381-17294381 on Build GRCh38
rs759741263 Chr.19: 17294362-17294362 on Build GRCh38
rs186428731 Chr.19: 17294407-17294407 on Build GRCh38
rs747576672 Chr.19: 17294460-17294460 on Build GRCh38
rs190922068 Chr.19: 17294449-17294449 on Build GRCh38
rs889098002 Chr.19: 17294506-17294506 on Build GRCh38
rs1013484743 Chr.19: 17294503-17294503 on Build GRCh38
rs868423446 Chr.19: 17294490-17294490 on Build GRCh38
rs370502455 Chr.19: 17294501-17294501 on Build GRCh38
rs553178516 Chr.19: 17294455-17294455 on Build GRCh38
rs202076995 Chr.19: 17294440-17294440 on Build GRCh38
rs754597785 Chr.19: 17294415-17294415 on Build GRCh38
rs781423967 Chr.19: 17294474-17294474 on Build GRCh38
rs767189361 Chr.19: 17294516-17294516 on Build GRCh38
rs778402701 Chr.19: 17294448-17294448 on Build GRCh38
rs894626012 Chr.19: 17294481-17294481 on Build GRCh38
rs761864032 Chr.19: 17294473-17294474 on Build GRCh38
rs915796204 Chr.19: 17294511-17294511 on Build GRCh38
rs761770440 Chr.19: 17294384-17294384 on Build GRCh38
rs767412487 Chr.19: 17294385-17294385 on Build GRCh38
rs750266959 Chr.19: 17294518-17294519 on Build GRCh38

View Full Product Details