Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46673065-46673302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX38
Entrez Gene ID
374973
Gene Name
testis expressed 38
Gene Aliases
ATPAF1-AS1, C1orf223, THEG4
UniGene
Hs.443299
Gene Chromosome Location
Chr.1: 46669344-46673603 on Build GRCh38
Transcripts
NM_001145474.3 NM_001300863.1 NM_001300864.1 XM_011541421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548724 c.616G>A p.E206K Chr.1: 46673289-46673289 on Build GRCh38
4423235 c.410C>T p.T137M Chr.1: 46673083-46673083 on Build GRCh38
1152111 c.528G>A p.A176A Chr.1: 46673201-46673201 on Build GRCh38
1584613 c.497G>A p.S166N Chr.1: 46673170-46673170 on Build GRCh38
6540430 c.422C>T p.P141L Chr.1: 46673095-46673095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763198126 Chr.1: 46673176-46673176 on Build GRCh38
rs761233526 Chr.1: 46673104-46673104 on Build GRCh38
rs949123189 Chr.1: 46673223-46673223 on Build GRCh38
rs1011434271 Chr.1: 46673126-46673126 on Build GRCh38
rs762623049 Chr.1: 46673124-46673124 on Build GRCh38
rs614486 Chr.1: 46673147-46673147 on Build GRCh38
rs767244664 Chr.1: 46673160-46673160 on Build GRCh38
rs776131772 Chr.1: 46673110-46673110 on Build GRCh38
rs749364461 Chr.1: 46673243-46673243 on Build GRCh38
rs529892680 Chr.1: 46673217-46673217 on Build GRCh38
rs960947295 Chr.1: 46673186-46673186 on Build GRCh38
rs557033219 Chr.1: 46673246-46673246 on Build GRCh38
rs965443580 Chr.1: 46673122-46673122 on Build GRCh38
rs369089422 Chr.1: 46673159-46673159 on Build GRCh38
rs988931326 Chr.1: 46673192-46673192 on Build GRCh38
rs916429158 Chr.1: 46673224-46673224 on Build GRCh38
rs200946921 Chr.1: 46673121-46673121 on Build GRCh38
rs377229148 Chr.1: 46673105-46673105 on Build GRCh38
rs750254026 Chr.1: 46673169-46673169 on Build GRCh38
rs756022932 Chr.1: 46673180-46673180 on Build GRCh38
rs1028309848 Chr.1: 46673134-46673134 on Build GRCh38
rs746317996 Chr.1: 46673129-46673129 on Build GRCh38
rs1038104626 Chr.1: 46673264-46673264 on Build GRCh38
rs372815487 Chr.1: 46673135-46673135 on Build GRCh38
rs918354322 Chr.1: 46673266-46673266 on Build GRCh38
rs571767389 Chr.1: 46673168-46673168 on Build GRCh38
rs151085882 Chr.1: 46673094-46673094 on Build GRCh38
rs865881439 Chr.1: 46673145-46673145 on Build GRCh38
rs927850515 Chr.1: 46673166-46673166 on Build GRCh38
rs139847766 Chr.1: 46673263-46673263 on Build GRCh38
rs890721100 Chr.1: 46673084-46673084 on Build GRCh38
rs929597193 Chr.1: 46673277-46673277 on Build GRCh38

View Full Product Details