Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86264616-86264865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3504933 c.583G>A p.D195N Chr.15: 86264699-86264699 on Build GRCh38
4528118 c.1511G>A p.R504Q Chr.15: 86264820-86264820 on Build GRCh38
3504937 c.662G>A p.W221* Chr.15: 86264778-86264778 on Build GRCh38
3816971 c.1364C>G p.T455R Chr.15: 86264673-86264673 on Build GRCh38
7081972 c.669C>A p.H223Q Chr.15: 86264785-86264785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766018404 Chr.15: 86264669-86264669 on Build GRCh38
rs201827530 Chr.15: 86264787-86264787 on Build GRCh38
rs139189895 Chr.15: 86264647-86264647 on Build GRCh38
rs11856833 Chr.15: 86264653-86264653 on Build GRCh38
rs964337675 Chr.15: 86264723-86264723 on Build GRCh38
rs758755951 Chr.15: 86264776-86264776 on Build GRCh38
rs746597393 Chr.15: 86264820-86264820 on Build GRCh38
rs765457924 Chr.15: 86264677-86264677 on Build GRCh38
rs778880442 Chr.15: 86264811-86264811 on Build GRCh38
rs767192869 Chr.15: 86264651-86264651 on Build GRCh38
rs765764602 Chr.15: 86264769-86264769 on Build GRCh38
rs749164915 Chr.15: 86264698-86264698 on Build GRCh38
rs373269237 Chr.15: 86264683-86264683 on Build GRCh38
rs770706712 Chr.15: 86264839-86264839 on Build GRCh38
rs199539728 Chr.15: 86264695-86264695 on Build GRCh38
rs753103295 Chr.15: 86264684-86264684 on Build GRCh38
rs1007294128 Chr.15: 86264795-86264795 on Build GRCh38
rs564644978 Chr.15: 86264786-86264786 on Build GRCh38
rs904561590 Chr.15: 86264789-86264789 on Build GRCh38
rs750932521 Chr.15: 86264810-86264810 on Build GRCh38
rs750093981 Chr.15: 86264652-86264652 on Build GRCh38
rs751727702 Chr.15: 86264784-86264784 on Build GRCh38
rs201541206 Chr.15: 86264699-86264699 on Build GRCh38
rs574460212 Chr.15: 86264693-86264693 on Build GRCh38
rs367677892 Chr.15: 86264752-86264752 on Build GRCh38
rs143113232 Chr.15: 86264812-86264812 on Build GRCh38
rs765874212 Chr.15: 86264741-86264742 on Build GRCh38
rs372186294 Chr.15: 86264754-86264754 on Build GRCh38
rs760427652 Chr.15: 86264746-86264746 on Build GRCh38
rs776289444 Chr.15: 86264760-86264760 on Build GRCh38
rs188199996 Chr.15: 86264782-86264782 on Build GRCh38
rs747955198 Chr.15: 86264724-86264724 on Build GRCh38
rs755559236 Chr.15: 86264671-86264671 on Build GRCh38
rs927174065 Chr.15: 86264829-86264829 on Build GRCh38
rs745941158 Chr.15: 86264790-86264790 on Build GRCh38
rs753190553 Chr.15: 86264774-86264774 on Build GRCh38
rs200425510 Chr.15: 86264685-86264685 on Build GRCh38
rs778030412 Chr.15: 86264687-86264687 on Build GRCh38
rs756204466 Chr.15: 86264810-86264810 on Build GRCh38
rs759097792 Chr.15: 86264846-86264846 on Build GRCh38
rs184053635 Chr.15: 86264739-86264739 on Build GRCh38
rs774220419 Chr.15: 86264717-86264717 on Build GRCh38
rs767849868 Chr.15: 86264773-86264773 on Build GRCh38
rs200845665 Chr.15: 86264762-86264762 on Build GRCh38
rs771850905 Chr.15: 86264818-86264818 on Build GRCh38
rs544094832 Chr.15: 86264819-86264819 on Build GRCh38
rs774810434 Chr.15: 86264704-86264704 on Build GRCh38

View Full Product Details