Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30758947-30759204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC189
Entrez Gene ID
90835
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93
UniGene
Hs.724010
Gene Chromosome Location
Chr.16: 30757226-30762277 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4981337 c.721G>C p.E241Q Chr.16: 30759184-30759184 on Build GRCh38
5660835 c.597G>A p.L199L Chr.16: 30759197-30759197 on Build GRCh38
5685242 c.694G>T p.E232* Chr.16: 30759024-30759024 on Build GRCh38
5901940 c.667G>A p.E223K Chr.16: 30759051-30759051 on Build GRCh38
5660834 c.708G>A p.L236L Chr.16: 30759197-30759197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775822711 Chr.16: 30759049-30759050 on Build GRCh38
rs750447347 Chr.16: 30759142-30759143 on Build GRCh38
rs371457610 Chr.16: 30758982-30758982 on Build GRCh38
rs368395391 Chr.16: 30759075-30759075 on Build GRCh38
rs749042317 Chr.16: 30759013-30759018 on Build GRCh38
rs910187950 Chr.16: 30758971-30758971 on Build GRCh38
rs750859134 Chr.16: 30759058-30759058 on Build GRCh38
rs141842194 Chr.16: 30759081-30759081 on Build GRCh38
rs780448832 Chr.16: 30759062-30759062 on Build GRCh38
rs538805878 Chr.16: 30759002-30759002 on Build GRCh38
rs561615712 Chr.16: 30758972-30758972 on Build GRCh38
rs533260900 Chr.16: 30759129-30759129 on Build GRCh38
rs553935133 Chr.16: 30759009-30759009 on Build GRCh38
rs770511048 Chr.16: 30759117-30759117 on Build GRCh38
rs759543408 Chr.16: 30759179-30759179 on Build GRCh38
rs765880267 Chr.16: 30759147-30759147 on Build GRCh38
rs759098573 Chr.16: 30759026-30759026 on Build GRCh38
rs765676123 Chr.16: 30759055-30759055 on Build GRCh38
rs764439368 Chr.16: 30759036-30759036 on Build GRCh38
rs755201655 Chr.16: 30759161-30759161 on Build GRCh38
rs916735133 Chr.16: 30759110-30759110 on Build GRCh38
rs776645687 Chr.16: 30758979-30758979 on Build GRCh38
rs61746942 Chr.16: 30759051-30759051 on Build GRCh38
rs762557343 Chr.16: 30758998-30758998 on Build GRCh38
rs748092461 Chr.16: 30759090-30759090 on Build GRCh38
rs187063168 Chr.16: 30759007-30759007 on Build GRCh38
rs774055979 Chr.16: 30759130-30759130 on Build GRCh38
rs373898885 Chr.16: 30759145-30759145 on Build GRCh38
rs751538068 Chr.16: 30759064-30759064 on Build GRCh38
rs777504706 Chr.16: 30759096-30759096 on Build GRCh38
rs930743006 Chr.16: 30759121-30759121 on Build GRCh38
rs551789158 Chr.16: 30759180-30759180 on Build GRCh38
rs947076521 Chr.16: 30759004-30759004 on Build GRCh38
rs368035673 Chr.16: 30759149-30759149 on Build GRCh38
rs374688295 Chr.16: 30759128-30759128 on Build GRCh38
rs750771634 Chr.16: 30759148-30759148 on Build GRCh38
rs755078275 Chr.16: 30759068-30759068 on Build GRCh38
rs769560917 Chr.16: 30758987-30758987 on Build GRCh38
rs144755146 Chr.16: 30759046-30759046 on Build GRCh38
rs761003739 Chr.16: 30759058-30759059 on Build GRCh38
rs368458049 Chr.16: 30759132-30759132 on Build GRCh38
rs749101763 Chr.16: 30759116-30759116 on Build GRCh38
rs773037049 Chr.16: 30758995-30758995 on Build GRCh38
rs371228244 Chr.16: 30759153-30759153 on Build GRCh38
rs774912276 Chr.16: 30759135-30759135 on Build GRCh38
rs565882864 Chr.16: 30759059-30759059 on Build GRCh38
rs967029072 Chr.16: 30759010-30759010 on Build GRCh38
rs1057347198 Chr.16: 30759089-30759089 on Build GRCh38
rs769758772 Chr.16: 30759091-30759091 on Build GRCh38
rs370922410 Chr.16: 30759134-30759134 on Build GRCh38
rs770878389 Chr.16: 30759015-30759016 on Build GRCh38

View Full Product Details