Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3498860-3499133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADI1
Entrez Gene ID
55256
Gene Name
acireductone dioxygenase 1
Gene Aliases
APL1, ARD, Fe-ARD, HMFT1638, MTCBP1, Ni-ARD, SIPL, mtnD
UniGene
Hs.502773
Gene Chromosome Location
Chr.2: 3497919-3519579 on Build GRCh38
Transcripts
NM_001306077.1 NM_018269.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
73594 c.430A>G p.K144E Chr.2: 3499073-3499073 on Build GRCh38
4094004 c.503G>A p.R168H Chr.2: 3499000-3499000 on Build GRCh38
6595230 c.502C>T p.R168C Chr.2: 3499001-3499001 on Build GRCh38
2943294 c.487G>A p.D163N Chr.2: 3499016-3499016 on Build GRCh38
4939110 c.525G>A p.L175L Chr.2: 3498978-3498978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748794526 Chr.2: 3499044-3499044 on Build GRCh38
rs143494796 Chr.2: 3499049-3499049 on Build GRCh38
rs986880427 Chr.2: 3498975-3498975 on Build GRCh38
rs777423115 Chr.2: 3498973-3498973 on Build GRCh38
rs755803454 Chr.2: 3499094-3499094 on Build GRCh38
rs771361700 Chr.2: 3498990-3498990 on Build GRCh38
rs146697305 Chr.2: 3499033-3499033 on Build GRCh38
rs781647971 Chr.2: 3498886-3498886 on Build GRCh38
rs1012162845 Chr.2: 3498932-3498932 on Build GRCh38
rs542113701 Chr.2: 3498893-3498893 on Build GRCh38
rs747698961 Chr.2: 3498918-3498918 on Build GRCh38
rs760815269 Chr.2: 3498946-3498946 on Build GRCh38
rs145825002 Chr.2: 3498919-3498919 on Build GRCh38
rs556650618 Chr.2: 3499026-3499026 on Build GRCh38
rs375578103 Chr.2: 3498970-3498970 on Build GRCh38
rs751335367 Chr.2: 3498965-3498965 on Build GRCh38
rs766324518 Chr.2: 3499074-3499074 on Build GRCh38
rs754878202 Chr.2: 3499024-3499024 on Build GRCh38
rs371879652 Chr.2: 3498913-3498913 on Build GRCh38
rs369579979 Chr.2: 3498933-3498933 on Build GRCh38
rs148975364 Chr.2: 3499063-3499063 on Build GRCh38
rs533234616 Chr.2: 3499004-3499004 on Build GRCh38
rs750318073 Chr.2: 3498955-3498955 on Build GRCh38
rs906416607 Chr.2: 3499111-3499111 on Build GRCh38
rs767376641 Chr.2: 3499084-3499084 on Build GRCh38
rs151206448 Chr.2: 3498966-3498966 on Build GRCh38
rs372843618 Chr.2: 3499025-3499025 on Build GRCh38
rs763691922 Chr.2: 3499106-3499106 on Build GRCh38
rs939083366 Chr.2: 3498909-3498909 on Build GRCh38
rs781308657 Chr.2: 3499070-3499070 on Build GRCh38
rs772279164 Chr.2: 3499000-3499000 on Build GRCh38
rs753897708 Chr.2: 3499019-3499019 on Build GRCh38
rs140822493 Chr.2: 3499022-3499022 on Build GRCh38
rs761864659 Chr.2: 3499069-3499069 on Build GRCh38
rs759550378 Chr.2: 3499076-3499076 on Build GRCh38
rs535892517 Chr.2: 3498959-3498959 on Build GRCh38
rs746234400 Chr.2: 3499055-3499055 on Build GRCh38
rs370738223 Chr.2: 3499064-3499064 on Build GRCh38
rs747648086 Chr.2: 3498969-3498969 on Build GRCh38
rs752558500 Chr.2: 3499091-3499091 on Build GRCh38
rs771440115 Chr.2: 3499045-3499045 on Build GRCh38
rs961401919 Chr.2: 3498968-3498968 on Build GRCh38
rs753387336 Chr.2: 3499110-3499110 on Build GRCh38
rs373506681 Chr.2: 3498940-3498940 on Build GRCh38
rs776752124 Chr.2: 3499067-3499067 on Build GRCh38
rs375338188 Chr.2: 3499065-3499065 on Build GRCh38
rs759752630 Chr.2: 3498939-3498939 on Build GRCh38
rs760904872 Chr.2: 3499002-3499002 on Build GRCh38
rs764093955 Chr.2: 3499014-3499014 on Build GRCh38
rs1003756035 Chr.2: 3498962-3498962 on Build GRCh38
rs10208494 Chr.2: 3498938-3498938 on Build GRCh38
rs749758704 Chr.2: 3498989-3498989 on Build GRCh38
rs748218756 Chr.2: 3499075-3499075 on Build GRCh38
rs752327437 Chr.2: 3499028-3499028 on Build GRCh38
rs774752301 Chr.2: 3498998-3498998 on Build GRCh38
rs765149986 Chr.2: 3498954-3498954 on Build GRCh38
rs200222805 Chr.2: 3499032-3499032 on Build GRCh38
rs752745822 Chr.2: 3498941-3498941 on Build GRCh38
rs775751643 Chr.2: 3499001-3499001 on Build GRCh38
rs567726498 Chr.2: 3499020-3499020 on Build GRCh38
rs763380271 Chr.2: 3498937-3498937 on Build GRCh38
rs144167517 Chr.2: 3498999-3498999 on Build GRCh38

View Full Product Details