Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3623604-3623863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACTIN
Entrez Gene ID
58509
Gene Name
cactin, spliceosome C complex subunit
Gene Aliases
C19orf29, NY-REN-24, fSAPc
UniGene
Hs.128425
Gene Chromosome Location
Chr.19: 3610628-3626815 on Build GRCh38
Transcripts
NM_001080543.1 NM_021231.1 XM_011528160.2 XM_011528161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5757859 c.572A>G p.Y191C Chr.19: 3623758-3623758 on Build GRCh38
995213 c.481G>A p.E161K Chr.19: 3623849-3623849 on Build GRCh38
4667154 c.528C>A p.A176A Chr.19: 3623802-3623802 on Build GRCh38
6314477 c.369C>A p.Y123* Chr.19: 3623757-3623757 on Build GRCh38
5683149 c.386C>T p.T129I Chr.19: 3623740-3623740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61740064 Chr.19: 3623787-3623787 on Build GRCh38
rs781706693 Chr.19: 3623781-3623781 on Build GRCh38
rs773035619 Chr.19: 3623669-3623669 on Build GRCh38
rs370366128 Chr.19: 3623784-3623784 on Build GRCh38
rs372820595 Chr.19: 3623739-3623739 on Build GRCh38
rs767422361 Chr.19: 3623679-3623679 on Build GRCh38
rs531232281 Chr.19: 3623676-3623676 on Build GRCh38
rs199984244 Chr.19: 3623796-3623796 on Build GRCh38
rs747519855 Chr.19: 3623752-3623752 on Build GRCh38
rs759027582 Chr.19: 3623805-3623805 on Build GRCh38
rs201335488 Chr.19: 3623683-3623683 on Build GRCh38
rs769002983 Chr.19: 3623757-3623757 on Build GRCh38
rs993066886 Chr.19: 3623627-3623627 on Build GRCh38
rs758487720 Chr.19: 3623742-3623742 on Build GRCh38
rs766510486 Chr.19: 3623844-3623844 on Build GRCh38
rs770714598 Chr.19: 3623791-3623791 on Build GRCh38
rs1005561090 Chr.19: 3623678-3623678 on Build GRCh38
rs771589130 Chr.19: 3623814-3623814 on Build GRCh38
rs757495702 Chr.19: 3623727-3623727 on Build GRCh38
rs779038820 Chr.19: 3623738-3623738 on Build GRCh38
rs917206842 Chr.19: 3623656-3623656 on Build GRCh38
rs769043155 Chr.19: 3623665-3623665 on Build GRCh38
rs753593309 Chr.19: 3623721-3623721 on Build GRCh38
rs377118932 Chr.19: 3623745-3623745 on Build GRCh38
rs763973263 Chr.19: 3623828-3623828 on Build GRCh38
rs760440624 Chr.19: 3623709-3623709 on Build GRCh38
rs890738741 Chr.19: 3623794-3623794 on Build GRCh38
rs137855632 Chr.19: 3623678-3623679 on Build GRCh38
rs1040092684 Chr.19: 3623673-3623673 on Build GRCh38
rs770188736 Chr.19: 3623681-3623681 on Build GRCh38
rs1052353733 Chr.19: 3623785-3623785 on Build GRCh38
rs747390179 Chr.19: 3623663-3623663 on Build GRCh38
rs780605213 Chr.19: 3623662-3623662 on Build GRCh38
rs191160617 Chr.19: 3623675-3623675 on Build GRCh38
rs369539603 Chr.19: 3623712-3623712 on Build GRCh38
rs951360848 Chr.19: 3623658-3623658 on Build GRCh38
rs752667656 Chr.19: 3623699-3623699 on Build GRCh38
rs775333735 Chr.19: 3623820-3623820 on Build GRCh38
rs187985455 Chr.19: 3623659-3623659 on Build GRCh38
rs760701708 Chr.19: 3623826-3623826 on Build GRCh38
rs371224074 Chr.19: 3623830-3623830 on Build GRCh38
rs758444035 Chr.19: 3623841-3623841 on Build GRCh38
rs763850291 Chr.19: 3623720-3623720 on Build GRCh38
rs761589675 Chr.19: 3623829-3623829 on Build GRCh38
rs558910599 Chr.19: 3623625-3623625 on Build GRCh38

View Full Product Details