Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29562489-29562750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD15
Entrez Gene ID
116236
Gene Name
abhydrolase domain containing 15
Gene Aliases
-
UniGene
Hs.106510
Gene Chromosome Location
Chr.17: 29560661-29567024 on Build GRCh38
Transcripts
NM_198147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464737 c.1307T>C p.L436P Chr.17: 29562661-29562661 on Build GRCh38
4830149 c.1406G>C p.*469S Chr.17: 29562562-29562562 on Build GRCh38
6464738 c.1306C>T p.L436F Chr.17: 29562662-29562662 on Build GRCh38
6523408 c.1271G>T p.R424M Chr.17: 29562697-29562697 on Build GRCh38
5102351 c.1262C>T p.T421M Chr.17: 29562706-29562706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542605305 Chr.17: 29562558-29562558 on Build GRCh38
rs148129691 Chr.17: 29562626-29562626 on Build GRCh38
rs755683613 Chr.17: 29562570-29562570 on Build GRCh38
rs752691493 Chr.17: 29562663-29562663 on Build GRCh38
rs377676039 Chr.17: 29562578-29562578 on Build GRCh38
rs747952644 Chr.17: 29562525-29562525 on Build GRCh38
rs770873228 Chr.17: 29562549-29562549 on Build GRCh38
rs954062938 Chr.17: 29562666-29562666 on Build GRCh38
rs1053751493 Chr.17: 29562709-29562709 on Build GRCh38
rs750012759 Chr.17: 29562565-29562565 on Build GRCh38
rs776106528 Chr.17: 29562710-29562710 on Build GRCh38
rs201718724 Chr.17: 29562584-29562584 on Build GRCh38
rs762914048 Chr.17: 29562669-29562669 on Build GRCh38
rs764090375 Chr.17: 29562675-29562675 on Build GRCh38
rs62070804 Chr.17: 29562625-29562625 on Build GRCh38
rs759144529 Chr.17: 29562658-29562658 on Build GRCh38
rs374615193 Chr.17: 29562715-29562715 on Build GRCh38
rs745946949 Chr.17: 29562604-29562604 on Build GRCh38
rs146551867 Chr.17: 29562659-29562659 on Build GRCh38
rs866479542 Chr.17: 29562704-29562704 on Build GRCh38
rs748501993 Chr.17: 29562728-29562728 on Build GRCh38
rs747148886 Chr.17: 29562594-29562594 on Build GRCh38
rs758232715 Chr.17: 29562590-29562590 on Build GRCh38
rs142986550 Chr.17: 29562705-29562705 on Build GRCh38
rs267604787 Chr.17: 29562650-29562650 on Build GRCh38
rs751372234 Chr.17: 29562678-29562678 on Build GRCh38
rs200330374 Chr.17: 29562637-29562637 on Build GRCh38
rs747262799 Chr.17: 29562644-29562644 on Build GRCh38
rs372691848 Chr.17: 29562710-29562710 on Build GRCh38
rs201924599 Chr.17: 29562653-29562653 on Build GRCh38
rs925906472 Chr.17: 29562697-29562697 on Build GRCh38
rs528380562 Chr.17: 29562577-29562577 on Build GRCh38
rs199677056 Chr.17: 29562648-29562648 on Build GRCh38
rs748004076 Chr.17: 29562585-29562585 on Build GRCh38
rs753345969 Chr.17: 29562631-29562631 on Build GRCh38
rs775958133 Chr.17: 29562610-29562610 on Build GRCh38
rs770347594 Chr.17: 29562605-29562605 on Build GRCh38
rs139069619 Chr.17: 29562682-29562682 on Build GRCh38
rs773280096 Chr.17: 29562543-29562543 on Build GRCh38
rs778083462 Chr.17: 29562593-29562593 on Build GRCh38
rs768584286 Chr.17: 29562514-29562514 on Build GRCh38
rs768376797 Chr.17: 29562639-29562640 on Build GRCh38
rs138073270 Chr.17: 29562651-29562651 on Build GRCh38
rs371401765 Chr.17: 29562706-29562706 on Build GRCh38
rs776511533 Chr.17: 29562599-29562599 on Build GRCh38
rs780053237 Chr.17: 29562711-29562711 on Build GRCh38
rs530464744 Chr.17: 29562517-29562517 on Build GRCh38
rs769099232 Chr.17: 29562714-29562714 on Build GRCh38
rs746445636 Chr.17: 29562566-29562567 on Build GRCh38
rs749364478 Chr.17: 29562713-29562713 on Build GRCh38
rs764326975 Chr.17: 29562613-29562613 on Build GRCh38
rs367567626 Chr.17: 29562652-29562652 on Build GRCh38
rs781467147 Chr.17: 29562646-29562646 on Build GRCh38
rs150816352 Chr.17: 29562730-29562730 on Build GRCh38
rs754514801 Chr.17: 29562633-29562633 on Build GRCh38
rs895939426 Chr.17: 29562567-29562567 on Build GRCh38
rs777371076 Chr.17: 29562643-29562643 on Build GRCh38
rs746001203 Chr.17: 29562647-29562647 on Build GRCh38
rs772479385 Chr.17: 29562526-29562526 on Build GRCh38
rs375930056 Chr.17: 29562544-29562544 on Build GRCh38
rs777214610 Chr.17: 29562556-29562556 on Build GRCh38
rs766049769 Chr.17: 29562627-29562627 on Build GRCh38
rs763348862 Chr.17: 29562612-29562612 on Build GRCh38
rs201730687 Chr.17: 29562649-29562649 on Build GRCh38
rs763059674 Chr.17: 29562602-29562602 on Build GRCh38
rs539307 Chr.17: 29562560-29562560 on Build GRCh38

View Full Product Details