Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95202008-95202243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM6
Entrez Gene ID
142827
Gene Name
acyl-CoA synthetase medium-chain family member 6
Gene Aliases
C10orf129, bA310E22.3
UniGene
Hs.134229
Gene Chromosome Location
Chr.10: 95194200-95228929 on Build GRCh38
Transcripts
NM_207321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229169 c.225G>A p.W75* Chr.10: 95202017-95202017 on Build GRCh38
7347964 c.295G>A p.A99T Chr.10: 95202087-95202087 on Build GRCh38
5989389 c.345G>T p.M115I Chr.10: 95202137-95202137 on Build GRCh38
7357932 c.228G>C p.K76N Chr.10: 95202020-95202020 on Build GRCh38
1970968 c.222C>T p.L74L Chr.10: 95202014-95202014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376416951 Chr.10: 95202211-95202211 on Build GRCh38
rs772462551 Chr.10: 95202057-95202057 on Build GRCh38
rs138613844 Chr.10: 95202212-95202212 on Build GRCh38
rs754035653 Chr.10: 95202220-95202221 on Build GRCh38
rs371778473 Chr.10: 95202152-95202152 on Build GRCh38
rs185879196 Chr.10: 95202130-95202130 on Build GRCh38
rs994791452 Chr.10: 95202158-95202158 on Build GRCh38
rs201108997 Chr.10: 95202037-95202037 on Build GRCh38
rs774092318 Chr.10: 95202167-95202167 on Build GRCh38
rs776587083 Chr.10: 95202044-95202044 on Build GRCh38
rs777627862 Chr.10: 95202139-95202139 on Build GRCh38
rs775034434 Chr.10: 95202221-95202221 on Build GRCh38
rs200202492 Chr.10: 95202148-95202148 on Build GRCh38
rs909823672 Chr.10: 95202034-95202034 on Build GRCh38
rs745542005 Chr.10: 95202190-95202190 on Build GRCh38
rs897652019 Chr.10: 95202154-95202154 on Build GRCh38
rs751791076 Chr.10: 95202091-95202091 on Build GRCh38
rs891189893 Chr.10: 95202044-95202044 on Build GRCh38
rs781280614 Chr.10: 95202107-95202107 on Build GRCh38
rs1036232743 Chr.10: 95202143-95202143 on Build GRCh38
rs748213716 Chr.10: 95202129-95202129 on Build GRCh38
rs377737073 Chr.10: 95202142-95202142 on Build GRCh38
rs755001167 Chr.10: 95202098-95202098 on Build GRCh38
rs900915376 Chr.10: 95202115-95202115 on Build GRCh38
rs201176783 Chr.10: 95202189-95202189 on Build GRCh38
rs774208243 Chr.10: 95202086-95202086 on Build GRCh38
rs939652838 Chr.10: 95202084-95202084 on Build GRCh38
rs780181629 Chr.10: 95202087-95202087 on Build GRCh38
rs933756545 Chr.10: 95202117-95202117 on Build GRCh38

View Full Product Details